Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91278864-91279403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88C
Entrez Gene ID
440193
Gene Name
coiled-coil domain containing 88C
Gene Aliases
DAPLE, HKRP2, KIAA1509, SCA40
UniGene
Hs.525536
Gene Chromosome Location
Chr.14: 91271323-91418498 on Build GRCh38
Transcripts
NM_001080414.3 XM_011536796.2 XM_017021335.1 XM_017021336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5924612 c.4700-3C>T p.? Chr.14: 91279309-91279309 on Build GRCh38
5946984 c.4768+1G>T p.? Chr.14: 91279237-91279237 on Build GRCh38
115963 c.4706G>A p.R1569Q Chr.14: 91279300-91279300 on Build GRCh38
3815498 c.4751C>T p.S1584L Chr.14: 91279255-91279255 on Build GRCh38
6457552 c.318C>T p.N106N Chr.14: 91279260-91279260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775761174 Chr.14: 91279226-91279226 on Build GRCh38
rs75284036 Chr.14: 91278917-91278917 on Build GRCh38
rs371969147 Chr.14: 91279302-91279302 on Build GRCh38
rs900981500 Chr.14: 91279297-91279297 on Build GRCh38
rs185405074 Chr.14: 91279001-91279001 on Build GRCh38
rs534486317 Chr.14: 91278998-91278998 on Build GRCh38
rs770658482 Chr.14: 91279327-91279327 on Build GRCh38
rs531829211 Chr.14: 91279352-91279352 on Build GRCh38
rs930653935 Chr.14: 91279214-91279214 on Build GRCh38
rs917389389 Chr.14: 91279059-91279059 on Build GRCh38
rs918722562 Chr.14: 91279347-91279347 on Build GRCh38
rs912810554 Chr.14: 91279300-91279300 on Build GRCh38
rs753727388 Chr.14: 91278889-91278890 on Build GRCh38
rs869116104 Chr.14: 91278890-91278904 on Build GRCh38
rs386382145 Chr.14: 91278890-91278891 on Build GRCh38
rs544395625 Chr.14: 91279301-91279301 on Build GRCh38
rs968372200 Chr.14: 91279010-91279010 on Build GRCh38
rs866007086 Chr.14: 91278916-91278916 on Build GRCh38
rs763665401 Chr.14: 91279268-91279268 on Build GRCh38
rs781648642 Chr.14: 91279295-91279295 on Build GRCh38
rs372725492 Chr.14: 91279298-91279298 on Build GRCh38
rs774806113 Chr.14: 91279058-91279058 on Build GRCh38
rs972250667 Chr.14: 91279135-91279135 on Build GRCh38
rs760861922 Chr.14: 91279250-91279250 on Build GRCh38
rs769791630 Chr.14: 91279334-91279334 on Build GRCh38
rs1032316567 Chr.14: 91279292-91279292 on Build GRCh38
rs985646682 Chr.14: 91279047-91279047 on Build GRCh38
rs773302314 Chr.14: 91279211-91279211 on Build GRCh38
rs765141591 Chr.14: 91279257-91279257 on Build GRCh38
rs978924527 Chr.14: 91279049-91279049 on Build GRCh38
rs918323537 Chr.14: 91279087-91279087 on Build GRCh38
rs781551975 Chr.14: 91279199-91279199 on Build GRCh38
rs776581160 Chr.14: 91279255-91279255 on Build GRCh38
rs766131713 Chr.14: 91279253-91279253 on Build GRCh38
rs1048092825 Chr.14: 91279101-91279101 on Build GRCh38
rs147375675 Chr.14: 91279051-91279051 on Build GRCh38
rs776462903 Chr.14: 91279329-91279329 on Build GRCh38
rs6575186 Chr.14: 91279000-91279000 on Build GRCh38
rs958497284 Chr.14: 91279349-91279349 on Build GRCh38
rs377035641 Chr.14: 91279235-91279235 on Build GRCh38
rs558759958 Chr.14: 91279096-91279096 on Build GRCh38
rs572045603 Chr.14: 91279249-91279249 on Build GRCh38
rs367891084 Chr.14: 91279191-91279191 on Build GRCh38
rs762138919 Chr.14: 91279061-91279061 on Build GRCh38
rs757924740 Chr.14: 91279267-91279267 on Build GRCh38
rs752247270 Chr.14: 91279260-91279260 on Build GRCh38
rs10671669 Chr.14: 91278916-91278917 on Build GRCh38
rs752668368 Chr.14: 91279303-91279303 on Build GRCh38
rs759425025 Chr.14: 91279256-91279256 on Build GRCh38
rs927062430 Chr.14: 91279050-91279050 on Build GRCh38
rs540907763 Chr.14: 91279264-91279264 on Build GRCh38
rs1040910288 Chr.14: 91279252-91279252 on Build GRCh38
rs1050982591 Chr.14: 91279216-91279216 on Build GRCh38
rs895691884 Chr.14: 91279183-91279183 on Build GRCh38
rs965275658 Chr.14: 91278956-91278956 on Build GRCh38
rs150065113 Chr.14: 91278921-91278921 on Build GRCh38
rs776918682 Chr.14: 91279273-91279273 on Build GRCh38
rs149283102 Chr.14: 91278923-91278923 on Build GRCh38
rs931073100 Chr.14: 91279377-91279377 on Build GRCh38
rs774441216 Chr.14: 91279318-91279318 on Build GRCh38
rs1001481604 Chr.14: 91279159-91279159 on Build GRCh38
rs538304756 Chr.14: 91279141-91279141 on Build GRCh38
rs539265889 Chr.14: 91279060-91279060 on Build GRCh38
rs183312399 Chr.14: 91279232-91279232 on Build GRCh38
rs1019566308 Chr.14: 91278941-91278941 on Build GRCh38
rs190066438 Chr.14: 91279124-91279124 on Build GRCh38
rs920604950 Chr.14: 91279157-91279157 on Build GRCh38
rs139697985 Chr.14: 91279137-91279137 on Build GRCh38
rs748362063 Chr.14: 91279324-91279324 on Build GRCh38
rs759298529 Chr.14: 91279330-91279330 on Build GRCh38
rs979851856 Chr.14: 91278992-91278992 on Build GRCh38
rs554362051 Chr.14: 91279281-91279281 on Build GRCh38
rs768825207 Chr.14: 91279240-91279240 on Build GRCh38
rs145210051 Chr.14: 91279299-91279299 on Build GRCh38
rs1057053775 Chr.14: 91279160-91279160 on Build GRCh38
rs1034544775 Chr.14: 91279328-91279328 on Build GRCh38
rs570506095 Chr.14: 91279002-91279002 on Build GRCh38
rs770016495 Chr.14: 91279221-91279221 on Build GRCh38
rs966771126 Chr.14: 91278913-91278913 on Build GRCh38
rs766898543 Chr.14: 91278961-91278961 on Build GRCh38

View Full Product Details