Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91942500-91943022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN5
Entrez Gene ID
10516
Gene Name
fibulin 5
Gene Aliases
ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, HNARMD, UP50
UniGene
Hs.332708
Gene Chromosome Location
Chr.14: 91869411-91947702 on Build GRCh38
Transcripts
NM_006329.3 XM_005267267.3 XM_011536356.1 XM_011536357.1 XM_011536358.1 XM_017020929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6214940 c.72+5G>C p.? Chr.14: 91942902-91942902 on Build GRCh38
6425161 c.62G>A p.G21E Chr.14: 91942917-91942917 on Build GRCh38
4959658 c.59C>T p.P20L Chr.14: 91942920-91942920 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs60976221 Chr.14: 91942560-91942560 on Build GRCh38
rs570162560 Chr.14: 91942851-91942851 on Build GRCh38
rs778999697 Chr.14: 91942918-91942918 on Build GRCh38
rs113167395 Chr.14: 91942881-91942881 on Build GRCh38
rs772595054 Chr.14: 91942972-91942972 on Build GRCh38
rs889455462 Chr.14: 91942578-91942578 on Build GRCh38
rs1044027539 Chr.14: 91942661-91942661 on Build GRCh38
rs139320827 Chr.14: 91942599-91942599 on Build GRCh38
rs973948359 Chr.14: 91942837-91942837 on Build GRCh38
rs1042669976 Chr.14: 91942791-91942791 on Build GRCh38
rs950994101 Chr.14: 91942933-91942933 on Build GRCh38
rs200525690 Chr.14: 91942809-91942809 on Build GRCh38
rs914056629 Chr.14: 91942957-91942957 on Build GRCh38
rs1033844540 Chr.14: 91942763-91942763 on Build GRCh38
rs920577337 Chr.14: 91942589-91942589 on Build GRCh38
rs565761187 Chr.14: 91942882-91942882 on Build GRCh38
rs150715038 Chr.14: 91942902-91942902 on Build GRCh38
rs930689741 Chr.14: 91942593-91942593 on Build GRCh38
rs944787790 Chr.14: 91942616-91942616 on Build GRCh38
rs531808729 Chr.14: 91942610-91942610 on Build GRCh38
rs945558917 Chr.14: 91942974-91942974 on Build GRCh38
rs371468999 Chr.14: 91942861-91942861 on Build GRCh38
rs960815290 Chr.14: 91942523-91942523 on Build GRCh38
rs1026941629 Chr.14: 91942932-91942932 on Build GRCh38
rs1040501479 Chr.14: 91942889-91942889 on Build GRCh38
rs949350528 Chr.14: 91942767-91942767 on Build GRCh38
rs771672858 Chr.14: 91942969-91942969 on Build GRCh38
rs986108144 Chr.14: 91942843-91942843 on Build GRCh38
rs770470522 Chr.14: 91942992-91942992 on Build GRCh38
rs910674129 Chr.14: 91942600-91942600 on Build GRCh38
rs1009799961 Chr.14: 91942648-91942648 on Build GRCh38
rs569683263 Chr.14: 91942916-91942916 on Build GRCh38
rs748040891 Chr.14: 91942925-91942925 on Build GRCh38
rs1015356231 Chr.14: 91942814-91942814 on Build GRCh38
rs561218063 Chr.14: 91942833-91942833 on Build GRCh38
rs973977947 Chr.14: 91942586-91942586 on Build GRCh38
rs1016427895 Chr.14: 91942984-91942984 on Build GRCh38
rs950911685 Chr.14: 91942625-91942625 on Build GRCh38
rs990847246 Chr.14: 91942711-91942711 on Build GRCh38
rs184502226 Chr.14: 91942838-91942838 on Build GRCh38
rs903505985 Chr.14: 91942717-91942717 on Build GRCh38
rs751863037 Chr.14: 91942872-91942872 on Build GRCh38
rs8014548 Chr.14: 91942759-91942759 on Build GRCh38
rs1040528880 Chr.14: 91942633-91942633 on Build GRCh38
rs961216486 Chr.14: 91942832-91942832 on Build GRCh38
rs532141720 Chr.14: 91942841-91942841 on Build GRCh38
rs960449743 Chr.14: 91942789-91942789 on Build GRCh38
rs545611868 Chr.14: 91942622-91942622 on Build GRCh38
rs191229827 Chr.14: 91942645-91942645 on Build GRCh38
rs17127768 Chr.14: 91942989-91942989 on Build GRCh38
rs966350140 Chr.14: 91942581-91942581 on Build GRCh38
rs758388867 Chr.14: 91942579-91942579 on Build GRCh38
rs992046521 Chr.14: 91942546-91942546 on Build GRCh38
rs904145472 Chr.14: 91942991-91942991 on Build GRCh38
rs147327466 Chr.14: 91942555-91942555 on Build GRCh38
rs566317946 Chr.14: 91942522-91942522 on Build GRCh38
rs766688945 Chr.14: 91942873-91942873 on Build GRCh38
rs749068976 Chr.14: 91942900-91942900 on Build GRCh38
rs892653916 Chr.14: 91942634-91942634 on Build GRCh38

View Full Product Details