Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103104624-103105155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L4
Entrez Gene ID
91828
Gene Name
exocyst complex component 3 like 4
Gene Aliases
C14orf73
UniGene
Hs.37712
Gene Chromosome Location
Chr.14: 103094202-103110562 on Build GRCh38
Transcripts
NM_001077594.1 XM_011537323.2 XM_011537324.2 XM_011537325.2 XM_011537327.2 XM_011537328.2 XM_011537329.2 XM_011537330.2 XM_011537331.1 XM_011537332.2 XM_011537333.2 XM_011537334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3377343 c.1326G>A p.A442A Chr.14: 103104779-103104779 on Build GRCh38
7255295 c.1466G>A p.R489K Chr.14: 103105072-103105072 on Build GRCh38
2025356 c.1396G>A p.A466T Chr.14: 103105002-103105002 on Build GRCh38
2150013 c.1422C>G p.S474R Chr.14: 103105028-103105028 on Build GRCh38
953613 c.1365G>T p.A455A Chr.14: 103104818-103104818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372795518 Chr.14: 103105058-103105058 on Build GRCh38
rs753573132 Chr.14: 103105108-103105128 on Build GRCh38
rs1020373648 Chr.14: 103104802-103104802 on Build GRCh38
rs372989603 Chr.14: 103104982-103104982 on Build GRCh38
rs142688553 Chr.14: 103105055-103105055 on Build GRCh38
rs189534180 Chr.14: 103104691-103104691 on Build GRCh38
rs975947584 Chr.14: 103104740-103104740 on Build GRCh38
rs776619848 Chr.14: 103104722-103104722 on Build GRCh38
rs959000508 Chr.14: 103104929-103104929 on Build GRCh38
rs780944899 Chr.14: 103104970-103104970 on Build GRCh38
rs746671849 Chr.14: 103104960-103104960 on Build GRCh38
rs767645409 Chr.14: 103104828-103104828 on Build GRCh38
rs772188063 Chr.14: 103105016-103105016 on Build GRCh38
rs944634621 Chr.14: 103104725-103104725 on Build GRCh38
rs763498373 Chr.14: 103105117-103105117 on Build GRCh38
rs968745964 Chr.14: 103104823-103104823 on Build GRCh38
rs919157016 Chr.14: 103104772-103104772 on Build GRCh38
rs954263516 Chr.14: 103104711-103104711 on Build GRCh38
rs772091584 Chr.14: 103105085-103105085 on Build GRCh38
rs749235337 Chr.14: 103105077-103105077 on Build GRCh38
rs759986341 Chr.14: 103105100-103105100 on Build GRCh38
rs907334327 Chr.14: 103105013-103105013 on Build GRCh38
rs930301380 Chr.14: 103104668-103104668 on Build GRCh38
rs747016106 Chr.14: 103105086-103105086 on Build GRCh38
rs200932924 Chr.14: 103104948-103104948 on Build GRCh38
rs770695781 Chr.14: 103104696-103104696 on Build GRCh38
rs780030946 Chr.14: 103105074-103105074 on Build GRCh38
rs1040655241 Chr.14: 103105129-103105129 on Build GRCh38
rs773363855 Chr.14: 103105020-103105020 on Build GRCh38
rs760508121 Chr.14: 103105049-103105050 on Build GRCh38
rs776932247 Chr.14: 103105092-103105092 on Build GRCh38
rs754442933 Chr.14: 103104851-103104851 on Build GRCh38
rs962486528 Chr.14: 103105012-103105012 on Build GRCh38
rs757060596 Chr.14: 103104964-103104964 on Build GRCh38
rs769756488 Chr.14: 103104979-103104979 on Build GRCh38
rs773417318 Chr.14: 103104830-103104830 on Build GRCh38
rs368534894 Chr.14: 103104971-103104971 on Build GRCh38
rs985746389 Chr.14: 103105110-103105110 on Build GRCh38
rs545016554 Chr.14: 103104856-103104856 on Build GRCh38
rs558942943 Chr.14: 103104945-103104945 on Build GRCh38
rs775359162 Chr.14: 103104980-103104980 on Build GRCh38
rs760925230 Chr.14: 103105022-103105022 on Build GRCh38
rs1010328086 Chr.14: 103104779-103104779 on Build GRCh38
rs761010320 Chr.14: 103104847-103104847 on Build GRCh38
rs367554801 Chr.14: 103104777-103104777 on Build GRCh38
rs775922626 Chr.14: 103105112-103105112 on Build GRCh38
rs910181114 Chr.14: 103105116-103105116 on Build GRCh38
rs956002529 Chr.14: 103104685-103104685 on Build GRCh38
rs763991823 Chr.14: 103105062-103105064 on Build GRCh38
rs1034630581 Chr.14: 103104922-103104922 on Build GRCh38
rs774384088 Chr.14: 103104800-103104800 on Build GRCh38
rs371693526 Chr.14: 103104882-103104882 on Build GRCh38
rs765856203 Chr.14: 103105037-103105037 on Build GRCh38
rs1053391683 Chr.14: 103105065-103105065 on Build GRCh38
rs765794977 Chr.14: 103104890-103104890 on Build GRCh38
rs1049197387 Chr.14: 103104913-103104913 on Build GRCh38
rs767551531 Chr.14: 103104909-103104909 on Build GRCh38
rs920266500 Chr.14: 103104666-103104666 on Build GRCh38
rs139438860 Chr.14: 103105033-103105033 on Build GRCh38
rs1050340673 Chr.14: 103104683-103104683 on Build GRCh38
rs777299619 Chr.14: 103104951-103104951 on Build GRCh38
rs564794915 Chr.14: 103105118-103105120 on Build GRCh38
rs762441291 Chr.14: 103105039-103105039 on Build GRCh38
rs780785493 Chr.14: 103105052-103105052 on Build GRCh38
rs1021742549 Chr.14: 103104694-103104694 on Build GRCh38
rs761913390 Chr.14: 103104814-103104814 on Build GRCh38
rs755838650 Chr.14: 103105070-103105070 on Build GRCh38
rs756872735 Chr.14: 103105049-103105049 on Build GRCh38
rs759928849 Chr.14: 103105035-103105035 on Build GRCh38
rs753184704 Chr.14: 103105038-103105038 on Build GRCh38
rs544623067 Chr.14: 103105088-103105088 on Build GRCh38
rs71470234 Chr.14: 103105072-103105073 on Build GRCh38
rs763649884 Chr.14: 103105045-103105045 on Build GRCh38
rs770281293 Chr.14: 103105105-103105105 on Build GRCh38
rs1003145946 Chr.14: 103105031-103105031 on Build GRCh38
rs112803052 Chr.14: 103105056-103105056 on Build GRCh38
rs200219064 Chr.14: 103105028-103105028 on Build GRCh38
rs1040219419 Chr.14: 103104738-103104738 on Build GRCh38
rs1011620742 Chr.14: 103105131-103105131 on Build GRCh38
rs755038398 Chr.14: 103104660-103104660 on Build GRCh38
rs892271069 Chr.14: 103104752-103104752 on Build GRCh38
rs377413179 Chr.14: 103104648-103104668 on Build GRCh38
rs986021363 Chr.14: 103104719-103104719 on Build GRCh38
rs865946883 Chr.14: 103105024-103105028 on Build GRCh38
rs768774941 Chr.14: 103104799-103104799 on Build GRCh38
rs751220987 Chr.14: 103105048-103105048 on Build GRCh38
rs1031690885 Chr.14: 103104681-103104681 on Build GRCh38

View Full Product Details