Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105248442-105248933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRF1
Entrez Gene ID
2972
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Gene Aliases
BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
UniGene
Hs.424484
Gene Chromosome Location
Chr.14: 105209286-105315577 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 XM_005267561.3 XM_005267563.3 XM_006720123.2 XM_011536672.2 XM_011536673.1 XM_011536674.1 NM_033271.2 XM_005268188.3

Gene Symbol
BTBD6
Entrez Gene ID
90135
Gene Name
BTB domain containing 6
Gene Aliases
BDPL
UniGene
Hs.7367
Gene Chromosome Location
Chr.14: 105248542-105251093 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 XM_005267561.3 XM_005267563.3 XM_006720123.2 XM_011536672.2 XM_011536673.1 XM_011536674.1 NM_033271.2 XM_005268188.3

SNP Detail

SNP ID Chromosome Location
rs961776784 Chr.14: 105248859-105248859 on Build GRCh38
rs1008951570 Chr.14: 105248606-105248606 on Build GRCh38
rs1050540143 Chr.14: 105248598-105248598 on Build GRCh38
rs944790430 Chr.14: 105248777-105248777 on Build GRCh38
rs1050487656 Chr.14: 105248482-105248482 on Build GRCh38
rs931351894 Chr.14: 105248579-105248590 on Build GRCh38
rs962002808 Chr.14: 105248689-105248689 on Build GRCh38
rs918044858 Chr.14: 105248578-105248578 on Build GRCh38
rs1034165692 Chr.14: 105248826-105248826 on Build GRCh38
rs904449101 Chr.14: 105248815-105248815 on Build GRCh38
rs986357143 Chr.14: 105248909-105248909 on Build GRCh38
rs973481427 Chr.14: 105248877-105248877 on Build GRCh38
rs890495604 Chr.14: 105248501-105248501 on Build GRCh38
rs968937220 Chr.14: 105248567-105248567 on Build GRCh38
rs992514821 Chr.14: 105248659-105248659 on Build GRCh38
rs587646414 Chr.14: 105248608-105248608 on Build GRCh38
rs1052499461 Chr.14: 105248785-105248785 on Build GRCh38
rs899307475 Chr.14: 105248474-105248474 on Build GRCh38
rs1010176371 Chr.14: 105248616-105248616 on Build GRCh38
rs1027288092 Chr.14: 105248887-105248887 on Build GRCh38
rs587742730 Chr.14: 105248511-105248511 on Build GRCh38
rs1021945949 Chr.14: 105248618-105248618 on Build GRCh38
rs920803248 Chr.14: 105248593-105248593 on Build GRCh38
rs587685764 Chr.14: 105248510-105248510 on Build GRCh38
rs1048482817 Chr.14: 105248604-105248613 on Build GRCh38
rs1013989461 Chr.14: 105248840-105248840 on Build GRCh38
rs868300982 Chr.14: 105248625-105248625 on Build GRCh38
rs912034649 Chr.14: 105248771-105248771 on Build GRCh38
rs1034114754 Chr.14: 105248636-105248636 on Build GRCh38
rs587697366 Chr.14: 105248708-105248708 on Build GRCh38
rs773705469 Chr.14: 105248573-105248574 on Build GRCh38
rs1052446796 Chr.14: 105248610-105248610 on Build GRCh38
rs973715024 Chr.14: 105248709-105248709 on Build GRCh38
rs1001319076 Chr.14: 105248632-105248632 on Build GRCh38
rs587616123 Chr.14: 105248526-105248526 on Build GRCh38
rs971199874 Chr.14: 105248662-105248662 on Build GRCh38
rs587676941 Chr.14: 105248900-105248900 on Build GRCh38
rs892436831 Chr.14: 105248612-105248612 on Build GRCh38
rs779801718 Chr.14: 105248564-105248564 on Build GRCh38
rs932182269 Chr.14: 105248742-105248742 on Build GRCh38
rs940973109 Chr.14: 105248584-105248584 on Build GRCh38
rs587622684 Chr.14: 105248475-105248475 on Build GRCh38
rs186857534 Chr.14: 105248833-105248833 on Build GRCh38
rs587760631 Chr.14: 105248570-105248573 on Build GRCh38
rs904388149 Chr.14: 105248628-105248628 on Build GRCh38
rs986306691 Chr.14: 105248759-105248759 on Build GRCh38
rs1038341860 Chr.14: 105248591-105248591 on Build GRCh38
rs927035755 Chr.14: 105248576-105248576 on Build GRCh38
rs953623186 Chr.14: 105248898-105248898 on Build GRCh38
rs867040308 Chr.14: 105248651-105248651 on Build GRCh38
rs1043016107 Chr.14: 105248812-105248818 on Build GRCh38
rs892494588 Chr.14: 105248801-105248801 on Build GRCh38
rs771660280 Chr.14: 105248518-105248518 on Build GRCh38
rs955989799 Chr.14: 105248539-105248539 on Build GRCh38
rs1014616687 Chr.14: 105248682-105248682 on Build GRCh38
rs920843323 Chr.14: 105248711-105248711 on Build GRCh38
rs979848042 Chr.14: 105248571-105248571 on Build GRCh38
rs587727144 Chr.14: 105248528-105248529 on Build GRCh38
rs867422238 Chr.14: 105248675-105248675 on Build GRCh38
rs946697726 Chr.14: 105248802-105248802 on Build GRCh38
rs932135045 Chr.14: 105248596-105248596 on Build GRCh38
rs113720774 Chr.14: 105248699-105248699 on Build GRCh38
rs1015080509 Chr.14: 105248857-105248857 on Build GRCh38
rs1001380786 Chr.14: 105248824-105248824 on Build GRCh38
rs1030967221 Chr.14: 105248521-105248521 on Build GRCh38
rs959745489 Chr.14: 105248577-105248577 on Build GRCh38
rs981325718 Chr.14: 105248677-105248677 on Build GRCh38
rs894667193 Chr.14: 105248556-105248571 on Build GRCh38
rs587744685 Chr.14: 105248574-105248574 on Build GRCh38
rs150454083 Chr.14: 105248550-105248550 on Build GRCh38

View Full Product Details