Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 35520953-35521467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH6
Entrez Gene ID
89978
Gene Name
diphthamine biosynthesis 6
Gene Aliases
ATPBD4
UniGene
Hs.107196
Gene Chromosome Location
Chr.15: 35217356-35546222 on Build GRCh38
Transcripts
NM_001141972.1 NM_080650.3 XM_011522160.1 XM_017022707.1 XM_017022708.1 XM_017022709.1

SNP Detail

SNP ID Chromosome Location
rs115574079 Chr.15: 35521406-35521406 on Build GRCh38
rs981823 Chr.15: 35521328-35521328 on Build GRCh38
rs542701312 Chr.15: 35521147-35521147 on Build GRCh38
rs930605110 Chr.15: 35521163-35521163 on Build GRCh38
rs1028779193 Chr.15: 35521254-35521254 on Build GRCh38
rs986130029 Chr.15: 35521397-35521397 on Build GRCh38
rs571392944 Chr.15: 35520994-35520994 on Build GRCh38
rs10520012 Chr.15: 35521177-35521177 on Build GRCh38
rs999046078 Chr.15: 35521022-35521022 on Build GRCh38
rs910770215 Chr.15: 35521183-35521183 on Build GRCh38
rs538470949 Chr.15: 35521029-35521029 on Build GRCh38
rs754988556 Chr.15: 35521028-35521028 on Build GRCh38
rs151327896 Chr.15: 35521276-35521276 on Build GRCh38
rs1040602444 Chr.15: 35521004-35521004 on Build GRCh38
rs952113862 Chr.15: 35521375-35521375 on Build GRCh38
rs998598412 Chr.15: 35521272-35521272 on Build GRCh38
rs1002778971 Chr.15: 35521339-35521339 on Build GRCh38
rs181161522 Chr.15: 35521039-35521039 on Build GRCh38
rs887373092 Chr.15: 35521168-35521168 on Build GRCh38
rs538869645 Chr.15: 35521024-35521024 on Build GRCh38
rs1050686398 Chr.15: 35520981-35520981 on Build GRCh38
rs571591354 Chr.15: 35521376-35521376 on Build GRCh38
rs577926979 Chr.15: 35521019-35521019 on Build GRCh38
rs938771779 Chr.15: 35521043-35521043 on Build GRCh38
rs189842878 Chr.15: 35521311-35521311 on Build GRCh38
rs1054590587 Chr.15: 35521275-35521275 on Build GRCh38
rs972585920 Chr.15: 35521077-35521077 on Build GRCh38
rs553996546 Chr.15: 35521063-35521063 on Build GRCh38
rs772424004 Chr.15: 35521127-35521127 on Build GRCh38
rs552587749 Chr.15: 35521364-35521364 on Build GRCh38
rs1048374279 Chr.15: 35521162-35521162 on Build GRCh38
rs530146104 Chr.15: 35521297-35521297 on Build GRCh38
rs560734023 Chr.15: 35521157-35521157 on Build GRCh38
rs540592079 Chr.15: 35521182-35521182 on Build GRCh38
rs534159908 Chr.15: 35521353-35521353 on Build GRCh38
rs928554678 Chr.15: 35521377-35521377 on Build GRCh38
rs962143494 Chr.15: 35521072-35521072 on Build GRCh38
rs180799889 Chr.15: 35521374-35521374 on Build GRCh38
rs112809854 Chr.15: 35521203-35521203 on Build GRCh38
rs1040540674 Chr.15: 35521219-35521219 on Build GRCh38
rs557169239 Chr.15: 35521053-35521053 on Build GRCh38
rs143864942 Chr.15: 35521111-35521111 on Build GRCh38
rs532652206 Chr.15: 35521222-35521222 on Build GRCh38
rs920585928 Chr.15: 35521125-35521125 on Build GRCh38
rs973949039 Chr.15: 35521368-35521368 on Build GRCh38
rs971295340 Chr.15: 35521045-35521045 on Build GRCh38
rs186257876 Chr.15: 35521296-35521296 on Build GRCh38
rs748716360 Chr.15: 35521114-35521114 on Build GRCh38
rs1022807731 Chr.15: 35521041-35521041 on Build GRCh38
rs894558960 Chr.15: 35521098-35521098 on Build GRCh38
rs1020208659 Chr.15: 35521185-35521185 on Build GRCh38
rs928454352 Chr.15: 35521031-35521031 on Build GRCh38
rs961525130 Chr.15: 35521354-35521354 on Build GRCh38
rs189627058 Chr.15: 35520999-35520999 on Build GRCh38

View Full Product Details