Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48755778-48756324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP152
Entrez Gene ID
22995
Gene Name
centrosomal protein 152
Gene Aliases
MCPH4, MCPH9, SCKL5
UniGene
Hs.443005
Gene Chromosome Location
Chr.15: 48662534-48811901 on Build GRCh38
Transcripts
NM_001194998.1 NM_014985.3 XM_006720437.3 XM_011521373.2 XM_011521374.2 XM_011521375.2 XM_011521378.2 XM_011521379.2 XM_011521381.2 XM_017022014.1 XM_017022015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5667054 c.3186C>A p.I1062I Chr.15: 48756062-48756062 on Build GRCh38
6189779 c.3325G>C p.A1109P Chr.15: 48755923-48755923 on Build GRCh38
3501840 c.3331C>G p.P1111A Chr.15: 48755917-48755917 on Build GRCh38
4937172 c.3057T>C p.C1019C Chr.15: 48756191-48756191 on Build GRCh38
3794216 c.3233C>T p.S1078F Chr.15: 48756015-48756015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779668798 Chr.15: 48755952-48755952 on Build GRCh38
rs770123187 Chr.15: 48756185-48756185 on Build GRCh38
rs764619845 Chr.15: 48755932-48755932 on Build GRCh38
rs972643438 Chr.15: 48755972-48755972 on Build GRCh38
rs747274010 Chr.15: 48756024-48756024 on Build GRCh38
rs780990996 Chr.15: 48755900-48755900 on Build GRCh38
rs763222881 Chr.15: 48756162-48756162 on Build GRCh38
rs554490098 Chr.15: 48756075-48756075 on Build GRCh38
rs754267846 Chr.15: 48755999-48755999 on Build GRCh38
rs866569090 Chr.15: 48756150-48756150 on Build GRCh38
rs758970441 Chr.15: 48756194-48756194 on Build GRCh38
rs372487939 Chr.15: 48756190-48756190 on Build GRCh38
rs375232696 Chr.15: 48756281-48756281 on Build GRCh38
rs765747552 Chr.15: 48756264-48756265 on Build GRCh38
rs369771086 Chr.15: 48755925-48755925 on Build GRCh38
rs267606718 Chr.15: 48756289-48756289 on Build GRCh38
rs200167001 Chr.15: 48756009-48756009 on Build GRCh38
rs775863754 Chr.15: 48755930-48755930 on Build GRCh38
rs537168507 Chr.15: 48755924-48755924 on Build GRCh38
rs549918537 Chr.15: 48755818-48755818 on Build GRCh38
rs74012142 Chr.15: 48755970-48755970 on Build GRCh38
rs754266800 Chr.15: 48755934-48755934 on Build GRCh38
rs772349993 Chr.15: 48755926-48755926 on Build GRCh38
rs16961557 Chr.15: 48755931-48755931 on Build GRCh38
rs779689504 Chr.15: 48755861-48755861 on Build GRCh38
rs201172776 Chr.15: 48756163-48756163 on Build GRCh38
rs754578791 Chr.15: 48756100-48756100 on Build GRCh38
rs754740345 Chr.15: 48755961-48755961 on Build GRCh38
rs767135964 Chr.15: 48756199-48756199 on Build GRCh38
rs748533734 Chr.15: 48756184-48756184 on Build GRCh38
rs756035852 Chr.15: 48755978-48755978 on Build GRCh38
rs767103785 Chr.15: 48756114-48756114 on Build GRCh38
rs745881226 Chr.15: 48756010-48756010 on Build GRCh38
rs200740117 Chr.15: 48755905-48755905 on Build GRCh38
rs199914670 Chr.15: 48756177-48756177 on Build GRCh38
rs553647106 Chr.15: 48755974-48755974 on Build GRCh38
rs953932004 Chr.15: 48756123-48756123 on Build GRCh38
rs765917624 Chr.15: 48756093-48756093 on Build GRCh38
rs762089839 Chr.15: 48756061-48756061 on Build GRCh38
rs773323076 Chr.15: 48755949-48755949 on Build GRCh38
rs587783422 Chr.15: 48756234-48756234 on Build GRCh38
rs1054434594 Chr.15: 48756226-48756226 on Build GRCh38
rs768502483 Chr.15: 48755872-48755872 on Build GRCh38
rs776732911 Chr.15: 48756178-48756178 on Build GRCh38
rs773681807 Chr.15: 48756188-48756188 on Build GRCh38
rs751283579 Chr.15: 48755953-48755953 on Build GRCh38
rs1029453867 Chr.15: 48756022-48756022 on Build GRCh38
rs758009260 Chr.15: 48755858-48755858 on Build GRCh38
rs566425914 Chr.15: 48755810-48755810 on Build GRCh38
rs1043960660 Chr.15: 48756219-48756219 on Build GRCh38
rs115832709 Chr.15: 48755923-48755923 on Build GRCh38
rs576054020 Chr.15: 48756131-48756131 on Build GRCh38
rs146955708 Chr.15: 48756264-48756264 on Build GRCh38
rs757694737 Chr.15: 48756217-48756219 on Build GRCh38
rs866582451 Chr.15: 48756147-48756147 on Build GRCh38
rs772335724 Chr.15: 48756014-48756014 on Build GRCh38
rs778999870 Chr.15: 48756267-48756267 on Build GRCh38
rs952431515 Chr.15: 48756044-48756044 on Build GRCh38
rs749882876 Chr.15: 48755941-48755941 on Build GRCh38
rs777016485 Chr.15: 48756054-48756054 on Build GRCh38
rs755071182 Chr.15: 48756288-48756288 on Build GRCh38
rs1018974682 Chr.15: 48755966-48755966 on Build GRCh38
rs765695843 Chr.15: 48755938-48755938 on Build GRCh38
rs746749686 Chr.15: 48755871-48755871 on Build GRCh38
rs184240876 Chr.15: 48756148-48756148 on Build GRCh38
rs997208404 Chr.15: 48756280-48756280 on Build GRCh38
rs1020780910 Chr.15: 48755981-48755981 on Build GRCh38
rs141275046 Chr.15: 48756058-48756058 on Build GRCh38
rs201655910 Chr.15: 48755884-48755884 on Build GRCh38
rs768878403 Chr.15: 48756040-48756040 on Build GRCh38
rs985110567 Chr.15: 48756126-48756126 on Build GRCh38
rs770176594 Chr.15: 48756290-48756290 on Build GRCh38
rs398122977 Chr.15: 48756099-48756099 on Build GRCh38
rs752467260 Chr.15: 48756129-48756129 on Build GRCh38
rs905854183 Chr.15: 48755811-48755811 on Build GRCh38
rs74553953 Chr.15: 48755935-48755935 on Build GRCh38
rs764433464 Chr.15: 48755970-48755971 on Build GRCh38
rs1046320145 Chr.15: 48756180-48756180 on Build GRCh38
rs377506426 Chr.15: 48756263-48756263 on Build GRCh38
rs945108502 Chr.15: 48756217-48756217 on Build GRCh38
rs981043818 Chr.15: 48756081-48756081 on Build GRCh38
rs773650149 Chr.15: 48756070-48756070 on Build GRCh38
rs587783423 Chr.15: 48756232-48756232 on Build GRCh38
rs202022140 Chr.15: 48756200-48756200 on Build GRCh38
rs775849447 Chr.15: 48756023-48756023 on Build GRCh38
rs773808366 Chr.15: 48756192-48756192 on Build GRCh38
rs869312853 Chr.15: 48756233-48756234 on Build GRCh38
rs777749052 Chr.15: 48756139-48756139 on Build GRCh38
rs752346294 Chr.15: 48756245-48756245 on Build GRCh38
rs755933405 Chr.15: 48756250-48756250 on Build GRCh38
rs765719526 Chr.15: 48756062-48756062 on Build GRCh38

View Full Product Details