Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51400282-51400788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDN
Entrez Gene ID
342035
Gene Name
gliomedin
Gene Aliases
CLOM, COLM, CRG-L2, CRGL2, UNC-112
UniGene
Hs.526441
Gene Chromosome Location
Chr.15: 51341516-51413365 on Build GRCh38
Transcripts
NM_181789.2 XM_011521501.2 XM_017022121.1 XM_017022122.1 XM_017022123.1 XM_017022124.1 XM_017022125.1 XM_017022126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2216917 c.934C>T p.Q312* Chr.15: 51400405-51400405 on Build GRCh38
1708169 c.976G>A p.E326K Chr.15: 51400447-51400447 on Build GRCh38
5583541 c.909G>A p.M303I Chr.15: 51400380-51400380 on Build GRCh38
7337727 c.1016A>T p.E339V Chr.15: 51400487-51400487 on Build GRCh38
1678457 c.1015G>T p.E339* Chr.15: 51400486-51400486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768273210 Chr.15: 51400522-51400522 on Build GRCh38
rs768664968 Chr.15: 51400437-51400437 on Build GRCh38
rs985958302 Chr.15: 51400608-51400608 on Build GRCh38
rs781669571 Chr.15: 51400311-51400311 on Build GRCh38
rs758156940 Chr.15: 51400515-51400515 on Build GRCh38
rs929126491 Chr.15: 51400447-51400447 on Build GRCh38
rs745503110 Chr.15: 51400413-51400413 on Build GRCh38
rs1031463148 Chr.15: 51400745-51400745 on Build GRCh38
rs150238396 Chr.15: 51400539-51400539 on Build GRCh38
rs756672506 Chr.15: 51400378-51400378 on Build GRCh38
rs767684508 Chr.15: 51400473-51400473 on Build GRCh38
rs911347827 Chr.15: 51400631-51400631 on Build GRCh38
rs199785314 Chr.15: 51400402-51400402 on Build GRCh38
rs776291474 Chr.15: 51400511-51400511 on Build GRCh38
rs779966031 Chr.15: 51400427-51400427 on Build GRCh38
rs560880760 Chr.15: 51400643-51400643 on Build GRCh38
rs879887295 Chr.15: 51400735-51400735 on Build GRCh38
rs764667400 Chr.15: 51400486-51400486 on Build GRCh38
rs771990009 Chr.15: 51400330-51400330 on Build GRCh38
rs759948387 Chr.15: 51400371-51400371 on Build GRCh38
rs771636580 Chr.15: 51400575-51400575 on Build GRCh38
rs370260835 Chr.15: 51400536-51400536 on Build GRCh38
rs189782108 Chr.15: 51400471-51400471 on Build GRCh38
rs563893526 Chr.15: 51400422-51400422 on Build GRCh38
rs762894985 Chr.15: 51400349-51400349 on Build GRCh38
rs777304415 Chr.15: 51400321-51400321 on Build GRCh38
rs756850716 Chr.15: 51400510-51400510 on Build GRCh38
rs753079395 Chr.15: 51400475-51400475 on Build GRCh38
rs773817522 Chr.15: 51400326-51400326 on Build GRCh38
rs763574477 Chr.15: 51400422-51400423 on Build GRCh38
rs569076676 Chr.15: 51400587-51400587 on Build GRCh38
rs74013207 Chr.15: 51400738-51400738 on Build GRCh38
rs944016924 Chr.15: 51400637-51400637 on Build GRCh38
rs376628257 Chr.15: 51400376-51400376 on Build GRCh38
rs765274993 Chr.15: 51400343-51400343 on Build GRCh38
rs527786736 Chr.15: 51400570-51400570 on Build GRCh38
rs752925482 Chr.15: 51400734-51400734 on Build GRCh38
rs775626584 Chr.15: 51400334-51400334 on Build GRCh38
rs753240580 Chr.15: 51400377-51400377 on Build GRCh38
rs369681680 Chr.15: 51400345-51400345 on Build GRCh38
rs748009552 Chr.15: 51400534-51400534 on Build GRCh38
rs753399748 Chr.15: 51400397-51400397 on Build GRCh38
rs373570825 Chr.15: 51400479-51400479 on Build GRCh38
rs767604332 Chr.15: 51400681-51400681 on Build GRCh38
rs1047513820 Chr.15: 51400726-51400726 on Build GRCh38
rs867806350 Chr.15: 51400498-51400498 on Build GRCh38
rs748704334 Chr.15: 51400324-51400324 on Build GRCh38
rs141102085 Chr.15: 51400528-51400528 on Build GRCh38
rs754295348 Chr.15: 51400496-51400496 on Build GRCh38
rs745662373 Chr.15: 51400328-51400328 on Build GRCh38
rs978229711 Chr.15: 51400584-51400584 on Build GRCh38
rs376878868 Chr.15: 51400337-51400337 on Build GRCh38
rs114537731 Chr.15: 51400706-51400706 on Build GRCh38
rs763102564 Chr.15: 51400463-51400463 on Build GRCh38
rs72729238 Chr.15: 51400350-51400350 on Build GRCh38
rs748636690 Chr.15: 51400312-51400312 on Build GRCh38
rs375652066 Chr.15: 51400518-51400518 on Build GRCh38
rs923160729 Chr.15: 51400646-51400646 on Build GRCh38
rs574215097 Chr.15: 51400393-51400393 on Build GRCh38
rs756655568 Chr.15: 51400401-51400401 on Build GRCh38
rs746742376 Chr.15: 51400432-51400432 on Build GRCh38
rs751717350 Chr.15: 51400352-51400352 on Build GRCh38
rs776377579 Chr.15: 51400457-51400457 on Build GRCh38
rs772293339 Chr.15: 51400550-51400550 on Build GRCh38
rs755420151 Chr.15: 51400450-51400451 on Build GRCh38
rs866030739 Chr.15: 51400642-51400642 on Build GRCh38
rs549499079 Chr.15: 51400599-51400599 on Build GRCh38
rs755624810 Chr.15: 51400320-51400320 on Build GRCh38
rs759560192 Chr.15: 51400540-51400540 on Build GRCh38
rs759719608 Chr.15: 51400472-51400472 on Build GRCh38
rs770636789 Chr.15: 51400325-51400325 on Build GRCh38
rs750142040 Chr.15: 51400513-51400513 on Build GRCh38
rs773374767 Chr.15: 51400344-51400344 on Build GRCh38

View Full Product Details