Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63625865-63626413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7201400 c.13106-8C>G p.? Chr.15: 63626162-63626162 on Build GRCh38
1209550 c.13163C>T p.A4388V Chr.15: 63626097-63626097 on Build GRCh38
4819523 c.13225G>A p.D4409N Chr.15: 63626035-63626035 on Build GRCh38
4056125 c.13236C>T p.Y4412Y Chr.15: 63626024-63626024 on Build GRCh38
4393982 c.13136C>T p.P4379L Chr.15: 63626124-63626124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927524363 Chr.15: 63626043-63626043 on Build GRCh38
rs892745365 Chr.15: 63626310-63626310 on Build GRCh38
rs770061334 Chr.15: 63626124-63626124 on Build GRCh38
rs751529066 Chr.15: 63626151-63626151 on Build GRCh38
rs777606490 Chr.15: 63626181-63626181 on Build GRCh38
rs570689875 Chr.15: 63626242-63626242 on Build GRCh38
rs190991795 Chr.15: 63626060-63626060 on Build GRCh38
rs747559050 Chr.15: 63626200-63626200 on Build GRCh38
rs971702979 Chr.15: 63625964-63625964 on Build GRCh38
rs754539328 Chr.15: 63626091-63626091 on Build GRCh38
rs796595717 Chr.15: 63626359-63626359 on Build GRCh38
rs775930285 Chr.15: 63626073-63626073 on Build GRCh38
rs773401851 Chr.15: 63626127-63626127 on Build GRCh38
rs756741206 Chr.15: 63626216-63626218 on Build GRCh38
rs911032874 Chr.15: 63626364-63626364 on Build GRCh38
rs780926289 Chr.15: 63625960-63625960 on Build GRCh38
rs1034482216 Chr.15: 63626258-63626258 on Build GRCh38
rs1140422 Chr.15: 63626108-63626108 on Build GRCh38
rs908480292 Chr.15: 63626172-63626172 on Build GRCh38
rs958862196 Chr.15: 63626281-63626281 on Build GRCh38
rs1020307564 Chr.15: 63626241-63626241 on Build GRCh38
rs1009497231 Chr.15: 63626292-63626292 on Build GRCh38
rs747429095 Chr.15: 63626114-63626114 on Build GRCh38
rs780508835 Chr.15: 63626093-63626093 on Build GRCh38
rs772789325 Chr.15: 63626106-63626107 on Build GRCh38
rs556075335 Chr.15: 63626312-63626312 on Build GRCh38
rs769386386 Chr.15: 63625934-63625960 on Build GRCh38
rs1021962575 Chr.15: 63626099-63626099 on Build GRCh38
rs753978228 Chr.15: 63625917-63625917 on Build GRCh38
rs772690412 Chr.15: 63625972-63625972 on Build GRCh38
rs762837087 Chr.15: 63626138-63626138 on Build GRCh38
rs918296735 Chr.15: 63626347-63626347 on Build GRCh38
rs139461010 Chr.15: 63626198-63626198 on Build GRCh38
rs899167343 Chr.15: 63626229-63626229 on Build GRCh38
rs199544390 Chr.15: 63626109-63626109 on Build GRCh38
rs768798083 Chr.15: 63626116-63626116 on Build GRCh38
rs752398916 Chr.15: 63626096-63626096 on Build GRCh38
rs757079331 Chr.15: 63625918-63625918 on Build GRCh38
rs369029117 Chr.15: 63626055-63626055 on Build GRCh38
rs2228516 Chr.15: 63626080-63626080 on Build GRCh38
rs183347780 Chr.15: 63626326-63626326 on Build GRCh38
rs775834711 Chr.15: 63626018-63626018 on Build GRCh38
rs759324456 Chr.15: 63626158-63626158 on Build GRCh38
rs1051025798 Chr.15: 63626355-63626359 on Build GRCh38
rs766037239 Chr.15: 63626148-63626148 on Build GRCh38
rs746280736 Chr.15: 63626107-63626107 on Build GRCh38
rs767554358 Chr.15: 63626170-63626170 on Build GRCh38
rs774786065 Chr.15: 63625971-63625971 on Build GRCh38
rs765021229 Chr.15: 63626068-63626068 on Build GRCh38
rs751411027 Chr.15: 63626084-63626084 on Build GRCh38
rs927701403 Chr.15: 63626110-63626110 on Build GRCh38
rs200491054 Chr.15: 63626067-63626067 on Build GRCh38
rs760851641 Chr.15: 63626049-63626049 on Build GRCh38
rs1029154469 Chr.15: 63626027-63626027 on Build GRCh38
rs941270231 Chr.15: 63626184-63626184 on Build GRCh38
rs755660453 Chr.15: 63626178-63626178 on Build GRCh38
rs1003161775 Chr.15: 63626246-63626246 on Build GRCh38
rs990544655 Chr.15: 63626291-63626291 on Build GRCh38
rs754413826 Chr.15: 63625947-63625947 on Build GRCh38
rs1057518376 Chr.15: 63626118-63626118 on Build GRCh38
rs758815284 Chr.15: 63626195-63626195 on Build GRCh38
rs771434430 Chr.15: 63625961-63625961 on Build GRCh38
rs746588160 Chr.15: 63625945-63625945 on Build GRCh38
rs550919679 Chr.15: 63625929-63625929 on Build GRCh38
rs746029933 Chr.15: 63626380-63626380 on Build GRCh38
rs909644853 Chr.15: 63626294-63626294 on Build GRCh38
rs753504344 Chr.15: 63626186-63626186 on Build GRCh38
rs375721003 Chr.15: 63626022-63626022 on Build GRCh38
rs1031149896 Chr.15: 63626308-63626308 on Build GRCh38
rs982614751 Chr.15: 63626147-63626147 on Build GRCh38
rs777743186 Chr.15: 63626056-63626056 on Build GRCh38
rs144162223 Chr.15: 63626348-63626348 on Build GRCh38
rs372746021 Chr.15: 63626072-63626072 on Build GRCh38
rs776626146 Chr.15: 63626119-63626119 on Build GRCh38
rs532386086 Chr.15: 63625901-63625901 on Build GRCh38
rs761877909 Chr.15: 63626120-63626120 on Build GRCh38
rs777551579 Chr.15: 63626098-63626098 on Build GRCh38
rs374706985 Chr.15: 63626171-63626171 on Build GRCh38
rs149868040 Chr.15: 63626244-63626244 on Build GRCh38
rs764966855 Chr.15: 63625923-63625923 on Build GRCh38
rs779852700 Chr.15: 63625942-63625942 on Build GRCh38
rs1010465639 Chr.15: 63625934-63625934 on Build GRCh38
rs755893599 Chr.15: 63626097-63626097 on Build GRCh38
rs939196743 Chr.15: 63626118-63626118 on Build GRCh38
rs62012825 Chr.15: 63626369-63626369 on Build GRCh38
rs890517200 Chr.15: 63626275-63626275 on Build GRCh38
rs893112243 Chr.15: 63626212-63626212 on Build GRCh38

View Full Product Details