Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66293188-66293674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L
Entrez Gene ID
115752
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
Gene Aliases
DIS3L1
UniGene
Hs.446251
Gene Chromosome Location
Chr.15: 66293257-66333898 on Build GRCh38
Transcripts
NM_001143688.2 NM_001323936.1 NM_001323937.1 NM_001323938.1 NM_001323939.1 NM_001323940.1 NM_133375.4 XM_005254146.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5416949 c.37A>T p.T13S Chr.15: 66293633-66293633 on Build GRCh38
4627340 c.77A>G p.Y26C Chr.15: 66293673-66293673 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1010967808 Chr.15: 66293586-66293586 on Build GRCh38
rs769699419 Chr.15: 66293286-66293286 on Build GRCh38
rs1014954822 Chr.15: 66293288-66293288 on Build GRCh38
rs1034881630 Chr.15: 66293479-66293479 on Build GRCh38
rs932215494 Chr.15: 66293552-66293552 on Build GRCh38
rs981953956 Chr.15: 66293247-66293247 on Build GRCh38
rs966636576 Chr.15: 66293472-66293472 on Build GRCh38
rs1035355510 Chr.15: 66293597-66293597 on Build GRCh38
rs139096215 Chr.15: 66293533-66293533 on Build GRCh38
rs764518033 Chr.15: 66293565-66293565 on Build GRCh38
rs544398957 Chr.15: 66293457-66293457 on Build GRCh38
rs895099218 Chr.15: 66293604-66293604 on Build GRCh38
rs996337875 Chr.15: 66293255-66293255 on Build GRCh38
rs1003474878 Chr.15: 66293310-66293310 on Build GRCh38
rs75040294 Chr.15: 66293569-66293569 on Build GRCh38
rs990635708 Chr.15: 66293414-66293414 on Build GRCh38
rs900683523 Chr.15: 66293243-66293243 on Build GRCh38
rs775914855 Chr.15: 66293561-66293561 on Build GRCh38
rs185282271 Chr.15: 66293372-66293372 on Build GRCh38
rs1051903035 Chr.15: 66293358-66293358 on Build GRCh38
rs773422525 Chr.15: 66293263-66293263 on Build GRCh38
rs922170925 Chr.15: 66293536-66293536 on Build GRCh38
rs1049268489 Chr.15: 66293554-66293554 on Build GRCh38
rs907220653 Chr.15: 66293581-66293581 on Build GRCh38
rs776883066 Chr.15: 66293620-66293620 on Build GRCh38
rs971374849 Chr.15: 66293292-66293292 on Build GRCh38
rs902503635 Chr.15: 66293633-66293633 on Build GRCh38
rs1002834848 Chr.15: 66293591-66293591 on Build GRCh38
rs1036663656 Chr.15: 66293302-66293302 on Build GRCh38
rs561104597 Chr.15: 66293502-66293502 on Build GRCh38
rs1011332649 Chr.15: 66293449-66293449 on Build GRCh38
rs1022152791 Chr.15: 66293618-66293618 on Build GRCh38
rs1018101704 Chr.15: 66293403-66293403 on Build GRCh38
rs200010364 Chr.15: 66293611-66293611 on Build GRCh38
rs966583260 Chr.15: 66293236-66293236 on Build GRCh38
rs149917229 Chr.15: 66293534-66293534 on Build GRCh38
rs923640484 Chr.15: 66293495-66293495 on Build GRCh38
rs927759533 Chr.15: 66293260-66293260 on Build GRCh38
rs1036418729 Chr.15: 66293575-66293575 on Build GRCh38
rs943535236 Chr.15: 66293565-66293565 on Build GRCh38
rs1055270939 Chr.15: 66293269-66293270 on Build GRCh38
rs565223776 Chr.15: 66293328-66293328 on Build GRCh38
rs887900392 Chr.15: 66293270-66293270 on Build GRCh38
rs994432627 Chr.15: 66293277-66293277 on Build GRCh38
rs893850634 Chr.15: 66293417-66293420 on Build GRCh38
rs565794629 Chr.15: 66293562-66293562 on Build GRCh38
rs1051957787 Chr.15: 66293559-66293559 on Build GRCh38
rs1007630439 Chr.15: 66293373-66293373 on Build GRCh38
rs993685199 Chr.15: 66293353-66293353 on Build GRCh38
rs929586464 Chr.15: 66293544-66293544 on Build GRCh38
rs370213594 Chr.15: 66293300-66293300 on Build GRCh38
rs761761907 Chr.15: 66293651-66293651 on Build GRCh38
rs959186642 Chr.15: 66293493-66293493 on Build GRCh38
rs898081511 Chr.15: 66293319-66293319 on Build GRCh38
rs982007735 Chr.15: 66293477-66293477 on Build GRCh38
rs868833814 Chr.15: 66293317-66293317 on Build GRCh38
rs915052811 Chr.15: 66293455-66293455 on Build GRCh38
rs977792776 Chr.15: 66293458-66293458 on Build GRCh38

View Full Product Details