Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90890876-90891380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FES
Entrez Gene ID
2242
Gene Name
FES proto-oncogene, tyrosine kinase
Gene Aliases
FPS
UniGene
Hs.7636
Gene Chromosome Location
Chr.15: 90884421-90895776 on Build GRCh38
Transcripts
NM_001143783.1 NM_001143784.1 NM_001143785.1 NM_002005.3 XM_005254880.1 XM_005254882.1 XM_017022005.1 XM_017022006.1 XM_017022007.1 XM_017022008.1 XM_017022009.1 XM_017022010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787788 c.1430C>T p.S477F Chr.15: 90891091-90891091 on Build GRCh38
5790496 c.1331C>T p.P444L Chr.15: 90890992-90890992 on Build GRCh38
3741735 c.1481C>T p.S494L Chr.15: 90891142-90891142 on Build GRCh38
1563291 c.1328C>A p.P443Q Chr.15: 90890989-90890989 on Build GRCh38
6838889 c.1354C>T p.Q452* Chr.15: 90891015-90891015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772624199 Chr.15: 90890988-90890988 on Build GRCh38
rs755250119 Chr.15: 90890967-90890967 on Build GRCh38
rs755183833 Chr.15: 90891065-90891065 on Build GRCh38
rs901339449 Chr.15: 90891221-90891221 on Build GRCh38
rs764010129 Chr.15: 90890943-90890943 on Build GRCh38
rs56387260 Chr.15: 90891038-90891038 on Build GRCh38
rs555184567 Chr.15: 90891286-90891286 on Build GRCh38
rs780219022 Chr.15: 90890964-90890964 on Build GRCh38
rs773083019 Chr.15: 90891189-90891189 on Build GRCh38
rs72755390 Chr.15: 90891237-90891237 on Build GRCh38
rs756122752 Chr.15: 90890958-90890958 on Build GRCh38
rs751551364 Chr.15: 90890954-90890954 on Build GRCh38
rs75632404 Chr.15: 90890940-90890940 on Build GRCh38
rs765449538 Chr.15: 90891206-90891206 on Build GRCh38
rs769445323 Chr.15: 90891021-90891021 on Build GRCh38
rs367707420 Chr.15: 90891201-90891201 on Build GRCh38
rs34086251 Chr.15: 90890992-90890992 on Build GRCh38
rs752187808 Chr.15: 90891174-90891174 on Build GRCh38
rs775187033 Chr.15: 90891027-90891027 on Build GRCh38
rs890814829 Chr.15: 90890926-90890926 on Build GRCh38
rs1039798978 Chr.15: 90891180-90891180 on Build GRCh38
rs758806464 Chr.15: 90891094-90891094 on Build GRCh38
rs188984508 Chr.15: 90891142-90891142 on Build GRCh38
rs774122108 Chr.15: 90891057-90891057 on Build GRCh38
rs757798296 Chr.15: 90891108-90891108 on Build GRCh38
rs753965684 Chr.15: 90890965-90890965 on Build GRCh38
rs1022353194 Chr.15: 90891099-90891099 on Build GRCh38
rs1036305263 Chr.15: 90890923-90890923 on Build GRCh38
rs370487535 Chr.15: 90891159-90891159 on Build GRCh38
rs1008273508 Chr.15: 90890928-90890928 on Build GRCh38
rs932238304 Chr.15: 90891230-90891230 on Build GRCh38
rs767493516 Chr.15: 90891184-90891184 on Build GRCh38
rs759192417 Chr.15: 90891115-90891115 on Build GRCh38
rs769283692 Chr.15: 90890916-90890916 on Build GRCh38
rs774217594 Chr.15: 90891163-90891163 on Build GRCh38
rs113748785 Chr.15: 90891211-90891211 on Build GRCh38
rs745457962 Chr.15: 90891008-90891008 on Build GRCh38
rs869160421 Chr.15: 90890939-90890939 on Build GRCh38
rs927834924 Chr.15: 90890915-90890915 on Build GRCh38
rs1006979502 Chr.15: 90890983-90890983 on Build GRCh38
rs192241384 Chr.15: 90891165-90891165 on Build GRCh38
rs146754390 Chr.15: 90891058-90891058 on Build GRCh38
rs779361595 Chr.15: 90890981-90890981 on Build GRCh38
rs1039362573 Chr.15: 90890929-90890929 on Build GRCh38
rs569901538 Chr.15: 90891214-90891214 on Build GRCh38
rs753016539 Chr.15: 90891073-90891073 on Build GRCh38
rs897470860 Chr.15: 90890930-90890930 on Build GRCh38
rs753965166 Chr.15: 90891059-90891059 on Build GRCh38
rs1034352374 Chr.15: 90891052-90891052 on Build GRCh38
rs1020402719 Chr.15: 90891331-90891331 on Build GRCh38
rs377147995 Chr.15: 90891069-90891069 on Build GRCh38
rs115603049 Chr.15: 90891338-90891338 on Build GRCh38
rs778314889 Chr.15: 90891095-90891095 on Build GRCh38
rs1010327455 Chr.15: 90891251-90891251 on Build GRCh38
rs888868014 Chr.15: 90890952-90890952 on Build GRCh38
rs796353096 Chr.15: 90891210-90891210 on Build GRCh38
rs1054611979 Chr.15: 90891234-90891234 on Build GRCh38
rs377056494 Chr.15: 90890993-90890993 on Build GRCh38
rs760787408 Chr.15: 90890955-90890955 on Build GRCh38
rs139460083 Chr.15: 90891134-90891134 on Build GRCh38
rs373220201 Chr.15: 90891048-90891048 on Build GRCh38
rs568632074 Chr.15: 90891143-90891143 on Build GRCh38
rs757164397 Chr.15: 90891317-90891317 on Build GRCh38
rs753716736 Chr.15: 90891276-90891276 on Build GRCh38
rs748519202 Chr.15: 90890986-90890986 on Build GRCh38
rs892725026 Chr.15: 90891316-90891316 on Build GRCh38
rs776230369 Chr.15: 90891007-90891007 on Build GRCh38
rs777214342 Chr.15: 90890917-90890917 on Build GRCh38
rs775097175 Chr.15: 90891155-90891155 on Build GRCh38
rs2229074 Chr.15: 90891047-90891047 on Build GRCh38
rs747428833 Chr.15: 90891101-90891101 on Build GRCh38
rs747145494 Chr.15: 90890898-90890898 on Build GRCh38
rs779860529 Chr.15: 90891343-90891343 on Build GRCh38
rs767203953 Chr.15: 90891166-90891166 on Build GRCh38

View Full Product Details