Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 282205-282693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIG
Entrez Gene ID
398
Gene Name
Rho GDP dissociation inhibitor gamma
Gene Aliases
RHOGDI-3
UniGene
Hs.647388
Gene Chromosome Location
Chr.16: 280606-283003 on Build GRCh38
Transcripts
NM_001176.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5990348 c.357G>A p.K119K Chr.16: 282316-282316 on Build GRCh38
3370420 c.552G>A p.E184E Chr.16: 282688-282688 on Build GRCh38
6257405 c.415G>A p.V139I Chr.16: 282467-282467 on Build GRCh38
39849 c.424G>A p.E142K Chr.16: 282476-282476 on Build GRCh38
1283485 c.475C>T p.R159C Chr.16: 282527-282527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757985558 Chr.16: 282569-282569 on Build GRCh38
rs752239284 Chr.16: 282423-282424 on Build GRCh38
rs146858509 Chr.16: 282529-282529 on Build GRCh38
rs781704087 Chr.16: 282487-282487 on Build GRCh38
rs776991799 Chr.16: 282332-282332 on Build GRCh38
rs370015159 Chr.16: 282606-282606 on Build GRCh38
rs762523264 Chr.16: 282352-282352 on Build GRCh38
rs757331123 Chr.16: 282464-282464 on Build GRCh38
rs775334163 Chr.16: 282350-282350 on Build GRCh38
rs893158466 Chr.16: 282413-282413 on Build GRCh38
rs746288496 Chr.16: 282340-282340 on Build GRCh38
rs374517960 Chr.16: 282435-282435 on Build GRCh38
rs762129264 Chr.16: 282527-282527 on Build GRCh38
rs868311072 Chr.16: 282305-282305 on Build GRCh38
rs772551912 Chr.16: 282250-282250 on Build GRCh38
rs766514614 Chr.16: 282604-282604 on Build GRCh38
rs753113349 Chr.16: 282567-282567 on Build GRCh38
rs758644519 Chr.16: 282385-282385 on Build GRCh38
rs769738212 Chr.16: 282577-282577 on Build GRCh38
rs138141299 Chr.16: 282655-282655 on Build GRCh38
rs191861590 Chr.16: 282576-282576 on Build GRCh38
rs769474713 Chr.16: 282344-282344 on Build GRCh38
rs765644440 Chr.16: 282667-282667 on Build GRCh38
rs199970245 Chr.16: 282493-282493 on Build GRCh38
rs200937476 Chr.16: 282270-282270 on Build GRCh38
rs145921827 Chr.16: 282632-282632 on Build GRCh38
rs755991216 Chr.16: 282651-282651 on Build GRCh38
rs150946433 Chr.16: 282329-282329 on Build GRCh38
rs767420406 Chr.16: 282367-282367 on Build GRCh38
rs747659030 Chr.16: 282506-282506 on Build GRCh38
rs764280047 Chr.16: 282519-282519 on Build GRCh38
rs963397973 Chr.16: 282516-282516 on Build GRCh38
rs774010528 Chr.16: 282364-282364 on Build GRCh38
rs759388293 Chr.16: 282518-282518 on Build GRCh38
rs536785569 Chr.16: 282229-282229 on Build GRCh38
rs772798542 Chr.16: 282509-282509 on Build GRCh38
rs771280898 Chr.16: 282656-282656 on Build GRCh38
rs754352953 Chr.16: 282563-282563 on Build GRCh38
rs746221953 Chr.16: 282407-282407 on Build GRCh38
rs758849902 Chr.16: 282619-282619 on Build GRCh38
rs749164257 Chr.16: 282599-282599 on Build GRCh38
rs964268146 Chr.16: 282475-282475 on Build GRCh38
rs921664670 Chr.16: 282258-282258 on Build GRCh38
rs750187935 Chr.16: 282290-282290 on Build GRCh38
rs745448650 Chr.16: 282648-282648 on Build GRCh38
rs200392108 Chr.16: 282542-282542 on Build GRCh38
rs772166065 Chr.16: 282568-282568 on Build GRCh38
rs995834252 Chr.16: 282481-282481 on Build GRCh38
rs771766864 Chr.16: 282546-282546 on Build GRCh38
rs868150795 Chr.16: 282482-282482 on Build GRCh38
rs765842218 Chr.16: 282607-282607 on Build GRCh38
rs764370393 Chr.16: 282451-282451 on Build GRCh38
rs950333827 Chr.16: 282276-282276 on Build GRCh38
rs760475193 Chr.16: 282510-282510 on Build GRCh38
rs200441576 Chr.16: 282293-282293 on Build GRCh38
rs201140557 Chr.16: 282394-282394 on Build GRCh38
rs776886528 Chr.16: 282263-282263 on Build GRCh38
rs140777703 Chr.16: 282373-282373 on Build GRCh38
rs759751565 Chr.16: 282663-282663 on Build GRCh38
rs200673694 Chr.16: 282395-282395 on Build GRCh38
rs771441255 Chr.16: 282508-282508 on Build GRCh38
rs903906809 Chr.16: 282386-282386 on Build GRCh38
rs756683598 Chr.16: 282531-282531 on Build GRCh38
rs766109887 Chr.16: 282442-282442 on Build GRCh38
rs746482632 Chr.16: 282572-282572 on Build GRCh38
rs777084649 Chr.16: 282550-282550 on Build GRCh38
rs554741124 Chr.16: 282252-282252 on Build GRCh38
rs759172417 Chr.16: 282269-282269 on Build GRCh38
rs554911623 Chr.16: 282652-282652 on Build GRCh38
rs772162740 Chr.16: 282601-282601 on Build GRCh38
rs796944167 Chr.16: 282567-282567 on Build GRCh38
rs535873961 Chr.16: 282547-282547 on Build GRCh38
rs770846119 Chr.16: 282554-282554 on Build GRCh38
rs757781938 Chr.16: 282398-282398 on Build GRCh38
rs751588124 Chr.16: 282275-282275 on Build GRCh38
rs201745267 Chr.16: 282603-282603 on Build GRCh38
rs772174910 Chr.16: 282301-282301 on Build GRCh38
rs771290643 Chr.16: 282254-282254 on Build GRCh38
rs61572786 Chr.16: 282559-282559 on Build GRCh38
rs373716691 Chr.16: 282291-282291 on Build GRCh38
rs775501723 Chr.16: 282582-282583 on Build GRCh38
rs765459963 Chr.16: 282565-282565 on Build GRCh38
rs748890806 Chr.16: 282412-282412 on Build GRCh38
rs77035514 Chr.16: 282566-282567 on Build GRCh38
rs769539283 Chr.16: 282560-282560 on Build GRCh38
rs761286394 Chr.16: 282429-282429 on Build GRCh38
rs111793116 Chr.16: 282271-282271 on Build GRCh38
rs756656270 Chr.16: 282489-282489 on Build GRCh38
rs201950145 Chr.16: 282503-282503 on Build GRCh38
rs376948933 Chr.16: 282553-282553 on Build GRCh38
rs770098316 Chr.16: 282408-282408 on Build GRCh38
rs777544650 Chr.16: 282571-282571 on Build GRCh38
rs755585891 Chr.16: 282513-282515 on Build GRCh38
rs748958834 Chr.16: 282558-282558 on Build GRCh38
rs752994129 Chr.16: 282610-282610 on Build GRCh38
rs780624082 Chr.16: 282634-282634 on Build GRCh38
rs916396850 Chr.16: 282376-282376 on Build GRCh38
rs199594877 Chr.16: 282411-282411 on Build GRCh38
rs768425562 Chr.16: 282415-282415 on Build GRCh38
rs768409191 Chr.16: 282495-282495 on Build GRCh38
rs369485603 Chr.16: 282343-282343 on Build GRCh38
rs763648457 Chr.16: 282357-282357 on Build GRCh38
rs765465452 Chr.16: 282383-282383 on Build GRCh38
rs778982863 Chr.16: 282247-282247 on Build GRCh38
rs778827738 Chr.16: 282558-282559 on Build GRCh38
rs1023523006 Chr.16: 282537-282537 on Build GRCh38
rs1016554903 Chr.16: 282347-282347 on Build GRCh38
rs776862450 Chr.16: 282605-282605 on Build GRCh38
rs758551479 Chr.16: 282448-282448 on Build GRCh38
rs773993072 Chr.16: 282417-282417 on Build GRCh38
rs1017762514 Chr.16: 282231-282231 on Build GRCh38
rs748185415 Chr.16: 282584-282585 on Build GRCh38
rs747704322 Chr.16: 282545-282545 on Build GRCh38
rs779448151 Chr.16: 282544-282544 on Build GRCh38
rs142556476 Chr.16: 282325-282325 on Build GRCh38
rs900648285 Chr.16: 282445-282445 on Build GRCh38
rs1030051322 Chr.16: 282460-282460 on Build GRCh38
rs748291024 Chr.16: 282248-282248 on Build GRCh38
rs780497810 Chr.16: 282491-282491 on Build GRCh38
rs753649528 Chr.16: 282447-282447 on Build GRCh38
rs777305625 Chr.16: 282529-282530 on Build GRCh38
rs776848516 Chr.16: 282657-282657 on Build GRCh38
rs200907964 Chr.16: 282441-282441 on Build GRCh38
rs775142881 Chr.16: 282589-282589 on Build GRCh38
rs900698884 Chr.16: 282251-282251 on Build GRCh38
rs777858497 Chr.16: 282311-282311 on Build GRCh38
rs750455744 Chr.16: 282562-282562 on Build GRCh38
rs781609081 Chr.16: 282402-282402 on Build GRCh38
rs761872944 Chr.16: 282561-282561 on Build GRCh38
rs776603367 Chr.16: 282559-282559 on Build GRCh38
rs199532923 Chr.16: 282382-282382 on Build GRCh38
rs940528772 Chr.16: 282278-282278 on Build GRCh38
rs201741302 Chr.16: 282626-282626 on Build GRCh38
rs780763784 Chr.16: 282535-282535 on Build GRCh38
rs764101027 Chr.16: 282274-282274 on Build GRCh38
rs572337864 Chr.16: 282228-282228 on Build GRCh38
rs140706640 Chr.16: 282625-282625 on Build GRCh38
rs202008361 Chr.16: 282438-282438 on Build GRCh38
rs780983164 Chr.16: 282281-282281 on Build GRCh38
rs745607553 Chr.16: 282559-282560 on Build GRCh38
rs143766514 Chr.16: 282488-282488 on Build GRCh38
rs368770509 Chr.16: 282574-282574 on Build GRCh38
rs751629355 Chr.16: 282457-282457 on Build GRCh38
rs201067267 Chr.16: 282541-282541 on Build GRCh38
rs574851094 Chr.16: 282279-282279 on Build GRCh38
rs376348467 Chr.16: 282515-282515 on Build GRCh38

View Full Product Details