Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66603950-66604441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM3
Entrez Gene ID
123920
Gene Name
CKLF like MARVEL transmembrane domain containing 3
Gene Aliases
BNAS2, CKLFSF3
UniGene
Hs.298198
Gene Chromosome Location
Chr.16: 66604032-66613892 on Build GRCh38
Transcripts
NM_144601.3 NR_037613.1 XM_006721131.1 XM_017022954.1

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
UniGene
Hs.643961
Gene Chromosome Location
Chr.16: 66598178-66696707 on Build GRCh38
Transcripts
NM_144601.3 NR_037613.1 XM_006721131.1 XM_017022954.1

SNP Detail

SNP ID Chromosome Location
rs1032928796 Chr.16: 66604188-66604188 on Build GRCh38
rs1056871887 Chr.16: 66604357-66604357 on Build GRCh38
rs1039803584 Chr.16: 66604261-66604261 on Build GRCh38
rs888738443 Chr.16: 66604102-66604102 on Build GRCh38
rs774241178 Chr.16: 66604033-66604034 on Build GRCh38
rs918934880 Chr.16: 66604054-66604054 on Build GRCh38
rs569210233 Chr.16: 66604168-66604168 on Build GRCh38
rs546925441 Chr.16: 66604053-66604053 on Build GRCh38
rs771889351 Chr.16: 66604134-66604135 on Build GRCh38
rs944078921 Chr.16: 66604230-66604230 on Build GRCh38
rs1014774670 Chr.16: 66604134-66604134 on Build GRCh38
rs963370026 Chr.16: 66604421-66604421 on Build GRCh38
rs760882213 Chr.16: 66604142-66604143 on Build GRCh38
rs45446298 Chr.16: 66604128-66604128 on Build GRCh38
rs796439152 Chr.16: 66604047-66604051 on Build GRCh38
rs929001961 Chr.16: 66604055-66604055 on Build GRCh38
rs1056374808 Chr.16: 66604127-66604136 on Build GRCh38
rs998881977 Chr.16: 66604174-66604174 on Build GRCh38
rs895667893 Chr.16: 66604380-66604380 on Build GRCh38
rs1042986737 Chr.16: 66604073-66604073 on Build GRCh38
rs944005139 Chr.16: 66604033-66604033 on Build GRCh38
rs977350641 Chr.16: 66604031-66604032 on Build GRCh38
rs35320143 Chr.16: 66604342-66604342 on Build GRCh38
rs944454339 Chr.16: 66604052-66604053 on Build GRCh38
rs75682199 Chr.16: 66604032-66604032 on Build GRCh38
rs535720058 Chr.16: 66604140-66604140 on Build GRCh38
rs921229925 Chr.16: 66604287-66604287 on Build GRCh38
rs1014222376 Chr.16: 66603972-66603972 on Build GRCh38
rs1024315561 Chr.16: 66603995-66603995 on Build GRCh38
rs749278211 Chr.16: 66604032-66604033 on Build GRCh38
rs941530729 Chr.16: 66604125-66604132 on Build GRCh38
rs947450218 Chr.16: 66604289-66604289 on Build GRCh38
rs912632079 Chr.16: 66604217-66604217 on Build GRCh38
rs1035390201 Chr.16: 66604136-66604136 on Build GRCh38
rs903022546 Chr.16: 66604080-66604080 on Build GRCh38
rs1034435944 Chr.16: 66603969-66603969 on Build GRCh38
rs941614746 Chr.16: 66604417-66604417 on Build GRCh38
rs910095037 Chr.16: 66604402-66604402 on Build GRCh38
rs182253952 Chr.16: 66604396-66604396 on Build GRCh38
rs965801858 Chr.16: 66604282-66604282 on Build GRCh38
rs903173763 Chr.16: 66604161-66604161 on Build GRCh38
rs146585124 Chr.16: 66604316-66604316 on Build GRCh38
rs985949541 Chr.16: 66604208-66604208 on Build GRCh38
rs903138588 Chr.16: 66604337-66604337 on Build GRCh38
rs1022718680 Chr.16: 66604413-66604413 on Build GRCh38
rs368774831 Chr.16: 66604416-66604416 on Build GRCh38
rs549444133 Chr.16: 66604142-66604142 on Build GRCh38
rs974693291 Chr.16: 66604153-66604153 on Build GRCh38
rs1047173461 Chr.16: 66604019-66604019 on Build GRCh38
rs113755360 Chr.16: 66604269-66604269 on Build GRCh38
rs112822114 Chr.16: 66604127-66604134 on Build GRCh38
rs34561974 Chr.16: 66604283-66604284 on Build GRCh38
rs538339836 Chr.16: 66604169-66604169 on Build GRCh38
rs72790434 Chr.16: 66604051-66604051 on Build GRCh38
rs770714336 Chr.16: 66604141-66604142 on Build GRCh38
rs950379884 Chr.16: 66604324-66604324 on Build GRCh38
rs554156501 Chr.16: 66604279-66604279 on Build GRCh38
rs117010956 Chr.16: 66604049-66604049 on Build GRCh38
rs562484398 Chr.16: 66603999-66603999 on Build GRCh38
rs894586127 Chr.16: 66603970-66603970 on Build GRCh38
rs75704624 Chr.16: 66604189-66604189 on Build GRCh38
rs959807115 Chr.16: 66604138-66604138 on Build GRCh38
rs373830459 Chr.16: 66604126-66604127 on Build GRCh38
rs963086633 Chr.16: 66604145-66604145 on Build GRCh38
rs117750000 Chr.16: 66604047-66604047 on Build GRCh38
rs139365786 Chr.16: 66604187-66604187 on Build GRCh38

View Full Product Details