Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75631061-75631569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KARS
Entrez Gene ID
3735
Gene Name
lysyl-tRNA synthetase
Gene Aliases
CMTRIB, DFNB89, KARS1, KARS2, KRS
UniGene
Hs.3100
Gene Chromosome Location
Chr.16: 75627724-75647687 on Build GRCh38
Transcripts
NM_001130089.1 NM_005548.2 XM_017023217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5627068 c.1130G>A p.G377D Chr.16: 75631538-75631538 on Build GRCh38
5016524 c.1311delA p.E438fs*45 Chr.16: 75631441-75631441 on Build GRCh38
1379827 c.1351C>A p.L451I Chr.16: 75631239-75631239 on Build GRCh38
5016522 c.1314_1323del10 p.E438fs*42 Chr.16: 75631429-75631438 on Build GRCh38
5706225 c.1217_1219delTGA p.M406delM Chr.16: 75631449-75631451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764308156 Chr.16: 75631231-75631233 on Build GRCh38
rs762426497 Chr.16: 75631158-75631158 on Build GRCh38
rs374965622 Chr.16: 75631256-75631256 on Build GRCh38
rs764289340 Chr.16: 75631283-75631283 on Build GRCh38
rs754707116 Chr.16: 75631488-75631488 on Build GRCh38
rs3214635 Chr.16: 75631330-75631330 on Build GRCh38
rs763304502 Chr.16: 75631395-75631395 on Build GRCh38
rs7189895 Chr.16: 75631121-75631121 on Build GRCh38
rs773950858 Chr.16: 75631381-75631381 on Build GRCh38
rs1046648952 Chr.16: 75631094-75631094 on Build GRCh38
rs1000526670 Chr.16: 75631500-75631500 on Build GRCh38
rs749026592 Chr.16: 75631372-75631372 on Build GRCh38
rs746253005 Chr.16: 75631296-75631296 on Build GRCh38
rs770990869 Chr.16: 75631499-75631499 on Build GRCh38
rs918577830 Chr.16: 75631379-75631379 on Build GRCh38
rs776157629 Chr.16: 75631144-75631144 on Build GRCh38
rs774397948 Chr.16: 75631517-75631517 on Build GRCh38
rs761209727 Chr.16: 75631547-75631547 on Build GRCh38
rs571239491 Chr.16: 75631112-75631112 on Build GRCh38
rs145047133 Chr.16: 75631519-75631519 on Build GRCh38
rs201957065 Chr.16: 75631156-75631157 on Build GRCh38
rs775019458 Chr.16: 75631157-75631157 on Build GRCh38
rs747369627 Chr.16: 75631130-75631130 on Build GRCh38
rs751711802 Chr.16: 75631407-75631407 on Build GRCh38
rs201944195 Chr.16: 75631453-75631453 on Build GRCh38
rs776532898 Chr.16: 75631505-75631505 on Build GRCh38
rs370244075 Chr.16: 75631493-75631493 on Build GRCh38
rs773240290 Chr.16: 75631388-75631388 on Build GRCh38
rs768189185 Chr.16: 75631376-75631376 on Build GRCh38
rs146588850 Chr.16: 75631208-75631208 on Build GRCh38
rs758468945 Chr.16: 75631219-75631219 on Build GRCh38
rs371668969 Chr.16: 75631228-75631228 on Build GRCh38
rs368077299 Chr.16: 75631411-75631411 on Build GRCh38
rs753257579 Chr.16: 75631311-75631311 on Build GRCh38
rs543972962 Chr.16: 75631338-75631338 on Build GRCh38
rs369748685 Chr.16: 75631390-75631390 on Build GRCh38
rs779257362 Chr.16: 75631204-75631204 on Build GRCh38
rs190187794 Chr.16: 75631392-75631392 on Build GRCh38
rs369043605 Chr.16: 75631127-75631127 on Build GRCh38
rs369283454 Chr.16: 75631481-75631481 on Build GRCh38
rs918862686 Chr.16: 75631101-75631101 on Build GRCh38
rs771256082 Chr.16: 75631134-75631134 on Build GRCh38
rs374802658 Chr.16: 75631455-75631455 on Build GRCh38
rs779685501 Chr.16: 75631303-75631303 on Build GRCh38
rs942859264 Chr.16: 75631477-75631477 on Build GRCh38
rs747004598 Chr.16: 75631498-75631498 on Build GRCh38
rs201574004 Chr.16: 75631247-75631247 on Build GRCh38
rs964516917 Chr.16: 75631193-75631193 on Build GRCh38
rs113570860 Chr.16: 75631435-75631435 on Build GRCh38
rs201192725 Chr.16: 75631430-75631430 on Build GRCh38
rs758417518 Chr.16: 75631282-75631282 on Build GRCh38
rs900859529 Chr.16: 75631162-75631162 on Build GRCh38
rs751474072 Chr.16: 75631171-75631171 on Build GRCh38
rs982105596 Chr.16: 75631356-75631356 on Build GRCh38
rs560091813 Chr.16: 75631507-75631507 on Build GRCh38
rs760021784 Chr.16: 75631495-75631495 on Build GRCh38
rs77573084 Chr.16: 75631248-75631248 on Build GRCh38
rs749584191 Chr.16: 75631487-75631487 on Build GRCh38
rs764194013 Chr.16: 75631406-75631406 on Build GRCh38
rs761527468 Chr.16: 75631194-75631194 on Build GRCh38
rs756599606 Chr.16: 75631464-75631464 on Build GRCh38
rs771325159 Chr.16: 75631229-75631229 on Build GRCh38
rs772155670 Chr.16: 75631387-75631387 on Build GRCh38
rs761083503 Chr.16: 75631222-75631223 on Build GRCh38
rs398078761 Chr.16: 75631331-75631331 on Build GRCh38
rs750240137 Chr.16: 75631200-75631200 on Build GRCh38
rs747586573 Chr.16: 75631382-75631382 on Build GRCh38
rs867340865 Chr.16: 75631494-75631494 on Build GRCh38
rs759241436 Chr.16: 75631152-75631152 on Build GRCh38
rs753532467 Chr.16: 75631263-75631263 on Build GRCh38
rs1049079642 Chr.16: 75631109-75631109 on Build GRCh38
rs756429793 Chr.16: 75631297-75631297 on Build GRCh38
rs149772470 Chr.16: 75631490-75631490 on Build GRCh38
rs576143208 Chr.16: 75631337-75631337 on Build GRCh38
rs1037782218 Chr.16: 75631309-75631310 on Build GRCh38
rs746198033 Chr.16: 75631235-75631235 on Build GRCh38
rs779203788 Chr.16: 75631118-75631118 on Build GRCh38
rs755292539 Chr.16: 75631202-75631202 on Build GRCh38
rs201255570 Chr.16: 75631330-75631330 on Build GRCh38
rs761886340 Chr.16: 75631518-75631518 on Build GRCh38
rs367724914 Chr.16: 75631212-75631212 on Build GRCh38
rs769429317 Chr.16: 75631155-75631155 on Build GRCh38
rs886052328 Chr.16: 75631419-75631419 on Build GRCh38
rs762481405 Chr.16: 75631241-75631241 on Build GRCh38
rs767405343 Chr.16: 75631198-75631198 on Build GRCh38
rs7187847 Chr.16: 75631107-75631107 on Build GRCh38
rs748210287 Chr.16: 75631120-75631120 on Build GRCh38
rs3211301 Chr.16: 75631525-75631525 on Build GRCh38
rs773192982 Chr.16: 75631250-75631250 on Build GRCh38
rs766215239 Chr.16: 75631391-75631391 on Build GRCh38
rs780462719 Chr.16: 75631484-75631484 on Build GRCh38
rs554125088 Chr.16: 75631244-75631244 on Build GRCh38
rs397734981 Chr.16: 75631332-75631332 on Build GRCh38
rs532040317 Chr.16: 75631510-75631510 on Build GRCh38
rs754928857 Chr.16: 75631108-75631108 on Build GRCh38
rs765591392 Chr.16: 75631257-75631257 on Build GRCh38
rs768155959 Chr.16: 75631242-75631242 on Build GRCh38
rs757236374 Chr.16: 75631432-75631432 on Build GRCh38
rs768044754 Chr.16: 75631436-75631436 on Build GRCh38
rs763960268 Chr.16: 75631159-75631159 on Build GRCh38
rs779241883 Chr.16: 75631231-75631231 on Build GRCh38
rs368293521 Chr.16: 75631258-75631258 on Build GRCh38
rs778142378 Chr.16: 75631220-75631220 on Build GRCh38
rs997876680 Chr.16: 75631221-75631221 on Build GRCh38
rs752904090 Chr.16: 75631278-75631278 on Build GRCh38
rs769484182 Chr.16: 75631236-75631236 on Build GRCh38
rs556271072 Chr.16: 75631324-75631324 on Build GRCh38
rs970767608 Chr.16: 75631332-75631332 on Build GRCh38
rs748887124 Chr.16: 75631266-75631266 on Build GRCh38
rs750496729 Chr.16: 75631546-75631546 on Build GRCh38
rs751583623 Chr.16: 75631292-75631292 on Build GRCh38
rs548817586 Chr.16: 75631111-75631111 on Build GRCh38
rs200070851 Chr.16: 75631439-75631439 on Build GRCh38
rs372028053 Chr.16: 75631294-75631294 on Build GRCh38

View Full Product Details