Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3640102-3640595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNS
Entrez Gene ID
1497
Gene Name
cystinosin, lysosomal cystine transporter
Gene Aliases
CTNS-LSB, PQLC4
UniGene
Hs.187667
Gene Chromosome Location
Chr.17: 3636391-3663103 on Build GRCh38
Transcripts
NM_001031681.2 NM_004937.2 XM_005256485.2 XM_006721463.2 XM_006721464.2 XM_011523691.2 XM_011523692.2 XM_017024254.1 XM_017024255.1 XM_017024256.1 XM_017024257.1 XM_017024258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1382833 c.16C>T p.L6L Chr.17: 3640222-3640222 on Build GRCh38
5852307 c.32delT p.L14fs*1 Chr.17: 3640238-3640238 on Build GRCh38
1382834 c.16C>T p.L6L Chr.17: 3640222-3640222 on Build GRCh38
5852308 c.32delT p.L14fs*1 Chr.17: 3640238-3640238 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540162864 Chr.17: 3640482-3640482 on Build GRCh38
rs1036572322 Chr.17: 3640529-3640529 on Build GRCh38
rs927622961 Chr.17: 3640495-3640495 on Build GRCh38
rs760604266 Chr.17: 3640316-3640316 on Build GRCh38
rs971499213 Chr.17: 3640428-3640428 on Build GRCh38
rs1000814886 Chr.17: 3640366-3640366 on Build GRCh38
rs1030872379 Chr.17: 3640353-3640353 on Build GRCh38
rs1012958252 Chr.17: 3640420-3640420 on Build GRCh38
rs777799893 Chr.17: 3640234-3640234 on Build GRCh38
rs762229525 Chr.17: 3640163-3640166 on Build GRCh38
rs899334148 Chr.17: 3640126-3640126 on Build GRCh38
rs771153117 Chr.17: 3640277-3640277 on Build GRCh38
rs903368768 Chr.17: 3640254-3640254 on Build GRCh38
rs377004742 Chr.17: 3640530-3640530 on Build GRCh38
rs753269553 Chr.17: 3640308-3640308 on Build GRCh38
rs982907254 Chr.17: 3640442-3640442 on Build GRCh38
rs765523702 Chr.17: 3640183-3640183 on Build GRCh38
rs545580222 Chr.17: 3640153-3640153 on Build GRCh38
rs786204501 Chr.17: 3640224-3640227 on Build GRCh38
rs766774059 Chr.17: 3640173-3640173 on Build GRCh38
rs111824707 Chr.17: 3640134-3640134 on Build GRCh38
rs930813517 Chr.17: 3640562-3640562 on Build GRCh38
rs754092691 Chr.17: 3640235-3640235 on Build GRCh38
rs1050586419 Chr.17: 3640565-3640565 on Build GRCh38
rs374068354 Chr.17: 3640286-3640286 on Build GRCh38
rs556689070 Chr.17: 3640519-3640519 on Build GRCh38
rs567999441 Chr.17: 3640253-3640253 on Build GRCh38
rs758995279 Chr.17: 3640227-3640228 on Build GRCh38
rs762537406 Chr.17: 3640146-3640146 on Build GRCh38
rs779919175 Chr.17: 3640238-3640238 on Build GRCh38
rs762701585 Chr.17: 3640274-3640274 on Build GRCh38
rs889215917 Chr.17: 3640567-3640567 on Build GRCh38
rs768188337 Chr.17: 3640211-3640211 on Build GRCh38
rs550375830 Chr.17: 3640218-3640218 on Build GRCh38
rs547987979 Chr.17: 3640228-3640228 on Build GRCh38
rs767289120 Chr.17: 3640269-3640269 on Build GRCh38
rs769364200 Chr.17: 3640257-3640257 on Build GRCh38
rs1024376053 Chr.17: 3640421-3640421 on Build GRCh38
rs752795299 Chr.17: 3640225-3640225 on Build GRCh38
rs921230580 Chr.17: 3640538-3640538 on Build GRCh38
rs768135137 Chr.17: 3640304-3640304 on Build GRCh38
rs1008963260 Chr.17: 3640571-3640571 on Build GRCh38
rs760019018 Chr.17: 3640287-3640287 on Build GRCh38
rs148820665 Chr.17: 3640416-3640416 on Build GRCh38
rs758087826 Chr.17: 3640237-3640237 on Build GRCh38
rs747647287 Chr.17: 3640255-3640255 on Build GRCh38
rs372701935 Chr.17: 3640199-3640199 on Build GRCh38
rs1033273522 Chr.17: 3640414-3640414 on Build GRCh38
rs940982825 Chr.17: 3640502-3640502 on Build GRCh38
rs370068809 Chr.17: 3640245-3640245 on Build GRCh38
rs1040426811 Chr.17: 3640572-3640572 on Build GRCh38
rs143424493 Chr.17: 3640561-3640561 on Build GRCh38
rs377010311 Chr.17: 3640200-3640200 on Build GRCh38
rs531411098 Chr.17: 3640226-3640226 on Build GRCh38
rs756041888 Chr.17: 3640531-3640531 on Build GRCh38
rs958107702 Chr.17: 3640388-3640388 on Build GRCh38
rs571143928 Chr.17: 3640443-3640443 on Build GRCh38
rs994972597 Chr.17: 3640163-3640163 on Build GRCh38

View Full Product Details