Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29942033-29942577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB5
Entrez Gene ID
374786
Gene Name
EF-hand calcium binding domain 5
Gene Aliases
-
UniGene
Hs.662411
Gene Chromosome Location
Chr.17: 29929856-30108452 on Build GRCh38
Transcripts
NM_001145053.1 NM_198529.3 NR_026738.1 XM_011524759.1 XM_011524763.2 XM_011524765.1 XM_011524768.2 XM_017024592.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5177113 c.63_64delAG p.R23fs*6 Chr.17: 29942260-29942261 on Build GRCh38
122781 c.95A>T p.E32V Chr.17: 29942292-29942292 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901833628 Chr.17: 29942517-29942517 on Build GRCh38
rs896223699 Chr.17: 29942332-29942332 on Build GRCh38
rs763860314 Chr.17: 29942254-29942254 on Build GRCh38
rs751355455 Chr.17: 29942272-29942272 on Build GRCh38
rs373838999 Chr.17: 29942343-29942343 on Build GRCh38
rs372931755 Chr.17: 29942536-29942536 on Build GRCh38
rs368601605 Chr.17: 29942325-29942325 on Build GRCh38
rs762261664 Chr.17: 29942453-29942454 on Build GRCh38
rs979345322 Chr.17: 29942171-29942171 on Build GRCh38
rs750563003 Chr.17: 29942309-29942309 on Build GRCh38
rs968635057 Chr.17: 29942287-29942287 on Build GRCh38
rs1018986284 Chr.17: 29942089-29942089 on Build GRCh38
rs886993553 Chr.17: 29942165-29942165 on Build GRCh38
rs950267073 Chr.17: 29942369-29942369 on Build GRCh38
rs1044974473 Chr.17: 29942404-29942404 on Build GRCh38
rs756215605 Chr.17: 29942311-29942311 on Build GRCh38
rs921895581 Chr.17: 29942136-29942136 on Build GRCh38
rs898217754 Chr.17: 29942426-29942426 on Build GRCh38
rs556094061 Chr.17: 29942158-29942158 on Build GRCh38
rs562980502 Chr.17: 29942522-29942522 on Build GRCh38
rs745958702 Chr.17: 29942193-29942193 on Build GRCh38
rs140549659 Chr.17: 29942070-29942070 on Build GRCh38
rs533072180 Chr.17: 29942478-29942478 on Build GRCh38
rs974968719 Chr.17: 29942132-29942132 on Build GRCh38
rs775335897 Chr.17: 29942229-29942229 on Build GRCh38
rs769199820 Chr.17: 29942518-29942518 on Build GRCh38
rs887065478 Chr.17: 29942462-29942462 on Build GRCh38
rs772153164 Chr.17: 29942200-29942200 on Build GRCh38
rs541623524 Chr.17: 29942401-29942401 on Build GRCh38
rs917573555 Chr.17: 29942385-29942385 on Build GRCh38
rs770561941 Chr.17: 29942192-29942192 on Build GRCh38
rs771595921 Chr.17: 29942270-29942271 on Build GRCh38
rs1008091986 Chr.17: 29942471-29942471 on Build GRCh38
rs186217060 Chr.17: 29942175-29942175 on Build GRCh38
rs991856974 Chr.17: 29942378-29942378 on Build GRCh38
rs1029053645 Chr.17: 29942255-29942255 on Build GRCh38
rs947806572 Chr.17: 29942415-29942415 on Build GRCh38
rs995330136 Chr.17: 29942072-29942072 on Build GRCh38
rs545629214 Chr.17: 29942358-29942358 on Build GRCh38
rs761187614 Chr.17: 29942490-29942490 on Build GRCh38
rs965886724 Chr.17: 29942115-29942115 on Build GRCh38
rs995502945 Chr.17: 29942451-29942451 on Build GRCh38
rs576488314 Chr.17: 29942061-29942061 on Build GRCh38
rs79411908 Chr.17: 29942425-29942425 on Build GRCh38
rs749618405 Chr.17: 29942258-29942259 on Build GRCh38
rs544759519 Chr.17: 29942509-29942509 on Build GRCh38
rs868759538 Chr.17: 29942545-29942545 on Build GRCh38
rs767781292 Chr.17: 29942307-29942307 on Build GRCh38
rs757566302 Chr.17: 29942293-29942293 on Build GRCh38
rs765178630 Chr.17: 29942220-29942220 on Build GRCh38
rs773721116 Chr.17: 29942250-29942252 on Build GRCh38
rs530418026 Chr.17: 29942525-29942525 on Build GRCh38

View Full Product Details