Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19744878-19745382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3A1
Entrez Gene ID
218
Gene Name
aldehyde dehydrogenase 3 family member A1
Gene Aliases
ALDH3, ALDHIII
UniGene
Hs.531682
Gene Chromosome Location
Chr.17: 19737984-19748433 on Build GRCh38
Transcripts
NM_000691.4 NM_001135167.1 NM_001135168.1 XM_005256522.1 XM_005256523.2 XM_005256524.3 XM_011523730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415367 c.56G>T p.R19M Chr.17: 19745074-19745074 on Build GRCh38
7084683 c.9G>A p.K3K Chr.17: 19745121-19745121 on Build GRCh38
6404194 c.125A>T p.Q42L Chr.17: 19745005-19745005 on Build GRCh38
5852017 c.65C>T p.P22L Chr.17: 19745065-19745065 on Build GRCh38
6598258 c.26A>G p.K9R Chr.17: 19745104-19745104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368233404 Chr.17: 19745146-19745146 on Build GRCh38
rs200717464 Chr.17: 19745113-19745113 on Build GRCh38
rs980893840 Chr.17: 19745268-19745268 on Build GRCh38
rs201504992 Chr.17: 19745036-19745036 on Build GRCh38
rs769768762 Chr.17: 19745044-19745044 on Build GRCh38
rs780939059 Chr.17: 19745108-19745108 on Build GRCh38
rs112422668 Chr.17: 19744904-19744904 on Build GRCh38
rs936262526 Chr.17: 19744908-19744908 on Build GRCh38
rs763570276 Chr.17: 19745093-19745093 on Build GRCh38
rs60032532 Chr.17: 19744912-19744913 on Build GRCh38
rs118057522 Chr.17: 19745287-19745287 on Build GRCh38
rs747724829 Chr.17: 19744947-19744947 on Build GRCh38
rs745863174 Chr.17: 19744970-19744970 on Build GRCh38
rs749245878 Chr.17: 19745116-19745116 on Build GRCh38
rs901659474 Chr.17: 19744935-19744935 on Build GRCh38
rs116754663 Chr.17: 19744989-19744989 on Build GRCh38
rs570805569 Chr.17: 19745361-19745361 on Build GRCh38
rs147298045 Chr.17: 19745102-19745102 on Build GRCh38
rs777181749 Chr.17: 19745124-19745124 on Build GRCh38
rs957565918 Chr.17: 19745245-19745245 on Build GRCh38
rs749325435 Chr.17: 19745054-19745054 on Build GRCh38
rs762980938 Chr.17: 19744925-19744928 on Build GRCh38
rs1029380751 Chr.17: 19744999-19744999 on Build GRCh38
rs966391010 Chr.17: 19745195-19745195 on Build GRCh38
rs201999351 Chr.17: 19745148-19745148 on Build GRCh38
rs939868677 Chr.17: 19744971-19744971 on Build GRCh38
rs4646789 Chr.17: 19745316-19745316 on Build GRCh38
rs756727090 Chr.17: 19745009-19745009 on Build GRCh38
rs757597237 Chr.17: 19744920-19744920 on Build GRCh38
rs750984975 Chr.17: 19744923-19744923 on Build GRCh38
rs1000858181 Chr.17: 19745177-19745177 on Build GRCh38
rs544620393 Chr.17: 19745011-19745011 on Build GRCh38
rs765286453 Chr.17: 19744920-19744921 on Build GRCh38
rs777415702 Chr.17: 19745023-19745023 on Build GRCh38
rs924835465 Chr.17: 19744907-19744908 on Build GRCh38
rs778904408 Chr.17: 19745117-19745117 on Build GRCh38
rs892746174 Chr.17: 19745227-19745227 on Build GRCh38
rs1032556384 Chr.17: 19745194-19745194 on Build GRCh38
rs910514485 Chr.17: 19745347-19745347 on Build GRCh38
rs997514261 Chr.17: 19744991-19744991 on Build GRCh38
rs59720526 Chr.17: 19744958-19744958 on Build GRCh38
rs1040928388 Chr.17: 19745138-19745138 on Build GRCh38
rs760027682 Chr.17: 19745127-19745127 on Build GRCh38
rs772372375 Chr.17: 19745119-19745119 on Build GRCh38
rs771212258 Chr.17: 19745121-19745121 on Build GRCh38
rs771807870 Chr.17: 19744950-19744950 on Build GRCh38
rs1040889947 Chr.17: 19744931-19744931 on Build GRCh38
rs539635463 Chr.17: 19744907-19744907 on Build GRCh38
rs754595647 Chr.17: 19744999-19745047 on Build GRCh38
rs745610092 Chr.17: 19745109-19745109 on Build GRCh38
rs970921427 Chr.17: 19745255-19745255 on Build GRCh38
rs769856693 Chr.17: 19744972-19744972 on Build GRCh38
rs768634987 Chr.17: 19745064-19745064 on Build GRCh38
rs370178415 Chr.17: 19744901-19744902 on Build GRCh38
rs781738420 Chr.17: 19744921-19744921 on Build GRCh38
rs939817656 Chr.17: 19745132-19745132 on Build GRCh38
rs917410422 Chr.17: 19744903-19744903 on Build GRCh38
rs201253459 Chr.17: 19744905-19744906 on Build GRCh38
rs923270180 Chr.17: 19745308-19745308 on Build GRCh38
rs201273273 Chr.17: 19744954-19744954 on Build GRCh38
rs922390051 Chr.17: 19745339-19745339 on Build GRCh38
rs750078082 Chr.17: 19745135-19745135 on Build GRCh38
rs889083010 Chr.17: 19745170-19745170 on Build GRCh38
rs745769411 Chr.17: 19745039-19745039 on Build GRCh38
rs772284661 Chr.17: 19745171-19745171 on Build GRCh38
rs908446700 Chr.17: 19745302-19745302 on Build GRCh38
rs374241717 Chr.17: 19745129-19745129 on Build GRCh38
rs767162387 Chr.17: 19744999-19745000 on Build GRCh38
rs1047872574 Chr.17: 19745317-19745317 on Build GRCh38
rs910678957 Chr.17: 19745355-19745355 on Build GRCh38
rs747290788 Chr.17: 19745175-19745175 on Build GRCh38
rs754303573 Chr.17: 19745084-19745084 on Build GRCh38
rs551002574 Chr.17: 19745350-19745350 on Build GRCh38
rs56234923 Chr.17: 19744906-19744906 on Build GRCh38
rs746652504 Chr.17: 19745024-19745024 on Build GRCh38
rs533309462 Chr.17: 19745090-19745090 on Build GRCh38
rs750965150 Chr.17: 19745004-19745004 on Build GRCh38
rs887643470 Chr.17: 19745133-19745133 on Build GRCh38
rs181118535 Chr.17: 19745279-19745279 on Build GRCh38
rs754920415 Chr.17: 19745151-19745151 on Build GRCh38
rs773761996 Chr.17: 19745079-19745079 on Build GRCh38
rs1006129315 Chr.17: 19745140-19745140 on Build GRCh38
rs1054343079 Chr.17: 19744910-19744911 on Build GRCh38
rs777314022 Chr.17: 19744951-19744951 on Build GRCh38
rs748200146 Chr.17: 19745155-19745155 on Build GRCh38
rs199787983 Chr.17: 19744906-19744906 on Build GRCh38
rs773324314 Chr.17: 19744920-19744928 on Build GRCh38
rs1025539193 Chr.17: 19745231-19745231 on Build GRCh38
rs185327398 Chr.17: 19745359-19745359 on Build GRCh38
rs754526583 Chr.17: 19744932-19744932 on Build GRCh38
rs757670168 Chr.17: 19745137-19745143 on Build GRCh38
rs767114108 Chr.17: 19745134-19745134 on Build GRCh38
rs778633741 Chr.17: 19744942-19744942 on Build GRCh38
rs754226704 Chr.17: 19745104-19745104 on Build GRCh38
rs1042172 Chr.17: 19745095-19745095 on Build GRCh38
rs371701018 Chr.17: 19745149-19745149 on Build GRCh38
rs766871933 Chr.17: 19745083-19745083 on Build GRCh38
rs999268048 Chr.17: 19745207-19745207 on Build GRCh38
rs374854383 Chr.17: 19745152-19745152 on Build GRCh38
rs765863902 Chr.17: 19744970-19744971 on Build GRCh38
rs757032351 Chr.17: 19745033-19745033 on Build GRCh38
rs1033467902 Chr.17: 19745278-19745278 on Build GRCh38
rs943461245 Chr.17: 19744912-19744912 on Build GRCh38
rs761145572 Chr.17: 19745081-19745081 on Build GRCh38
rs751025286 Chr.17: 19744996-19744997 on Build GRCh38
rs1009785789 Chr.17: 19745228-19745228 on Build GRCh38
rs751220464 Chr.17: 19745094-19745094 on Build GRCh38
rs774623416 Chr.17: 19745068-19745068 on Build GRCh38
rs950286323 Chr.17: 19744903-19744904 on Build GRCh38
rs778081851 Chr.17: 19745172-19745172 on Build GRCh38
rs889157210 Chr.17: 19744910-19744910 on Build GRCh38
rs57485106 Chr.17: 19745003-19745003 on Build GRCh38
rs779986721 Chr.17: 19745115-19745115 on Build GRCh38
rs762182523 Chr.17: 19745001-19745001 on Build GRCh38
rs976684446 Chr.17: 19745327-19745327 on Build GRCh38
rs562444690 Chr.17: 19745300-19745300 on Build GRCh38
rs776492802 Chr.17: 19744959-19744959 on Build GRCh38
rs763629080 Chr.17: 19745128-19745128 on Build GRCh38
rs1050933415 Chr.17: 19745354-19745354 on Build GRCh38
rs542345176 Chr.17: 19744983-19744983 on Build GRCh38
rs774444883 Chr.17: 19744993-19744993 on Build GRCh38
rs761055890 Chr.17: 19745120-19745120 on Build GRCh38
rs895081305 Chr.17: 19745150-19745150 on Build GRCh38

View Full Product Details