Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45150349-45150850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXIM1
Entrez Gene ID
10614
Gene Name
hexamethylene bisacetamide inducible 1
Gene Aliases
CLP1, EDG1, HIS1, MAQ1
UniGene
Hs.586945
Gene Chromosome Location
Chr.17: 45147317-45152101 on Build GRCh38
Transcripts
NM_006460.2

SNP Detail

SNP ID Chromosome Location
rs545300118 Chr.17: 45150642-45150642 on Build GRCh38
rs942039837 Chr.17: 45150545-45150550 on Build GRCh38
rs1008745099 Chr.17: 45150494-45150494 on Build GRCh38
rs906155592 Chr.17: 45150602-45150604 on Build GRCh38
rs1801920 Chr.17: 45150784-45150784 on Build GRCh38
rs1032282796 Chr.17: 45150471-45150471 on Build GRCh38
rs750471057 Chr.17: 45150544-45150544 on Build GRCh38
rs763246975 Chr.17: 45150698-45150698 on Build GRCh38
rs1801923 Chr.17: 45150751-45150751 on Build GRCh38
rs891231675 Chr.17: 45150489-45150489 on Build GRCh38
rs76559206 Chr.17: 45150406-45150406 on Build GRCh38
rs370469173 Chr.17: 45150731-45150731 on Build GRCh38
rs377439069 Chr.17: 45150481-45150481 on Build GRCh38
rs995665715 Chr.17: 45150744-45150744 on Build GRCh38
rs949911513 Chr.17: 45150392-45150392 on Build GRCh38
rs971148449 Chr.17: 45150633-45150633 on Build GRCh38
rs1054376092 Chr.17: 45150420-45150420 on Build GRCh38
rs78243847 Chr.17: 45150399-45150400 on Build GRCh38
rs1037081940 Chr.17: 45150720-45150720 on Build GRCh38
rs923104639 Chr.17: 45150501-45150501 on Build GRCh38
rs951709813 Chr.17: 45150592-45150592 on Build GRCh38
rs907931115 Chr.17: 45150538-45150538 on Build GRCh38
rs560476592 Chr.17: 45150652-45150652 on Build GRCh38
rs934529204 Chr.17: 45150408-45150408 on Build GRCh38
rs915627231 Chr.17: 45150621-45150621 on Build GRCh38
rs200646346 Chr.17: 45150407-45150407 on Build GRCh38
rs1045593702 Chr.17: 45150395-45150395 on Build GRCh38
rs897327649 Chr.17: 45150739-45150739 on Build GRCh38
rs907982976 Chr.17: 45150816-45150816 on Build GRCh38
rs752698270 Chr.17: 45150406-45150407 on Build GRCh38
rs369748312 Chr.17: 45150724-45150724 on Build GRCh38
rs893139064 Chr.17: 45150433-45150433 on Build GRCh38
rs548392817 Chr.17: 45150680-45150680 on Build GRCh38
rs547733345 Chr.17: 45150469-45150469 on Build GRCh38
rs563974577 Chr.17: 45150397-45150397 on Build GRCh38
rs1032414356 Chr.17: 45150727-45150727 on Build GRCh38
rs566367119 Chr.17: 45150500-45150500 on Build GRCh38
rs71764432 Chr.17: 45150400-45150401 on Build GRCh38
rs964375115 Chr.17: 45150773-45150773 on Build GRCh38
rs761816311 Chr.17: 45150403-45150407 on Build GRCh38
rs954540384 Chr.17: 45150789-45150789 on Build GRCh38
rs71136073 Chr.17: 45150405-45150406 on Build GRCh38
rs944101281 Chr.17: 45150711-45150711 on Build GRCh38
rs1020687402 Chr.17: 45150681-45150681 on Build GRCh38
rs1801921 Chr.17: 45150822-45150822 on Build GRCh38
rs990055896 Chr.17: 45150675-45150675 on Build GRCh38
rs114205350 Chr.17: 45150523-45150523 on Build GRCh38
rs11558626 Chr.17: 45150470-45150470 on Build GRCh38
rs961534715 Chr.17: 45150508-45150508 on Build GRCh38
rs983271320 Chr.17: 45150803-45150803 on Build GRCh38
rs996890706 Chr.17: 45150458-45150458 on Build GRCh38
rs995678604 Chr.17: 45150518-45150518 on Build GRCh38
rs932625172 Chr.17: 45150687-45150687 on Build GRCh38
rs973464517 Chr.17: 45150823-45150823 on Build GRCh38
rs1030224175 Chr.17: 45150786-45150786 on Build GRCh38
rs386386160 Chr.17: 45150402-45150403 on Build GRCh38
rs545797773 Chr.17: 45150374-45150374 on Build GRCh38
rs891132261 Chr.17: 45150706-45150706 on Build GRCh38
rs1000524639 Chr.17: 45150713-45150713 on Build GRCh38
rs981613123 Chr.17: 45150639-45150639 on Build GRCh38
rs991200037 Chr.17: 45150593-45150593 on Build GRCh38
rs934519099 Chr.17: 45150648-45150648 on Build GRCh38
rs927296562 Chr.17: 45150825-45150825 on Build GRCh38
rs200818670 Chr.17: 45150653-45150654 on Build GRCh38
rs983554202 Chr.17: 45150525-45150525 on Build GRCh38

View Full Product Details