Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55315141-55315650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLF
Entrez Gene ID
3131
Gene Name
HLF, PAR bZIP transcription factor
Gene Aliases
-
UniGene
Hs.196952
Gene Chromosome Location
Chr.17: 55264960-55325065 on Build GRCh38
Transcripts
NM_002126.4 XM_005257269.2 XM_011524705.1 XM_017024556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764615 c.661G>A p.D221N Chr.17: 55315436-55315436 on Build GRCh38
6680865 c.520G>A p.G174S Chr.17: 55315295-55315295 on Build GRCh38
360829 c.472C>T p.R158C Chr.17: 55315247-55315247 on Build GRCh38
74928 c.586C>G p.R196G Chr.17: 55315361-55315361 on Build GRCh38
6298259 c.528G>T p.E176D Chr.17: 55315303-55315303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768509008 Chr.17: 55315295-55315295 on Build GRCh38
rs773037201 Chr.17: 55315488-55315488 on Build GRCh38
rs771695827 Chr.17: 55315248-55315248 on Build GRCh38
rs1048154091 Chr.17: 55315234-55315234 on Build GRCh38
rs745656628 Chr.17: 55315245-55315245 on Build GRCh38
rs745584891 Chr.17: 55315246-55315246 on Build GRCh38
rs750441913 Chr.17: 55315178-55315178 on Build GRCh38
rs1035259397 Chr.17: 55315560-55315560 on Build GRCh38
rs140685489 Chr.17: 55315367-55315367 on Build GRCh38
rs200972587 Chr.17: 55315212-55315212 on Build GRCh38
rs778339604 Chr.17: 55315378-55315378 on Build GRCh38
rs2287222 Chr.17: 55315183-55315183 on Build GRCh38
rs1040249035 Chr.17: 55315247-55315247 on Build GRCh38
rs751630445 Chr.17: 55315350-55315350 on Build GRCh38
rs3841527 Chr.17: 55315548-55315548 on Build GRCh38
rs75163131 Chr.17: 55315435-55315435 on Build GRCh38
rs938143926 Chr.17: 55315191-55315191 on Build GRCh38
rs532268866 Chr.17: 55315540-55315540 on Build GRCh38
rs779581406 Chr.17: 55315408-55315408 on Build GRCh38
rs752165259 Chr.17: 55315512-55315512 on Build GRCh38
rs544831113 Chr.17: 55315169-55315169 on Build GRCh38
rs770657492 Chr.17: 55315231-55315231 on Build GRCh38
rs374894572 Chr.17: 55315351-55315351 on Build GRCh38
rs765374641 Chr.17: 55315317-55315317 on Build GRCh38
rs780171226 Chr.17: 55315184-55315184 on Build GRCh38
rs886449160 Chr.17: 55315240-55315240 on Build GRCh38
rs900454568 Chr.17: 55315309-55315309 on Build GRCh38
rs746525611 Chr.17: 55315418-55315418 on Build GRCh38
rs746924145 Chr.17: 55315411-55315411 on Build GRCh38
rs865981796 Chr.17: 55315377-55315377 on Build GRCh38
rs993123218 Chr.17: 55315327-55315327 on Build GRCh38
rs2287223 Chr.17: 55315491-55315491 on Build GRCh38
rs760633766 Chr.17: 55315286-55315286 on Build GRCh38
rs371976247 Chr.17: 55315349-55315349 on Build GRCh38
rs755486098 Chr.17: 55315193-55315193 on Build GRCh38
rs371704687 Chr.17: 55315177-55315177 on Build GRCh38
rs561233390 Chr.17: 55315403-55315403 on Build GRCh38
rs776441842 Chr.17: 55315296-55315296 on Build GRCh38
rs761802214 Chr.17: 55315480-55315480 on Build GRCh38
rs200005618 Chr.17: 55315550-55315550 on Build GRCh38
rs1007186929 Chr.17: 55315617-55315617 on Build GRCh38
rs369313188 Chr.17: 55315366-55315366 on Build GRCh38
rs756544740 Chr.17: 55315368-55315368 on Build GRCh38
rs371745405 Chr.17: 55315375-55315375 on Build GRCh38
rs1465353 Chr.17: 55315551-55315551 on Build GRCh38
rs1051706566 Chr.17: 55315577-55315577 on Build GRCh38
rs930687914 Chr.17: 55315592-55315592 on Build GRCh38
rs71385382 Chr.17: 55315470-55315470 on Build GRCh38
rs189391743 Chr.17: 55315610-55315610 on Build GRCh38
rs781589675 Chr.17: 55315210-55315210 on Build GRCh38
rs375544785 Chr.17: 55315241-55315241 on Build GRCh38
rs991372886 Chr.17: 55315565-55315565 on Build GRCh38
rs745494728 Chr.17: 55315402-55315402 on Build GRCh38
rs959625625 Chr.17: 55315561-55315561 on Build GRCh38
rs368230625 Chr.17: 55315334-55315334 on Build GRCh38
rs920776157 Chr.17: 55315571-55315571 on Build GRCh38
rs755470158 Chr.17: 55315352-55315352 on Build GRCh38
rs769561073 Chr.17: 55315481-55315481 on Build GRCh38
rs971051846 Chr.17: 55315545-55315545 on Build GRCh38
rs774912373 Chr.17: 55315278-55315278 on Build GRCh38
rs368814378 Chr.17: 55315192-55315192 on Build GRCh38
rs750635452 Chr.17: 55315324-55315324 on Build GRCh38
rs758242519 Chr.17: 55315361-55315361 on Build GRCh38
rs761523454 Chr.17: 55315314-55315314 on Build GRCh38
rs983553333 Chr.17: 55315600-55315600 on Build GRCh38

View Full Product Details