Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58304914-58305412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPOAP1
Entrez Gene ID
9256
Gene Name
TSPO associated protein 1
Gene Aliases
BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
UniGene
Hs.112499
Gene Chromosome Location
Chr.17: 58301227-58329030 on Build GRCh38
Transcripts
NM_001261835.1 NM_004758.3 NM_024418.2 XM_005257807.2 XM_006722173.2 XM_006722174.2 XM_006722175.2 XM_006722176.2 XM_006722177.3 XM_006722178.3 XM_011525471.2 XM_011525472.1 XM_017025326.1 XM_017025327.1 XM_017025328.1 XM_017025329.1 XM_017025330.1 XM_017025331.1 XM_017025332.1 XM_017025333.1 XM_017025334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130440 c.5454G>A p.R1818R Chr.17: 58305151-58305151 on Build GRCh38
3520064 c.5453G>A p.R1818K Chr.17: 58305152-58305152 on Build GRCh38
3520062 c.5455G>A p.G1819S Chr.17: 58305150-58305150 on Build GRCh38
3520060 c.5475C>T p.F1825F Chr.17: 58305130-58305130 on Build GRCh38
3520058 c.5514G>A p.R1838R Chr.17: 58305091-58305091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761190766 Chr.17: 58305268-58305268 on Build GRCh38
rs772456594 Chr.17: 58305327-58305327 on Build GRCh38
rs764971219 Chr.17: 58305274-58305274 on Build GRCh38
rs536903691 Chr.17: 58304982-58304982 on Build GRCh38
rs936306924 Chr.17: 58305193-58305193 on Build GRCh38
rs773243561 Chr.17: 58305120-58305120 on Build GRCh38
rs755763398 Chr.17: 58304963-58304963 on Build GRCh38
rs780723601 Chr.17: 58305189-58305189 on Build GRCh38
rs778521172 Chr.17: 58305099-58305099 on Build GRCh38
rs760088131 Chr.17: 58305155-58305155 on Build GRCh38
rs749468730 Chr.17: 58305310-58305310 on Build GRCh38
rs768277493 Chr.17: 58305112-58305112 on Build GRCh38
rs764371374 Chr.17: 58305012-58305012 on Build GRCh38
rs573547867 Chr.17: 58305072-58305072 on Build GRCh38
rs757004736 Chr.17: 58305096-58305096 on Build GRCh38
rs763172388 Chr.17: 58305230-58305230 on Build GRCh38
rs774277267 Chr.17: 58305318-58305318 on Build GRCh38
rs776520679 Chr.17: 58305114-58305114 on Build GRCh38
rs766854785 Chr.17: 58305149-58305149 on Build GRCh38
rs779381503 Chr.17: 58305284-58305284 on Build GRCh38
rs765262192 Chr.17: 58305047-58305047 on Build GRCh38
rs748376520 Chr.17: 58305044-58305044 on Build GRCh38
rs779924056 Chr.17: 58305107-58305107 on Build GRCh38
rs749313994 Chr.17: 58305272-58305272 on Build GRCh38
rs962678137 Chr.17: 58305029-58305029 on Build GRCh38
rs759915316 Chr.17: 58305258-58305258 on Build GRCh38
rs758128452 Chr.17: 58305281-58305281 on Build GRCh38
rs770049020 Chr.17: 58305117-58305117 on Build GRCh38
rs11867412 Chr.17: 58305151-58305151 on Build GRCh38
rs755844010 Chr.17: 58305300-58305300 on Build GRCh38
rs372177919 Chr.17: 58305076-58305076 on Build GRCh38
rs776775715 Chr.17: 58305017-58305017 on Build GRCh38
rs576587495 Chr.17: 58305204-58305204 on Build GRCh38
rs368799246 Chr.17: 58305355-58305355 on Build GRCh38
rs529225806 Chr.17: 58305337-58305337 on Build GRCh38
rs543746288 Chr.17: 58305278-58305278 on Build GRCh38
rs368773487 Chr.17: 58305052-58305052 on Build GRCh38
rs754572229 Chr.17: 58305366-58305366 on Build GRCh38
rs765718958 Chr.17: 58304935-58304935 on Build GRCh38
rs773450537 Chr.17: 58305196-58305197 on Build GRCh38
rs374075771 Chr.17: 58305195-58305195 on Build GRCh38
rs760426173 Chr.17: 58305006-58305006 on Build GRCh38
rs770292905 Chr.17: 58304983-58304983 on Build GRCh38
rs202239867 Chr.17: 58305348-58305348 on Build GRCh38
rs200173933 Chr.17: 58305188-58305188 on Build GRCh38
rs750523132 Chr.17: 58305051-58305051 on Build GRCh38
rs751026038 Chr.17: 58305360-58305360 on Build GRCh38
rs72839983 Chr.17: 58304947-58304947 on Build GRCh38
rs759533839 Chr.17: 58305326-58305327 on Build GRCh38
rs754839047 Chr.17: 58305288-58305288 on Build GRCh38
rs752975910 Chr.17: 58305266-58305266 on Build GRCh38
rs749036265 Chr.17: 58305331-58305332 on Build GRCh38
rs779630628 Chr.17: 58305196-58305196 on Build GRCh38
rs201045149 Chr.17: 58305103-58305103 on Build GRCh38
rs955841097 Chr.17: 58304945-58304945 on Build GRCh38
rs762098992 Chr.17: 58305041-58305041 on Build GRCh38
rs142129538 Chr.17: 58305187-58305187 on Build GRCh38
rs777167245 Chr.17: 58305344-58305344 on Build GRCh38
rs770927643 Chr.17: 58305242-58305242 on Build GRCh38
rs377363761 Chr.17: 58305336-58305336 on Build GRCh38
rs777358825 Chr.17: 58304974-58304974 on Build GRCh38
rs777545713 Chr.17: 58305301-58305301 on Build GRCh38
rs766094721 Chr.17: 58305357-58305357 on Build GRCh38
rs768660831 Chr.17: 58305223-58305223 on Build GRCh38
rs907351257 Chr.17: 58305283-58305283 on Build GRCh38
rs781627509 Chr.17: 58305078-58305078 on Build GRCh38
rs750863788 Chr.17: 58304939-58304939 on Build GRCh38
rs771211083 Chr.17: 58305313-58305313 on Build GRCh38
rs751128800 Chr.17: 58305287-58305287 on Build GRCh38
rs747384094 Chr.17: 58304950-58304950 on Build GRCh38
rs771011592 Chr.17: 58305288-58305288 on Build GRCh38
rs1052591527 Chr.17: 58304934-58304934 on Build GRCh38
rs773477427 Chr.17: 58305229-58305229 on Build GRCh38
rs774206614 Chr.17: 58305317-58305317 on Build GRCh38
rs780825648 Chr.17: 58305224-58305224 on Build GRCh38
rs774525327 Chr.17: 58305245-58305245 on Build GRCh38
rs369935522 Chr.17: 58305374-58305374 on Build GRCh38
rs769997188 Chr.17: 58305227-58305227 on Build GRCh38
rs867869527 Chr.17: 58305219-58305219 on Build GRCh38
rs918313264 Chr.17: 58305363-58305363 on Build GRCh38
rs747922692 Chr.17: 58305293-58305293 on Build GRCh38
rs758936598 Chr.17: 58304941-58304941 on Build GRCh38
rs775401475 Chr.17: 58305001-58305001 on Build GRCh38
rs1045959926 Chr.17: 58305270-58305270 on Build GRCh38
rs746002928 Chr.17: 58305323-58305323 on Build GRCh38
rs753131731 Chr.17: 58305174-58305174 on Build GRCh38
rs773998521 Chr.17: 58304984-58304984 on Build GRCh38
rs372905504 Chr.17: 58305175-58305175 on Build GRCh38
rs767737282 Chr.17: 58305170-58305170 on Build GRCh38
rs746806807 Chr.17: 58305108-58305108 on Build GRCh38
rs772155652 Chr.17: 58304992-58304992 on Build GRCh38
rs766946300 Chr.17: 58305389-58305389 on Build GRCh38
rs1021442982 Chr.17: 58305291-58305291 on Build GRCh38
rs768121627 Chr.17: 58305259-58305259 on Build GRCh38
rs201985313 Chr.17: 58305065-58305065 on Build GRCh38
rs745756702 Chr.17: 58304991-58304991 on Build GRCh38
rs9893754 Chr.17: 58305134-58305134 on Build GRCh38
rs375624445 Chr.17: 58305102-58305102 on Build GRCh38
rs769946823 Chr.17: 58305061-58305064 on Build GRCh38
rs748829276 Chr.17: 58305081-58305081 on Build GRCh38

View Full Product Details