Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82080057-82080560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
UniGene
Hs.83190
Gene Chromosome Location
Chr.17: 82078338-82098303 on Build GRCh38
Transcripts
NM_004104.4 XM_011523538.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668084 c.7011C>T p.F2337F Chr.17: 82080406-82080406 on Build GRCh38
986027 c.7038C>A p.A2346A Chr.17: 82080379-82080379 on Build GRCh38
5620792 c.6880A>G p.R2294G Chr.17: 82080537-82080537 on Build GRCh38
460173 c.7084G>A p.E2362K Chr.17: 82080202-82080202 on Build GRCh38
5942716 c.7048-8C>T p.? Chr.17: 82080246-82080246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007673690 Chr.17: 82080507-82080507 on Build GRCh38
rs760913867 Chr.17: 82080384-82080384 on Build GRCh38
rs747420204 Chr.17: 82080484-82080484 on Build GRCh38
rs865870969 Chr.17: 82080525-82080525 on Build GRCh38
rs763770708 Chr.17: 82080285-82080285 on Build GRCh38
rs755122528 Chr.17: 82080456-82080456 on Build GRCh38
rs375551556 Chr.17: 82080114-82080114 on Build GRCh38
rs774718641 Chr.17: 82080349-82080349 on Build GRCh38
rs767046423 Chr.17: 82080408-82080408 on Build GRCh38
rs767269232 Chr.17: 82080332-82080332 on Build GRCh38
rs772167116 Chr.17: 82080360-82080360 on Build GRCh38
rs543466668 Chr.17: 82080088-82080088 on Build GRCh38
rs967174544 Chr.17: 82080328-82080328 on Build GRCh38
rs561844046 Chr.17: 82080158-82080158 on Build GRCh38
rs759570023 Chr.17: 82080107-82080107 on Build GRCh38
rs773891955 Chr.17: 82080531-82080531 on Build GRCh38
rs147197890 Chr.17: 82080172-82080172 on Build GRCh38
rs559727453 Chr.17: 82080300-82080300 on Build GRCh38
rs767855765 Chr.17: 82080262-82080262 on Build GRCh38
rs138854181 Chr.17: 82080173-82080173 on Build GRCh38
rs764409477 Chr.17: 82080122-82080122 on Build GRCh38
rs149292828 Chr.17: 82080397-82080397 on Build GRCh38
rs971170761 Chr.17: 82080343-82080343 on Build GRCh38
rs981096750 Chr.17: 82080294-82080294 on Build GRCh38
rs762298431 Chr.17: 82080249-82080249 on Build GRCh38
rs773282003 Chr.17: 82080466-82080466 on Build GRCh38
rs772890309 Chr.17: 82080087-82080087 on Build GRCh38
rs769050337 Chr.17: 82080218-82080218 on Build GRCh38
rs765729117 Chr.17: 82080523-82080523 on Build GRCh38
rs541213604 Chr.17: 82080295-82080295 on Build GRCh38
rs377707508 Chr.17: 82080253-82080253 on Build GRCh38
rs371010579 Chr.17: 82080236-82080236 on Build GRCh38
rs538886615 Chr.17: 82080510-82080510 on Build GRCh38
rs766372892 Chr.17: 82080269-82080269 on Build GRCh38
rs929258693 Chr.17: 82080258-82080258 on Build GRCh38
rs368803094 Chr.17: 82080346-82080346 on Build GRCh38
rs1045825830 Chr.17: 82080204-82080204 on Build GRCh38
rs778424897 Chr.17: 82080151-82080151 on Build GRCh38
rs773689162 Chr.17: 82080362-82080362 on Build GRCh38
rs776585959 Chr.17: 82080103-82080103 on Build GRCh38
rs767929924 Chr.17: 82080130-82080130 on Build GRCh38
rs777370582 Chr.17: 82080159-82080159 on Build GRCh38
rs112359298 Chr.17: 82080317-82080317 on Build GRCh38
rs763325618 Chr.17: 82080115-82080115 on Build GRCh38
rs745764195 Chr.17: 82080176-82080176 on Build GRCh38
rs373842180 Chr.17: 82080277-82080277 on Build GRCh38
rs777086227 Chr.17: 82080388-82080388 on Build GRCh38
rs546752454 Chr.17: 82080461-82080461 on Build GRCh38
rs769606313 Chr.17: 82080191-82080191 on Build GRCh38
rs762457050 Chr.17: 82080395-82080395 on Build GRCh38
rs754450538 Chr.17: 82080135-82080135 on Build GRCh38
rs187716690 Chr.17: 82080335-82080335 on Build GRCh38
rs748322623 Chr.17: 82080354-82080354 on Build GRCh38
rs925836770 Chr.17: 82080141-82080141 on Build GRCh38
rs750906105 Chr.17: 82080266-82080266 on Build GRCh38
rs762245340 Chr.17: 82080125-82080125 on Build GRCh38
rs868753757 Chr.17: 82080522-82080522 on Build GRCh38
rs985178560 Chr.17: 82080493-82080493 on Build GRCh38
rs866527364 Chr.17: 82080526-82080526 on Build GRCh38
rs534742499 Chr.17: 82080427-82080427 on Build GRCh38
rs868511953 Chr.17: 82080534-82080534 on Build GRCh38
rs45466394 Chr.17: 82080339-82080339 on Build GRCh38
rs371919401 Chr.17: 82080251-82080251 on Build GRCh38
rs770892859 Chr.17: 82080095-82080095 on Build GRCh38
rs371343146 Chr.17: 82080403-82080403 on Build GRCh38
rs749169363 Chr.17: 82080445-82080445 on Build GRCh38
rs773826339 Chr.17: 82080121-82080121 on Build GRCh38
rs11550612 Chr.17: 82080476-82080476 on Build GRCh38
rs746831757 Chr.17: 82080161-82080161 on Build GRCh38
rs376852008 Chr.17: 82080185-82080185 on Build GRCh38
rs929436144 Chr.17: 82080481-82080481 on Build GRCh38
rs766955776 Chr.17: 82080387-82080387 on Build GRCh38
rs367649401 Chr.17: 82080321-82080321 on Build GRCh38
rs775705403 Chr.17: 82080192-82080192 on Build GRCh38
rs749400816 Chr.17: 82080340-82080340 on Build GRCh38
rs144647991 Chr.17: 82080406-82080406 on Build GRCh38
rs752241314 Chr.17: 82080278-82080278 on Build GRCh38
rs370692974 Chr.17: 82080112-82080112 on Build GRCh38
rs8078234 Chr.17: 82080313-82080313 on Build GRCh38
rs567359161 Chr.17: 82080359-82080359 on Build GRCh38
rs755818892 Chr.17: 82080426-82080426 on Build GRCh38
rs777999942 Chr.17: 82080502-82080502 on Build GRCh38
rs879049159 Chr.17: 82080281-82080281 on Build GRCh38
rs372206206 Chr.17: 82080326-82080326 on Build GRCh38
rs779191690 Chr.17: 82080458-82080458 on Build GRCh38
rs75952379 Chr.17: 82080422-82080422 on Build GRCh38
rs757878543 Chr.17: 82080280-82080280 on Build GRCh38
rs45546335 Chr.17: 82080210-82080210 on Build GRCh38
rs752013052 Chr.17: 82080123-82080123 on Build GRCh38
rs757019530 Chr.17: 82080320-82080320 on Build GRCh38
rs569590740 Chr.17: 82080333-82080333 on Build GRCh38
rs750901486 Chr.17: 82080133-82080133 on Build GRCh38
rs1049073434 Chr.17: 82080270-82080270 on Build GRCh38
rs761208637 Chr.17: 82080537-82080537 on Build GRCh38
rs774665189 Chr.17: 82080241-82080241 on Build GRCh38
rs866272811 Chr.17: 82080486-82080486 on Build GRCh38
rs771222130 Chr.17: 82080490-82080490 on Build GRCh38
rs763095373 Chr.17: 82080212-82080212 on Build GRCh38
rs772375406 Chr.17: 82080462-82080462 on Build GRCh38
rs751150468 Chr.17: 82080319-82080319 on Build GRCh38
rs752185944 Chr.17: 82080153-82080153 on Build GRCh38
rs1140619 Chr.17: 82080475-82080475 on Build GRCh38

View Full Product Details