Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82716452-82716951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN3KRP
Entrez Gene ID
79672
Gene Name
fructosamine 3 kinase related protein
Gene Aliases
FN3KL
UniGene
Hs.31431
Gene Chromosome Location
Chr.17: 82716706-82728017 on Build GRCh38
Transcripts
NM_024619.3 NR_046408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296990 c.59_60insG p.G23fs*31 Chr.17: 82716814-82716815 on Build GRCh38
6189997 c.59C>T p.S20L Chr.17: 82716814-82716814 on Build GRCh38
7189015 c.19C>T p.R7C Chr.17: 82716774-82716774 on Build GRCh38
3989452 c.86G>A p.R29Q Chr.17: 82716841-82716841 on Build GRCh38
5853847 c.107G>A p.G36E Chr.17: 82716862-82716862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922792663 Chr.17: 82716696-82716696 on Build GRCh38
rs2250000 Chr.17: 82716767-82716767 on Build GRCh38
rs904496416 Chr.17: 82716510-82716510 on Build GRCh38
rs149332044 Chr.17: 82716861-82716861 on Build GRCh38
rs754224792 Chr.17: 82716720-82716720 on Build GRCh38
rs368407230 Chr.17: 82716762-82716762 on Build GRCh38
rs202177012 Chr.17: 82716886-82716886 on Build GRCh38
rs1013637941 Chr.17: 82716684-82716684 on Build GRCh38
rs770978856 Chr.17: 82716814-82716814 on Build GRCh38
rs763449059 Chr.17: 82716865-82716865 on Build GRCh38
rs191224552 Chr.17: 82716819-82716819 on Build GRCh38
rs780370237 Chr.17: 82716917-82716917 on Build GRCh38
rs201869559 Chr.17: 82716750-82716750 on Build GRCh38
rs964523894 Chr.17: 82716651-82716651 on Build GRCh38
rs774869869 Chr.17: 82716922-82716922 on Build GRCh38
rs758945172 Chr.17: 82716772-82716772 on Build GRCh38
rs1012028427 Chr.17: 82716634-82716634 on Build GRCh38
rs952961935 Chr.17: 82716706-82716706 on Build GRCh38
rs926063859 Chr.17: 82716702-82716702 on Build GRCh38
rs752571850 Chr.17: 82716837-82716837 on Build GRCh38
rs755838050 Chr.17: 82716739-82716739 on Build GRCh38
rs755745895 Chr.17: 82716825-82716825 on Build GRCh38
rs779330365 Chr.17: 82716728-82716728 on Build GRCh38
rs777739924 Chr.17: 82716839-82716839 on Build GRCh38
rs978568866 Chr.17: 82716876-82716876 on Build GRCh38
rs946134540 Chr.17: 82716485-82716485 on Build GRCh38
rs552171189 Chr.17: 82716672-82716672 on Build GRCh38
rs112898333 Chr.17: 82716561-82716561 on Build GRCh38
rs767854325 Chr.17: 82716883-82716883 on Build GRCh38
rs183144630 Chr.17: 82716915-82716915 on Build GRCh38
rs976511691 Chr.17: 82716759-82716759 on Build GRCh38
rs965410916 Chr.17: 82716742-82716742 on Build GRCh38
rs138953335 Chr.17: 82716841-82716841 on Build GRCh38
rs978537671 Chr.17: 82716771-82716771 on Build GRCh38
rs756762062 Chr.17: 82716815-82716815 on Build GRCh38
rs936036727 Chr.17: 82716596-82716596 on Build GRCh38
rs1027237130 Chr.17: 82716472-82716472 on Build GRCh38
rs773293010 Chr.17: 82716755-82716755 on Build GRCh38
rs1024804245 Chr.17: 82716641-82716642 on Build GRCh38
rs374236465 Chr.17: 82716760-82716760 on Build GRCh38
rs994294047 Chr.17: 82716656-82716656 on Build GRCh38
rs982452981 Chr.17: 82716483-82716483 on Build GRCh38
rs1025937726 Chr.17: 82716495-82716495 on Build GRCh38
rs773096689 Chr.17: 82716804-82716804 on Build GRCh38
rs771062744 Chr.17: 82716848-82716848 on Build GRCh38
rs922678178 Chr.17: 82716482-82716482 on Build GRCh38
rs1019790574 Chr.17: 82716553-82716553 on Build GRCh38
rs751285603 Chr.17: 82716906-82716906 on Build GRCh38
rs566514730 Chr.17: 82716588-82716588 on Build GRCh38
rs758250328 Chr.17: 82716904-82716904 on Build GRCh38
rs771933725 Chr.17: 82716753-82716753 on Build GRCh38
rs953911562 Chr.17: 82716770-82716770 on Build GRCh38
rs764638305 Chr.17: 82716774-82716774 on Build GRCh38
rs775533991 Chr.17: 82716864-82716864 on Build GRCh38
rs529694099 Chr.17: 82716558-82716558 on Build GRCh38
rs376480630 Chr.17: 82716849-82716849 on Build GRCh38
rs569379364 Chr.17: 82716756-82716756 on Build GRCh38
rs765909937 Chr.17: 82716826-82716826 on Build GRCh38
rs752327088 Chr.17: 82716898-82716898 on Build GRCh38
rs1002355103 Chr.17: 82716692-82716692 on Build GRCh38
rs988782476 Chr.17: 82716902-82716902 on Build GRCh38
rs1025411139 Chr.17: 82716660-82716660 on Build GRCh38
rs768478911 Chr.17: 82716747-82716747 on Build GRCh38
rs779800505 Chr.17: 82716740-82716740 on Build GRCh38
rs936680931 Chr.17: 82716669-82716669 on Build GRCh38
rs1041703569 Chr.17: 82716490-82716490 on Build GRCh38
rs749066225 Chr.17: 82716744-82716744 on Build GRCh38
rs200977511 Chr.17: 82716815-82716815 on Build GRCh38
rs376814932 Chr.17: 82716687-82716687 on Build GRCh38
rs754672660 Chr.17: 82716833-82716833 on Build GRCh38
rs755643968 Chr.17: 82716775-82716775 on Build GRCh38
rs925861617 Chr.17: 82716661-82716661 on Build GRCh38
rs2250015 Chr.17: 82716526-82716526 on Build GRCh38
rs1018528280 Chr.17: 82716682-82716682 on Build GRCh38
rs762415299 Chr.17: 82716869-82716869 on Build GRCh38
rs371743189 Chr.17: 82716730-82716730 on Build GRCh38
rs758933598 Chr.17: 82716897-82716897 on Build GRCh38
rs776883718 Chr.17: 82716757-82716757 on Build GRCh38
rs762220708 Chr.17: 82716927-82716927 on Build GRCh38
rs144595006 Chr.17: 82716887-82716887 on Build GRCh38
rs555164597 Chr.17: 82716785-82716785 on Build GRCh38
rs946212861 Chr.17: 82716697-82716697 on Build GRCh38
rs2280252 Chr.17: 82716679-82716679 on Build GRCh38
rs1028045197 Chr.17: 82716853-82716853 on Build GRCh38
rs527607307 Chr.17: 82716633-82716633 on Build GRCh38
rs185683685 Chr.17: 82716676-82716676 on Build GRCh38
rs541534059 Chr.17: 82716920-82716920 on Build GRCh38
rs745873900 Chr.17: 82716855-82716855 on Build GRCh38
rs753495515 Chr.17: 82716814-82716815 on Build GRCh38
rs554438539 Chr.17: 82716591-82716593 on Build GRCh38
rs560324846 Chr.17: 82716928-82716928 on Build GRCh38
rs562104367 Chr.17: 82716907-82716907 on Build GRCh38
rs778504668 Chr.17: 82716903-82716904 on Build GRCh38
rs768934012 Chr.17: 82716921-82716921 on Build GRCh38
rs573333278 Chr.17: 82716788-82716788 on Build GRCh38
rs550636711 Chr.17: 82716832-82716832 on Build GRCh38
rs758160781 Chr.17: 82716838-82716838 on Build GRCh38
rs952380282 Chr.17: 82716477-82716477 on Build GRCh38
rs200062745 Chr.17: 82716834-82716834 on Build GRCh38
rs997795753 Chr.17: 82716844-82716844 on Build GRCh38
rs548375428 Chr.17: 82716573-82716573 on Build GRCh38
rs1004302201 Chr.17: 82716727-82716727 on Build GRCh38
rs1042979974 Chr.17: 82716656-82716656 on Build GRCh38
rs975525722 Chr.17: 82716666-82716666 on Build GRCh38
rs905398504 Chr.17: 82716691-82716691 on Build GRCh38
rs763360394 Chr.17: 82716817-82716817 on Build GRCh38
rs75745129 Chr.17: 82716515-82716595 on Build GRCh38
rs755331578 Chr.17: 82716723-82716723 on Build GRCh38
rs779890434 Chr.17: 82716776-82716776 on Build GRCh38
rs752080042 Chr.17: 82716823-82716823 on Build GRCh38
rs773564281 Chr.17: 82716929-82716929 on Build GRCh38
rs754797996 Chr.17: 82716783-82716783 on Build GRCh38
rs543946176 Chr.17: 82716497-82716497 on Build GRCh38
rs993162793 Chr.17: 82716489-82716489 on Build GRCh38
rs921132585 Chr.17: 82716599-82716599 on Build GRCh38
rs895076198 Chr.17: 82716606-82716606 on Build GRCh38
rs759137629 Chr.17: 82716930-82716930 on Build GRCh38
rs906274544 Chr.17: 82716653-82716653 on Build GRCh38
rs1004432960 Chr.17: 82716835-82716835 on Build GRCh38
rs964327876 Chr.17: 82716683-82716683 on Build GRCh38
rs747135475 Chr.17: 82716806-82716806 on Build GRCh38
rs748116589 Chr.17: 82716751-82716751 on Build GRCh38
rs780942002 Chr.17: 82716910-82716910 on Build GRCh38
rs766531602 Chr.17: 82716713-82716713 on Build GRCh38
rs772023434 Chr.17: 82716793-82716793 on Build GRCh38
rs765305815 Chr.17: 82716816-82716816 on Build GRCh38
rs976951562 Chr.17: 82716474-82716474 on Build GRCh38
rs748958920 Chr.17: 82716778-82716778 on Build GRCh38
rs951588962 Chr.17: 82716513-82716513 on Build GRCh38
rs952442653 Chr.17: 82716673-82716673 on Build GRCh38
rs777537652 Chr.17: 82716905-82716905 on Build GRCh38
rs1027268306 Chr.17: 82716701-82716701 on Build GRCh38
rs745306587 Chr.17: 82716738-82716738 on Build GRCh38
rs780845169 Chr.17: 82716734-82716734 on Build GRCh38

View Full Product Details