Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63954328-63954834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMSD
Entrez Gene ID
284293
Gene Name
histocompatibility minor serpin domain containing
Gene Aliases
ACC-6, ACC6, C18orf53, HSMD-v
UniGene
Hs.721604
Gene Chromosome Location
Chr.18: 63949259-63969329 on Build GRCh38
Transcripts
NM_001123366.1 XM_011525930.2 XM_017025710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265945 c.222+5G>T p.? Chr.18: 63954562-63954562 on Build GRCh38
3527155 c.100G>A p.E34K Chr.18: 63954435-63954435 on Build GRCh38
3527156 c.211G>A p.D71N Chr.18: 63954546-63954546 on Build GRCh38
4441012 c.103G>C p.D35H Chr.18: 63954438-63954438 on Build GRCh38
6476718 c.136C>G p.L46V Chr.18: 63954471-63954471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764282333 Chr.18: 63954663-63954663 on Build GRCh38
rs539179386 Chr.18: 63954683-63954683 on Build GRCh38
rs367940545 Chr.18: 63954569-63954569 on Build GRCh38
rs559021231 Chr.18: 63954438-63954453 on Build GRCh38
rs778761552 Chr.18: 63954485-63954485 on Build GRCh38
rs115236448 Chr.18: 63954781-63954781 on Build GRCh38
rs764195947 Chr.18: 63954416-63954416 on Build GRCh38
rs751558615 Chr.18: 63954671-63954671 on Build GRCh38
rs61738713 Chr.18: 63954539-63954539 on Build GRCh38
rs201288276 Chr.18: 63954454-63954454 on Build GRCh38
rs916782110 Chr.18: 63954718-63954718 on Build GRCh38
rs1048249708 Chr.18: 63954793-63954793 on Build GRCh38
rs898494346 Chr.18: 63954722-63954723 on Build GRCh38
rs760913523 Chr.18: 63954590-63954590 on Build GRCh38
rs753674390 Chr.18: 63954474-63954474 on Build GRCh38
rs538640872 Chr.18: 63954715-63954715 on Build GRCh38
rs117841082 Chr.18: 63954429-63954429 on Build GRCh38
rs978280719 Chr.18: 63954646-63954646 on Build GRCh38
rs773360701 Chr.18: 63954584-63954584 on Build GRCh38
rs746673163 Chr.18: 63954532-63954532 on Build GRCh38
rs529735281 Chr.18: 63954554-63954554 on Build GRCh38
rs1046021328 Chr.18: 63954767-63954767 on Build GRCh38
rs749665971 Chr.18: 63954564-63954564 on Build GRCh38
rs750704640 Chr.18: 63954459-63954459 on Build GRCh38
rs555748551 Chr.18: 63954792-63954792 on Build GRCh38
rs955776769 Chr.18: 63954537-63954537 on Build GRCh38
rs200412677 Chr.18: 63954559-63954559 on Build GRCh38
rs949985604 Chr.18: 63954613-63954613 on Build GRCh38
rs1047628345 Chr.18: 63954382-63954382 on Build GRCh38
rs896932101 Chr.18: 63954779-63954779 on Build GRCh38
rs370534879 Chr.18: 63954403-63954403 on Build GRCh38
rs747704202 Chr.18: 63954405-63954405 on Build GRCh38
rs761923017 Chr.18: 63954453-63954453 on Build GRCh38
rs373693173 Chr.18: 63954423-63954423 on Build GRCh38
rs1007533081 Chr.18: 63954686-63954686 on Build GRCh38
rs570491134 Chr.18: 63954682-63954682 on Build GRCh38
rs527906909 Chr.18: 63954650-63954650 on Build GRCh38
rs868864345 Chr.18: 63954522-63954522 on Build GRCh38
rs905631524 Chr.18: 63954433-63954433 on Build GRCh38
rs888283496 Chr.18: 63954808-63954808 on Build GRCh38
rs758251436 Chr.18: 63954607-63954607 on Build GRCh38
rs569562630 Chr.18: 63954763-63954763 on Build GRCh38
rs372466147 Chr.18: 63954506-63954506 on Build GRCh38
rs141974510 Chr.18: 63954719-63954720 on Build GRCh38
rs141223010 Chr.18: 63954637-63954637 on Build GRCh38
rs896879696 Chr.18: 63954544-63954544 on Build GRCh38
rs976455351 Chr.18: 63954611-63954611 on Build GRCh38
rs777553623 Chr.18: 63954523-63954523 on Build GRCh38
rs188421799 Chr.18: 63954652-63954652 on Build GRCh38
rs1018304474 Chr.18: 63954721-63954721 on Build GRCh38
rs766036843 Chr.18: 63954594-63954594 on Build GRCh38
rs891964366 Chr.18: 63954463-63954463 on Build GRCh38
rs766170009 Chr.18: 63954472-63954472 on Build GRCh38
rs925545138 Chr.18: 63954428-63954428 on Build GRCh38
rs761871788 Chr.18: 63954575-63954575 on Build GRCh38
rs758046805 Chr.18: 63954521-63954521 on Build GRCh38
rs200019033 Chr.18: 63954596-63954596 on Build GRCh38
rs8089648 Chr.18: 63954571-63954571 on Build GRCh38
rs758510351 Chr.18: 63954661-63954661 on Build GRCh38
rs72943544 Chr.18: 63954734-63954734 on Build GRCh38
rs759290376 Chr.18: 63954451-63954451 on Build GRCh38
rs945536215 Chr.18: 63954385-63954385 on Build GRCh38
rs1010415319 Chr.18: 63954666-63954666 on Build GRCh38
rs762154669 Chr.18: 63954558-63954559 on Build GRCh38
rs754375094 Chr.18: 63954378-63954378 on Build GRCh38
rs772156442 Chr.18: 63954580-63954580 on Build GRCh38
rs951439547 Chr.18: 63954735-63954735 on Build GRCh38
rs888390104 Chr.18: 63954574-63954574 on Build GRCh38
rs200798007 Chr.18: 63954477-63954477 on Build GRCh38
rs771824897 Chr.18: 63954418-63954418 on Build GRCh38
rs375545599 Chr.18: 63954466-63954466 on Build GRCh38
rs548511753 Chr.18: 63954632-63954632 on Build GRCh38
rs139861303 Chr.18: 63954598-63954598 on Build GRCh38
rs746229587 Chr.18: 63954426-63954426 on Build GRCh38
rs372213136 Chr.18: 63954389-63954389 on Build GRCh38
rs763547452 Chr.18: 63954445-63954445 on Build GRCh38
rs1044091558 Chr.18: 63954629-63954629 on Build GRCh38

View Full Product Details