Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3936497-3936987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMRK2
Entrez Gene ID
27231
Gene Name
nicotinamide riboside kinase 2
Gene Aliases
ITGB1BP3, MIBP, NRK2
UniGene
Hs.135458
Gene Chromosome Location
Chr.19: 3933103-3942416 on Build GRCh38
Transcripts
NM_001289117.1 NM_170678.2 NR_110316.1 XM_006722725.3 XM_006722726.3 XM_017026597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895051 c.34A>T p.N12Y Chr.19: 3936582-3936582 on Build GRCh38
7250874 c.117+6C>T p.? Chr.19: 3936671-3936671 on Build GRCh38
4964871 c.103G>C p.D35H Chr.19: 3936651-3936651 on Build GRCh38
4619754 c.62G>T p.S21I Chr.19: 3936610-3936610 on Build GRCh38
5532859 c.117G>A p.K39K Chr.19: 3936665-3936665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528117037 Chr.19: 3936709-3936709 on Build GRCh38
rs1054031262 Chr.19: 3936618-3936618 on Build GRCh38
rs990982007 Chr.19: 3936962-3936962 on Build GRCh38
rs753987152 Chr.19: 3936561-3936561 on Build GRCh38
rs753170349 Chr.19: 3936549-3936549 on Build GRCh38
rs112942311 Chr.19: 3936666-3936666 on Build GRCh38
rs548659945 Chr.19: 3936588-3936588 on Build GRCh38
rs866844440 Chr.19: 3936659-3936659 on Build GRCh38
rs765403691 Chr.19: 3936682-3936682 on Build GRCh38
rs952572153 Chr.19: 3936739-3936739 on Build GRCh38
rs763776885 Chr.19: 3936656-3936658 on Build GRCh38
rs891246823 Chr.19: 3936878-3936878 on Build GRCh38
rs536752021 Chr.19: 3936795-3936795 on Build GRCh38
rs372608128 Chr.19: 3936737-3936737 on Build GRCh38
rs959499542 Chr.19: 3936752-3936752 on Build GRCh38
rs550811360 Chr.19: 3936758-3936758 on Build GRCh38
rs565276519 Chr.19: 3936534-3936534 on Build GRCh38
rs966709048 Chr.19: 3936917-3936917 on Build GRCh38
rs370939791 Chr.19: 3936671-3936671 on Build GRCh38
rs750622813 Chr.19: 3936687-3936687 on Build GRCh38
rs773754847 Chr.19: 3936520-3936520 on Build GRCh38
rs372962547 Chr.19: 3936736-3936736 on Build GRCh38
rs73530370 Chr.19: 3936554-3936554 on Build GRCh38
rs376129561 Chr.19: 3936944-3936944 on Build GRCh38
rs530696398 Chr.19: 3936757-3936757 on Build GRCh38
rs759900005 Chr.19: 3936631-3936631 on Build GRCh38
rs371069572 Chr.19: 3936840-3936840 on Build GRCh38
rs75174072 Chr.19: 3936808-3936808 on Build GRCh38
rs770671972 Chr.19: 3936600-3936600 on Build GRCh38
rs777406590 Chr.19: 3936589-3936589 on Build GRCh38
rs757459048 Chr.19: 3936562-3936562 on Build GRCh38
rs992718135 Chr.19: 3936767-3936767 on Build GRCh38
rs1032517318 Chr.19: 3936931-3936931 on Build GRCh38
rs879460383 Chr.19: 3936742-3936742 on Build GRCh38
rs1001533393 Chr.19: 3936919-3936919 on Build GRCh38
rs558800789 Chr.19: 3936846-3936846 on Build GRCh38
rs776134636 Chr.19: 3936963-3936963 on Build GRCh38
rs145402201 Chr.19: 3936727-3936727 on Build GRCh38
rs925468613 Chr.19: 3936747-3936747 on Build GRCh38
rs113359113 Chr.19: 3936958-3936958 on Build GRCh38
rs917885292 Chr.19: 3936780-3936780 on Build GRCh38
rs575552063 Chr.19: 3936941-3936941 on Build GRCh38
rs754735674 Chr.19: 3936703-3936703 on Build GRCh38
rs78453343 Chr.19: 3936827-3936827 on Build GRCh38
rs200504647 Chr.19: 3936574-3936574 on Build GRCh38
rs949017166 Chr.19: 3936538-3936538 on Build GRCh38
rs773903363 Chr.19: 3936612-3936612 on Build GRCh38
rs139991507 Chr.19: 3936587-3936587 on Build GRCh38
rs779155928 Chr.19: 3936570-3936570 on Build GRCh38
rs970707194 Chr.19: 3936794-3936794 on Build GRCh38
rs955358619 Chr.19: 3936598-3936598 on Build GRCh38
rs1008296208 Chr.19: 3936879-3936879 on Build GRCh38
rs748854209 Chr.19: 3936591-3936591 on Build GRCh38
rs376394284 Chr.19: 3936639-3936639 on Build GRCh38
rs752594163 Chr.19: 3936717-3936717 on Build GRCh38
rs567722758 Chr.19: 3936769-3936769 on Build GRCh38
rs1051592368 Chr.19: 3936847-3936847 on Build GRCh38
rs959612386 Chr.19: 3936947-3936947 on Build GRCh38
rs561372448 Chr.19: 3936645-3936645 on Build GRCh38
rs914089191 Chr.19: 3936835-3936835 on Build GRCh38
rs949444581 Chr.19: 3936576-3936576 on Build GRCh38
rs768025221 Chr.19: 3936543-3936543 on Build GRCh38
rs566849030 Chr.19: 3936826-3936826 on Build GRCh38
rs1049483620 Chr.19: 3936915-3936915 on Build GRCh38
rs551640272 Chr.19: 3936710-3936710 on Build GRCh38
rs201494732 Chr.19: 3936638-3936638 on Build GRCh38
rs1021394383 Chr.19: 3936899-3936899 on Build GRCh38
rs949155753 Chr.19: 3936764-3936764 on Build GRCh38
rs907515901 Chr.19: 3936796-3936796 on Build GRCh38
rs996945584 Chr.19: 3936584-3936584 on Build GRCh38
rs563014904 Chr.19: 3936577-3936577 on Build GRCh38
rs753894665 Chr.19: 3936672-3936672 on Build GRCh38
rs544428504 Chr.19: 3936556-3936556 on Build GRCh38
rs34364924 Chr.19: 3936821-3936822 on Build GRCh38
rs761921104 Chr.19: 3936681-3936681 on Build GRCh38
rs1019264846 Chr.19: 3936702-3936702 on Build GRCh38
rs375750253 Chr.19: 3936540-3936540 on Build GRCh38

View Full Product Details