Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6681850-6682350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
UniGene
Hs.529053
Gene Chromosome Location
Chr.19: 6677835-6720682 on Build GRCh38
Transcripts
NM_000064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2727838 c.4228G>A p.G1410S Chr.19: 6682174-6682174 on Build GRCh38
4547618 c.4271G>A p.G1424D Chr.19: 6682020-6682020 on Build GRCh38
2727839 c.4218C>T p.S1406S Chr.19: 6682184-6682184 on Build GRCh38
4288604 c.4322G>A p.R1441K Chr.19: 6681969-6681969 on Build GRCh38
6615968 c.4287C>A p.I1429I Chr.19: 6682004-6682004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775988121 Chr.19: 6682150-6682150 on Build GRCh38
rs111228502 Chr.19: 6682227-6682227 on Build GRCh38
rs200597513 Chr.19: 6682222-6682222 on Build GRCh38
rs1043541501 Chr.19: 6682291-6682291 on Build GRCh38
rs374704078 Chr.19: 6682102-6682102 on Build GRCh38
rs771359858 Chr.19: 6682138-6682138 on Build GRCh38
rs772599294 Chr.19: 6682124-6682124 on Build GRCh38
rs756586729 Chr.19: 6682276-6682276 on Build GRCh38
rs201597793 Chr.19: 6681914-6681914 on Build GRCh38
rs753897719 Chr.19: 6682166-6682166 on Build GRCh38
rs946583070 Chr.19: 6682286-6682286 on Build GRCh38
rs575119407 Chr.19: 6682224-6682224 on Build GRCh38
rs763380367 Chr.19: 6682025-6682025 on Build GRCh38
rs984274936 Chr.19: 6682023-6682023 on Build GRCh38
rs759927155 Chr.19: 6682036-6682036 on Build GRCh38
rs148227405 Chr.19: 6682225-6682225 on Build GRCh38
rs777880161 Chr.19: 6681959-6681959 on Build GRCh38
rs777432001 Chr.19: 6682245-6682245 on Build GRCh38
rs757313067 Chr.19: 6682180-6682180 on Build GRCh38
rs764140957 Chr.19: 6682059-6682059 on Build GRCh38
rs764310139 Chr.19: 6682157-6682157 on Build GRCh38
rs746439993 Chr.19: 6682281-6682281 on Build GRCh38
rs780959208 Chr.19: 6682107-6682107 on Build GRCh38
rs148695501 Chr.19: 6681934-6681934 on Build GRCh38
rs772763964 Chr.19: 6682024-6682024 on Build GRCh38
rs113281226 Chr.19: 6682139-6682139 on Build GRCh38
rs752000215 Chr.19: 6682028-6682028 on Build GRCh38
rs769427410 Chr.19: 6682119-6682119 on Build GRCh38
rs1055468845 Chr.19: 6681915-6681915 on Build GRCh38
rs751920993 Chr.19: 6682133-6682133 on Build GRCh38
rs764831445 Chr.19: 6681922-6681922 on Build GRCh38
rs529660041 Chr.19: 6682069-6682069 on Build GRCh38
rs147116781 Chr.19: 6681972-6681972 on Build GRCh38
rs370410378 Chr.19: 6682050-6682050 on Build GRCh38
rs775353799 Chr.19: 6681903-6681903 on Build GRCh38
rs753899226 Chr.19: 6682076-6682076 on Build GRCh38
rs773996375 Chr.19: 6682277-6682277 on Build GRCh38
rs990661864 Chr.19: 6682198-6682198 on Build GRCh38
rs757531444 Chr.19: 6682110-6682117 on Build GRCh38
rs760821811 Chr.19: 6682156-6682156 on Build GRCh38
rs768065757 Chr.19: 6682145-6682145 on Build GRCh38
rs756200539 Chr.19: 6681958-6681958 on Build GRCh38
rs143680252 Chr.19: 6681919-6681919 on Build GRCh38
rs746207752 Chr.19: 6682113-6682114 on Build GRCh38
rs776123692 Chr.19: 6681927-6681927 on Build GRCh38
rs1014475053 Chr.19: 6681971-6681971 on Build GRCh38
rs368531504 Chr.19: 6682113-6682113 on Build GRCh38
rs750932330 Chr.19: 6681940-6681940 on Build GRCh38
rs904617601 Chr.19: 6682299-6682299 on Build GRCh38
rs948764879 Chr.19: 6681922-6681922 on Build GRCh38
rs374607713 Chr.19: 6682152-6682152 on Build GRCh38
rs143070159 Chr.19: 6682144-6682144 on Build GRCh38
rs182162639 Chr.19: 6682064-6682064 on Build GRCh38
rs755251519 Chr.19: 6681950-6681950 on Build GRCh38
rs756357300 Chr.19: 6682058-6682058 on Build GRCh38
rs771805341 Chr.19: 6681899-6681899 on Build GRCh38
rs748867754 Chr.19: 6682248-6682248 on Build GRCh38
rs200878391 Chr.19: 6681977-6681977 on Build GRCh38
rs540818117 Chr.19: 6682298-6682298 on Build GRCh38
rs569469344 Chr.19: 6682292-6682292 on Build GRCh38
rs7951 Chr.19: 6681980-6681980 on Build GRCh38
rs937404920 Chr.19: 6682305-6682305 on Build GRCh38
rs191489530 Chr.19: 6681943-6681943 on Build GRCh38
rs781520118 Chr.19: 6681953-6681953 on Build GRCh38
rs45532534 Chr.19: 6682103-6682103 on Build GRCh38
rs369993135 Chr.19: 6681935-6681935 on Build GRCh38
rs112769578 Chr.19: 6682141-6682141 on Build GRCh38
rs753012307 Chr.19: 6682047-6682047 on Build GRCh38
rs758275799 Chr.19: 6682093-6682093 on Build GRCh38
rs781706996 Chr.19: 6682111-6682112 on Build GRCh38
rs752850583 Chr.19: 6681956-6681956 on Build GRCh38
rs149627133 Chr.19: 6682004-6682004 on Build GRCh38
rs751912518 Chr.19: 6681947-6681947 on Build GRCh38
rs754895856 Chr.19: 6682223-6682223 on Build GRCh38
rs960824132 Chr.19: 6682151-6682151 on Build GRCh38
rs745800783 Chr.19: 6681895-6681895 on Build GRCh38
rs763510721 Chr.19: 6681941-6681941 on Build GRCh38
rs751871723 Chr.19: 6682047-6682047 on Build GRCh38
rs372163622 Chr.19: 6682122-6682122 on Build GRCh38
rs747812523 Chr.19: 6682114-6682114 on Build GRCh38
rs867087895 Chr.19: 6682118-6682118 on Build GRCh38
rs768241258 Chr.19: 6681919-6681919 on Build GRCh38
rs761423793 Chr.19: 6681988-6681988 on Build GRCh38
rs113342628 Chr.19: 6681938-6681938 on Build GRCh38
rs770608489 Chr.19: 6682259-6682259 on Build GRCh38
rs193293763 Chr.19: 6682104-6682104 on Build GRCh38
rs377358917 Chr.19: 6682026-6682026 on Build GRCh38
rs772034590 Chr.19: 6681973-6681973 on Build GRCh38
rs779838912 Chr.19: 6681974-6681974 on Build GRCh38
rs933914091 Chr.19: 6682214-6682214 on Build GRCh38
rs767975717 Chr.19: 6682041-6682041 on Build GRCh38
rs867572584 Chr.19: 6682127-6682127 on Build GRCh38

View Full Product Details