Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44219715-44220208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMAP1
Entrez Gene ID
55929
Gene Name
DNA methyltransferase 1 associated protein 1
Gene Aliases
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4
UniGene
Hs.8008
Gene Chromosome Location
Chr.1: 44213428-44220679 on Build GRCh38
Transcripts
NM_001034023.1 NM_001034024.1 NM_019100.4 XM_005271037.4 XM_005271039.1 XM_005271040.1 XM_006710771.3 XM_011541784.2 XM_011541785.2 XM_017001807.1 XM_017001808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910000 c.1036C>G p.L346V Chr.1: 44219863-44219863 on Build GRCh38
910001 c.1175G>A p.R392Q Chr.1: 44220140-44220140 on Build GRCh38
6645146 c.1197C>A p.A399A Chr.1: 44220162-44220162 on Build GRCh38
4008367 c.1070C>T p.T357M Chr.1: 44220035-44220035 on Build GRCh38
2171605 c.1004_1006delAGA p.K337delK Chr.1: 44219831-44219833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766235875 Chr.1: 44219967-44219967 on Build GRCh38
rs750381568 Chr.1: 44219850-44219850 on Build GRCh38
rs146514924 Chr.1: 44220138-44220138 on Build GRCh38
rs763717827 Chr.1: 44220118-44220118 on Build GRCh38
rs771433988 Chr.1: 44219838-44219838 on Build GRCh38
rs753318592 Chr.1: 44220171-44220171 on Build GRCh38
rs746593320 Chr.1: 44219823-44219823 on Build GRCh38
rs906155455 Chr.1: 44219753-44219753 on Build GRCh38
rs746310575 Chr.1: 44219995-44219995 on Build GRCh38
rs748252842 Chr.1: 44220123-44220123 on Build GRCh38
rs752229758 Chr.1: 44220072-44220072 on Build GRCh38
rs767182825 Chr.1: 44220068-44220068 on Build GRCh38
rs12755972 Chr.1: 44219806-44219806 on Build GRCh38
rs755514530 Chr.1: 44220185-44220185 on Build GRCh38
rs368180564 Chr.1: 44219991-44219991 on Build GRCh38
rs1000046346 Chr.1: 44220156-44220156 on Build GRCh38
rs764172795 Chr.1: 44219755-44219755 on Build GRCh38
rs747659793 Chr.1: 44219837-44219837 on Build GRCh38
rs1032018963 Chr.1: 44219739-44219739 on Build GRCh38
rs569718563 Chr.1: 44220048-44220048 on Build GRCh38
rs756828038 Chr.1: 44220180-44220180 on Build GRCh38
rs189650822 Chr.1: 44220075-44220075 on Build GRCh38
rs750444247 Chr.1: 44219927-44219927 on Build GRCh38
rs759314123 Chr.1: 44220054-44220054 on Build GRCh38
rs768786023 Chr.1: 44219907-44219907 on Build GRCh38
rs767146610 Chr.1: 44220165-44220165 on Build GRCh38
rs760275631 Chr.1: 44220170-44220170 on Build GRCh38
rs770986672 Chr.1: 44220150-44220150 on Build GRCh38
rs377753318 Chr.1: 44219779-44219779 on Build GRCh38
rs969870238 Chr.1: 44219841-44219841 on Build GRCh38
rs779181470 Chr.1: 44219908-44219908 on Build GRCh38
rs74983620 Chr.1: 44219989-44219989 on Build GRCh38
rs762999018 Chr.1: 44219928-44219928 on Build GRCh38
rs116279448 Chr.1: 44220090-44220090 on Build GRCh38
rs576488630 Chr.1: 44219917-44219917 on Build GRCh38
rs746214283 Chr.1: 44219888-44219888 on Build GRCh38
rs775390346 Chr.1: 44220167-44220167 on Build GRCh38
rs930319726 Chr.1: 44219915-44219915 on Build GRCh38
rs917343309 Chr.1: 44220009-44220009 on Build GRCh38
rs149066039 Chr.1: 44220024-44220024 on Build GRCh38
rs772360829 Chr.1: 44219896-44219896 on Build GRCh38
rs368073383 Chr.1: 44220001-44220001 on Build GRCh38
rs762934376 Chr.1: 44220043-44220043 on Build GRCh38
rs753373766 Chr.1: 44220087-44220087 on Build GRCh38
rs761939880 Chr.1: 44220027-44220027 on Build GRCh38
rs1015359608 Chr.1: 44219938-44219938 on Build GRCh38
rs780703083 Chr.1: 44219835-44219835 on Build GRCh38
rs757172439 Chr.1: 44219769-44219769 on Build GRCh38
rs12760076 Chr.1: 44219807-44219807 on Build GRCh38
rs754618278 Chr.1: 44219834-44219834 on Build GRCh38
rs41270415 Chr.1: 44219992-44219992 on Build GRCh38
rs759526565 Chr.1: 44220163-44220163 on Build GRCh38
rs773525336 Chr.1: 44220002-44220002 on Build GRCh38
rs751745689 Chr.1: 44220157-44220157 on Build GRCh38
rs1049289052 Chr.1: 44219850-44219850 on Build GRCh38
rs979885958 Chr.1: 44219906-44219906 on Build GRCh38
rs777229352 Chr.1: 44220079-44220079 on Build GRCh38
rs1045004334 Chr.1: 44219751-44219751 on Build GRCh38
rs753897927 Chr.1: 44219765-44219765 on Build GRCh38
rs143355210 Chr.1: 44220182-44220182 on Build GRCh38
rs758418548 Chr.1: 44220010-44220010 on Build GRCh38
rs117340264 Chr.1: 44220096-44220096 on Build GRCh38
rs774902875 Chr.1: 44219863-44219863 on Build GRCh38
rs564972388 Chr.1: 44219979-44219979 on Build GRCh38
rs116432610 Chr.1: 44219971-44219971 on Build GRCh38
rs116305257 Chr.1: 44220053-44220053 on Build GRCh38
rs878922277 Chr.1: 44219941-44219941 on Build GRCh38
rs768962026 Chr.1: 44220098-44220098 on Build GRCh38
rs766472780 Chr.1: 44220153-44220153 on Build GRCh38
rs951792544 Chr.1: 44220120-44220120 on Build GRCh38
rs879241457 Chr.1: 44219951-44219951 on Build GRCh38
rs376830920 Chr.1: 44220152-44220152 on Build GRCh38
rs999952777 Chr.1: 44220037-44220037 on Build GRCh38
rs777274603 Chr.1: 44219994-44219994 on Build GRCh38
rs769937212 Chr.1: 44220029-44220029 on Build GRCh38
rs765263768 Chr.1: 44219918-44219918 on Build GRCh38
rs755218732 Chr.1: 44219788-44219788 on Build GRCh38
rs765138145 Chr.1: 44219774-44219774 on Build GRCh38
rs774210173 Chr.1: 44220160-44220160 on Build GRCh38
rs749417435 Chr.1: 44220149-44220149 on Build GRCh38
rs775839842 Chr.1: 44219902-44219902 on Build GRCh38
rs773455439 Chr.1: 44220036-44220036 on Build GRCh38
rs147610607 Chr.1: 44220005-44220005 on Build GRCh38
rs545072434 Chr.1: 44219740-44219740 on Build GRCh38
rs965373926 Chr.1: 44219953-44219953 on Build GRCh38
rs752244753 Chr.1: 44219993-44219993 on Build GRCh38
rs773404079 Chr.1: 44220146-44220146 on Build GRCh38
rs202183088 Chr.1: 44219988-44219988 on Build GRCh38
rs997898480 Chr.1: 44219959-44219959 on Build GRCh38
rs781630711 Chr.1: 44220102-44220102 on Build GRCh38
rs138593911 Chr.1: 44220023-44220023 on Build GRCh38
rs750213674 Chr.1: 44219776-44219776 on Build GRCh38
rs779841090 Chr.1: 44219802-44219802 on Build GRCh38

View Full Product Details