Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18931409-18931902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOMER3
Entrez Gene ID
9454
Gene Name
homer scaffolding protein 3
Gene Aliases
HOMER-3, VESL3
UniGene
Hs.720208
Gene Chromosome Location
Chr.19: 18929201-18941258 on Build GRCh38
Transcripts
NM_001145721.1 NM_001145722.1 NM_001145724.1 NM_004838.3 NR_027297.1 XM_006722943.1 XM_006722944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681516 c.748G>T p.G250W Chr.19: 18931568-18931568 on Build GRCh38
3822381 c.761G>C p.G254A Chr.19: 18931555-18931555 on Build GRCh38
6084163 c.768G>A p.S256S Chr.19: 18931548-18931548 on Build GRCh38
5833678 c.761G>C p.G254A Chr.19: 18931555-18931555 on Build GRCh38
6084164 c.768G>A p.S256S Chr.19: 18931548-18931548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139883016 Chr.19: 18931539-18931539 on Build GRCh38
rs772090218 Chr.19: 18931497-18931497 on Build GRCh38
rs764770947 Chr.19: 18931562-18931562 on Build GRCh38
rs764544065 Chr.19: 18931571-18931571 on Build GRCh38
rs759179202 Chr.19: 18931549-18931549 on Build GRCh38
rs763603379 Chr.19: 18931660-18931660 on Build GRCh38
rs112086166 Chr.19: 18931821-18931821 on Build GRCh38
rs201661602 Chr.19: 18931436-18931436 on Build GRCh38
rs770847447 Chr.19: 18931636-18931636 on Build GRCh38
rs557719727 Chr.19: 18931801-18931801 on Build GRCh38
rs953212838 Chr.19: 18931757-18931757 on Build GRCh38
rs757555627 Chr.19: 18931580-18931580 on Build GRCh38
rs908179658 Chr.19: 18931795-18931795 on Build GRCh38
rs975913594 Chr.19: 18931443-18931443 on Build GRCh38
rs1030981734 Chr.19: 18931706-18931706 on Build GRCh38
rs370632348 Chr.19: 18931498-18931498 on Build GRCh38
rs1046702723 Chr.19: 18931531-18931531 on Build GRCh38
rs746088116 Chr.19: 18931480-18931480 on Build GRCh38
rs948587392 Chr.19: 18931808-18931808 on Build GRCh38
rs915810098 Chr.19: 18931805-18931805 on Build GRCh38
rs368009860 Chr.19: 18931653-18931653 on Build GRCh38
rs745510533 Chr.19: 18931651-18931651 on Build GRCh38
rs78343319 Chr.19: 18931444-18931444 on Build GRCh38
rs756202089 Chr.19: 18931489-18931489 on Build GRCh38
rs748698153 Chr.19: 18931660-18931660 on Build GRCh38
rs145484146 Chr.19: 18931569-18931569 on Build GRCh38
rs1022535412 Chr.19: 18931719-18931719 on Build GRCh38
rs34229999 Chr.19: 18931866-18931867 on Build GRCh38
rs375317299 Chr.19: 18931454-18931454 on Build GRCh38
rs1045867230 Chr.19: 18931576-18931576 on Build GRCh38
rs75657844 Chr.19: 18931523-18931523 on Build GRCh38
rs774823124 Chr.19: 18931664-18931664 on Build GRCh38
rs928781117 Chr.19: 18931862-18931862 on Build GRCh38
rs73923187 Chr.19: 18931442-18931442 on Build GRCh38
rs375438043 Chr.19: 18931548-18931548 on Build GRCh38
rs758866803 Chr.19: 18931445-18931445 on Build GRCh38
rs1003558394 Chr.19: 18931672-18931672 on Build GRCh38
rs575503711 Chr.19: 18931716-18931716 on Build GRCh38
rs937521876 Chr.19: 18931872-18931872 on Build GRCh38
rs142700713 Chr.19: 18931588-18931588 on Build GRCh38
rs770309252 Chr.19: 18931443-18931443 on Build GRCh38
rs749291184 Chr.19: 18931608-18931608 on Build GRCh38
rs975591041 Chr.19: 18931750-18931750 on Build GRCh38
rs1006639848 Chr.19: 18931633-18931633 on Build GRCh38
rs762383290 Chr.19: 18931656-18931656 on Build GRCh38
rs776333499 Chr.19: 18931646-18931646 on Build GRCh38
rs925032650 Chr.19: 18931769-18931769 on Build GRCh38
rs757649518 Chr.19: 18931463-18931463 on Build GRCh38
rs778086699 Chr.19: 18931447-18931447 on Build GRCh38
rs377478783 Chr.19: 18931479-18931479 on Build GRCh38
rs746734578 Chr.19: 18931534-18931534 on Build GRCh38
rs1011127005 Chr.19: 18931717-18931717 on Build GRCh38
rs200520741 Chr.19: 18931625-18931625 on Build GRCh38
rs982431935 Chr.19: 18931782-18931782 on Build GRCh38
rs755118382 Chr.19: 18931620-18931620 on Build GRCh38
rs10410004 Chr.19: 18931799-18931799 on Build GRCh38
rs769134625 Chr.19: 18931613-18931613 on Build GRCh38
rs190406448 Chr.19: 18931659-18931659 on Build GRCh38
rs780062792 Chr.19: 18931492-18931492 on Build GRCh38
rs73529155 Chr.19: 18931879-18931879 on Build GRCh38
rs1012749120 Chr.19: 18931595-18931595 on Build GRCh38
rs964601159 Chr.19: 18931720-18931720 on Build GRCh38
rs375130225 Chr.19: 18931629-18931629 on Build GRCh38
rs752033379 Chr.19: 18931488-18931488 on Build GRCh38
rs775238819 Chr.19: 18931654-18931654 on Build GRCh38
rs780349520 Chr.19: 18931606-18931606 on Build GRCh38
rs955099613 Chr.19: 18931761-18931761 on Build GRCh38
rs779898277 Chr.19: 18931655-18931655 on Build GRCh38
rs922376650 Chr.19: 18931759-18931759 on Build GRCh38
rs755793159 Chr.19: 18931646-18931646 on Build GRCh38
rs762371193 Chr.19: 18931676-18931676 on Build GRCh38
rs767924157 Chr.19: 18931581-18931581 on Build GRCh38
rs57792670 Chr.19: 18931578-18931578 on Build GRCh38
rs946287539 Chr.19: 18931809-18931809 on Build GRCh38
rs149686899 Chr.19: 18931544-18931544 on Build GRCh38
rs985849926 Chr.19: 18931766-18931766 on Build GRCh38
rs748098701 Chr.19: 18931493-18931493 on Build GRCh38
rs933683258 Chr.19: 18931496-18931496 on Build GRCh38
rs931707929 Chr.19: 18931466-18931466 on Build GRCh38
rs976028243 Chr.19: 18931673-18931673 on Build GRCh38
rs766093745 Chr.19: 18931433-18931433 on Build GRCh38
rs750651658 Chr.19: 18931587-18931587 on Build GRCh38
rs201906107 Chr.19: 18931526-18931526 on Build GRCh38

View Full Product Details