Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47264540-47265032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
UniGene
Hs.227782
Gene Chromosome Location
Chr.19: 47256474-47271958 on Build GRCh38
Transcripts
NM_015603.2 XM_017026578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764755 c.575G>A p.R192Q Chr.19: 47264801-47264801 on Build GRCh38
5364726 c.675delC p.D227fs*26 Chr.19: 47264901-47264901 on Build GRCh38
1612472 c.499A>T p.M167L Chr.19: 47264639-47264639 on Build GRCh38
5031584 c.712G>A p.E238K Chr.19: 47264938-47264938 on Build GRCh38
3749524 c.645G>C p.E215D Chr.19: 47264871-47264871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766811639 Chr.19: 47264921-47264921 on Build GRCh38
rs923498933 Chr.19: 47265003-47265003 on Build GRCh38
rs761864246 Chr.19: 47264701-47264702 on Build GRCh38
rs897671203 Chr.19: 47264895-47264895 on Build GRCh38
rs774015477 Chr.19: 47264658-47264658 on Build GRCh38
rs201452989 Chr.19: 47264633-47264633 on Build GRCh38
rs113198762 Chr.19: 47264746-47264746 on Build GRCh38
rs567723167 Chr.19: 47264882-47264882 on Build GRCh38
rs200234430 Chr.19: 47264904-47264904 on Build GRCh38
rs755816844 Chr.19: 47264883-47264883 on Build GRCh38
rs202074409 Chr.19: 47264857-47264857 on Build GRCh38
rs529448151 Chr.19: 47264827-47264827 on Build GRCh38
rs201513324 Chr.19: 47264940-47264940 on Build GRCh38
rs761629950 Chr.19: 47264738-47264738 on Build GRCh38
rs377003541 Chr.19: 47264568-47264568 on Build GRCh38
rs760584373 Chr.19: 47264565-47264565 on Build GRCh38
rs988738067 Chr.19: 47264964-47264964 on Build GRCh38
rs1034832430 Chr.19: 47264594-47264595 on Build GRCh38
rs773212656 Chr.19: 47264824-47264824 on Build GRCh38
rs764635988 Chr.19: 47264737-47264737 on Build GRCh38
rs765668773 Chr.19: 47264875-47264875 on Build GRCh38
rs558779868 Chr.19: 47264979-47264979 on Build GRCh38
rs779619274 Chr.19: 47264800-47264800 on Build GRCh38
rs780268098 Chr.19: 47264591-47264591 on Build GRCh38
rs569098125 Chr.19: 47264858-47264858 on Build GRCh38
rs773558758 Chr.19: 47264917-47264917 on Build GRCh38
rs781671505 Chr.19: 47264605-47264605 on Build GRCh38
rs202156396 Chr.19: 47264947-47264947 on Build GRCh38
rs765257262 Chr.19: 47264743-47264743 on Build GRCh38
rs770988713 Chr.19: 47264892-47264892 on Build GRCh38
rs187437597 Chr.19: 47264708-47264708 on Build GRCh38
rs757919555 Chr.19: 47264799-47264799 on Build GRCh38
rs965800630 Chr.19: 47264838-47264838 on Build GRCh38
rs768449354 Chr.19: 47264735-47264735 on Build GRCh38
rs763027219 Chr.19: 47264680-47264680 on Build GRCh38
rs753096066 Chr.19: 47264702-47264702 on Build GRCh38
rs757120629 Chr.19: 47264958-47264958 on Build GRCh38
rs372655114 Chr.19: 47264734-47264734 on Build GRCh38
rs201629819 Chr.19: 47264707-47264707 on Build GRCh38
rs200098648 Chr.19: 47264836-47264836 on Build GRCh38
rs527261409 Chr.19: 47264758-47264758 on Build GRCh38
rs1001227480 Chr.19: 47264747-47264747 on Build GRCh38
rs901829129 Chr.19: 47264712-47264712 on Build GRCh38
rs749135400 Chr.19: 47264887-47264887 on Build GRCh38
rs768749441 Chr.19: 47264867-47264867 on Build GRCh38
rs777635498 Chr.19: 47264948-47264948 on Build GRCh38
rs778480319 Chr.19: 47264619-47264619 on Build GRCh38
rs756559161 Chr.19: 47264704-47264704 on Build GRCh38
rs750948280 Chr.19: 47264840-47264840 on Build GRCh38
rs184137525 Chr.19: 47264767-47264767 on Build GRCh38
rs968323400 Chr.19: 47264999-47264999 on Build GRCh38
rs375699108 Chr.19: 47264744-47264744 on Build GRCh38
rs988423771 Chr.19: 47264870-47264870 on Build GRCh38
rs769602417 Chr.19: 47264822-47264822 on Build GRCh38
rs759621777 Chr.19: 47264825-47264825 on Build GRCh38
rs199988747 Chr.19: 47264698-47264698 on Build GRCh38
rs993820245 Chr.19: 47264599-47264599 on Build GRCh38
rs746557968 Chr.19: 47264850-47264850 on Build GRCh38
rs202227718 Chr.19: 47264905-47264905 on Build GRCh38
rs369117321 Chr.19: 47264718-47264718 on Build GRCh38
rs754150222 Chr.19: 47264832-47264832 on Build GRCh38
rs539946123 Chr.19: 47264662-47264662 on Build GRCh38
rs766609342 Chr.19: 47264901-47264901 on Build GRCh38
rs74541218 Chr.19: 47264600-47264600 on Build GRCh38
rs752197270 Chr.19: 47264941-47264941 on Build GRCh38
rs926608422 Chr.19: 47264879-47264879 on Build GRCh38
rs1003353563 Chr.19: 47264567-47264567 on Build GRCh38
rs751757229 Chr.19: 47264595-47264595 on Build GRCh38
rs747837480 Chr.19: 47264856-47264856 on Build GRCh38
rs969773116 Chr.19: 47264571-47264571 on Build GRCh38
rs760906023 Chr.19: 47264864-47264864 on Build GRCh38
rs773990557 Chr.19: 47264633-47264635 on Build GRCh38
rs1006172532 Chr.19: 47264736-47264736 on Build GRCh38
rs779052907 Chr.19: 47264894-47264894 on Build GRCh38
rs377289724 Chr.19: 47264815-47264815 on Build GRCh38
rs775184887 Chr.19: 47264733-47264733 on Build GRCh38
rs780952824 Chr.19: 47264855-47264855 on Build GRCh38
rs191815512 Chr.19: 47264745-47264745 on Build GRCh38
rs762121355 Chr.19: 47264910-47264910 on Build GRCh38
rs757675273 Chr.19: 47264781-47264781 on Build GRCh38
rs760533096 Chr.19: 47264666-47264666 on Build GRCh38
rs377072160 Chr.19: 47264863-47264863 on Build GRCh38
rs200888797 Chr.19: 47264641-47264641 on Build GRCh38
rs200381759 Chr.19: 47264973-47264973 on Build GRCh38
rs964976063 Chr.19: 47264972-47264972 on Build GRCh38
rs758468860 Chr.19: 47264589-47264589 on Build GRCh38
rs151252612 Chr.19: 47264801-47264801 on Build GRCh38
rs767299559 Chr.19: 47264704-47264704 on Build GRCh38
rs763363009 Chr.19: 47264920-47264920 on Build GRCh38
rs1010021375 Chr.19: 47264915-47264915 on Build GRCh38
rs201111116 Chr.19: 47264833-47264833 on Build GRCh38
rs770351779 Chr.19: 47264654-47264654 on Build GRCh38
rs946670676 Chr.19: 47264755-47264755 on Build GRCh38
rs754617753 Chr.19: 47264853-47264853 on Build GRCh38
rs534168821 Chr.19: 47264877-47264877 on Build GRCh38
rs766388399 Chr.19: 47264684-47264684 on Build GRCh38
rs745425218 Chr.19: 47264719-47264719 on Build GRCh38
rs372884937 Chr.19: 47264793-47264793 on Build GRCh38
rs542800293 Chr.19: 47264663-47264663 on Build GRCh38
rs993436511 Chr.19: 47264823-47264823 on Build GRCh38
rs751705680 Chr.19: 47264687-47264687 on Build GRCh38
rs753680941 Chr.19: 47264952-47264952 on Build GRCh38
rs748559994 Chr.19: 47264616-47264616 on Build GRCh38
rs752063406 Chr.19: 47264929-47264929 on Build GRCh38
rs946933052 Chr.19: 47264872-47264872 on Build GRCh38
rs776255827 Chr.19: 47264820-47264820 on Build GRCh38
rs778934146 Chr.19: 47264962-47264962 on Build GRCh38
rs770418476 Chr.19: 47264618-47264618 on Build GRCh38
rs777602055 Chr.19: 47264884-47264884 on Build GRCh38
rs150553117 Chr.19: 47264814-47264814 on Build GRCh38
rs771763612 Chr.19: 47264665-47264665 on Build GRCh38
rs780641278 Chr.19: 47264881-47264881 on Build GRCh38
rs747030757 Chr.19: 47264903-47264903 on Build GRCh38
rs755121878 Chr.19: 47264697-47264697 on Build GRCh38
rs758928050 Chr.19: 47264937-47264937 on Build GRCh38
rs200857202 Chr.19: 47264849-47264849 on Build GRCh38
rs2032811 Chr.19: 47264871-47264871 on Build GRCh38
rs975952482 Chr.19: 47264785-47264785 on Build GRCh38
rs764418073 Chr.19: 47264831-47264831 on Build GRCh38
rs374702564 Chr.19: 47264880-47264880 on Build GRCh38
rs773792508 Chr.19: 47264604-47264604 on Build GRCh38
rs750236459 Chr.19: 47264741-47264741 on Build GRCh38
rs753790394 Chr.19: 47264676-47264676 on Build GRCh38
rs968436564 Chr.19: 47264942-47264942 on Build GRCh38
rs754278679 Chr.19: 47264779-47264779 on Build GRCh38
rs762020143 Chr.19: 47264874-47264874 on Build GRCh38
rs142258592 Chr.19: 47264615-47264615 on Build GRCh38
rs538184871 Chr.19: 47264861-47264861 on Build GRCh38
rs757868241 Chr.19: 47264848-47264848 on Build GRCh38
rs760957343 Chr.19: 47264828-47264828 on Build GRCh38
rs760344736 Chr.19: 47264893-47264894 on Build GRCh38
rs771710800 Chr.19: 47264722-47264722 on Build GRCh38
rs756507747 Chr.19: 47264761-47264761 on Build GRCh38
rs554313823 Chr.19: 47264878-47264878 on Build GRCh38
rs745359387 Chr.19: 47264798-47264798 on Build GRCh38
rs772258987 Chr.19: 47264675-47264675 on Build GRCh38
rs201241449 Chr.19: 47264914-47264914 on Build GRCh38
rs748039161 Chr.19: 47264821-47264821 on Build GRCh38

View Full Product Details