Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47751288-47751687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR2
Entrez Gene ID
29997
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
PICT-1, PICT1
UniGene
Hs.421907
Gene Chromosome Location
Chr.19: 47745536-47757066 on Build GRCh38
Transcripts
NM_015710.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3106730 c.639C>T p.F213F Chr.19: 47751560-47751560 on Build GRCh38
3106731 c.656A>G p.K219R Chr.19: 47751577-47751577 on Build GRCh38
712141 c.629A>C p.D210A Chr.19: 47751550-47751550 on Build GRCh38
6239538 c.652A>T p.K218* Chr.19: 47751573-47751573 on Build GRCh38
7318312 c.654G>C p.K218N Chr.19: 47751575-47751575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11538669 Chr.19: 47751542-47751542 on Build GRCh38
rs747609478 Chr.19: 47751551-47751551 on Build GRCh38
rs200973335 Chr.19: 47751589-47751589 on Build GRCh38
rs753859740 Chr.19: 47751524-47751524 on Build GRCh38
rs1015456708 Chr.19: 47751415-47751415 on Build GRCh38
rs779067725 Chr.19: 47751533-47751533 on Build GRCh38
rs142374779 Chr.19: 47751590-47751590 on Build GRCh38
rs1027837855 Chr.19: 47751451-47751452 on Build GRCh38
rs754966051 Chr.19: 47751512-47751512 on Build GRCh38
rs757351238 Chr.19: 47751530-47751530 on Build GRCh38
rs767396110 Chr.19: 47751582-47751582 on Build GRCh38
rs897962683 Chr.19: 47751445-47751445 on Build GRCh38
rs750600449 Chr.19: 47751603-47751603 on Build GRCh38
rs923688973 Chr.19: 47751473-47751473 on Build GRCh38
rs138696857 Chr.19: 47751449-47751450 on Build GRCh38
rs951063448 Chr.19: 47751462-47751462 on Build GRCh38
rs570192319 Chr.19: 47751588-47751588 on Build GRCh38
rs946727298 Chr.19: 47751651-47751651 on Build GRCh38
rs761913677 Chr.19: 47751613-47751613 on Build GRCh38
rs756139350 Chr.19: 47751417-47751417 on Build GRCh38
rs946701334 Chr.19: 47751343-47751343 on Build GRCh38
rs1026547915 Chr.19: 47751454-47751454 on Build GRCh38
rs552271624 Chr.19: 47751436-47751436 on Build GRCh38
rs750109611 Chr.19: 47751488-47751488 on Build GRCh38
rs75650709 Chr.19: 47751654-47751654 on Build GRCh38
rs762612269 Chr.19: 47751498-47751498 on Build GRCh38
rs906527074 Chr.19: 47751546-47751546 on Build GRCh38
rs781748751 Chr.19: 47751641-47751641 on Build GRCh38
rs765703426 Chr.19: 47751639-47751639 on Build GRCh38
rs892496919 Chr.19: 47751334-47751334 on Build GRCh38
rs939355404 Chr.19: 47751552-47751554 on Build GRCh38
rs948008427 Chr.19: 47751371-47751371 on Build GRCh38
rs766073839 Chr.19: 47751572-47751572 on Build GRCh38
rs965453909 Chr.19: 47751458-47751458 on Build GRCh38
rs759329135 Chr.19: 47751580-47751580 on Build GRCh38
rs927759368 Chr.19: 47751555-47751555 on Build GRCh38
rs755412963 Chr.19: 47751640-47751640 on Build GRCh38
rs769457075 Chr.19: 47751554-47751554 on Build GRCh38
rs780529466 Chr.19: 47751536-47751536 on Build GRCh38
rs140040455 Chr.19: 47751544-47751544 on Build GRCh38
rs754585342 Chr.19: 47751526-47751526 on Build GRCh38
rs1053664865 Chr.19: 47751330-47751330 on Build GRCh38
rs935161412 Chr.19: 47751317-47751317 on Build GRCh38
rs113310396 Chr.19: 47751450-47751450 on Build GRCh38
rs1007729361 Chr.19: 47751408-47751408 on Build GRCh38
rs976497384 Chr.19: 47751471-47751471 on Build GRCh38
rs747469849 Chr.19: 47751539-47751539 on Build GRCh38
rs112625343 Chr.19: 47751452-47751452 on Build GRCh38
rs58520435 Chr.19: 47751453-47751453 on Build GRCh38
rs548451384 Chr.19: 47751577-47751577 on Build GRCh38
rs368749381 Chr.19: 47751476-47751476 on Build GRCh38
rs1045060527 Chr.19: 47751375-47751375 on Build GRCh38
rs764922480 Chr.19: 47751483-47751483 on Build GRCh38
rs1036406955 Chr.19: 47751564-47751564 on Build GRCh38
rs375224226 Chr.19: 47751602-47751602 on Build GRCh38
rs775553343 Chr.19: 47751413-47751413 on Build GRCh38
rs751429514 Chr.19: 47751506-47751506 on Build GRCh38
rs762715929 Chr.19: 47751560-47751560 on Build GRCh38
rs893746397 Chr.19: 47751360-47751360 on Build GRCh38
rs879612714 Chr.19: 47751348-47751348 on Build GRCh38
rs763741683 Chr.19: 47751658-47751658 on Build GRCh38
rs772705450 Chr.19: 47751559-47751559 on Build GRCh38
rs764153370 Chr.19: 47751517-47751517 on Build GRCh38
rs151274684 Chr.19: 47751637-47751637 on Build GRCh38
rs149555718 Chr.19: 47751535-47751535 on Build GRCh38
rs889164793 Chr.19: 47751376-47751376 on Build GRCh38

View Full Product Details