Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9958576-9959104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL1
Entrez Gene ID
29841
Gene Name
grainyhead like transcription factor 1
Gene Aliases
LBP32, MGR, NH32, TFCP2L2
UniGene
Hs.418493
Gene Chromosome Location
Chr.2: 9951663-10002284 on Build GRCh38
Transcripts
NM_198182.2 XM_005246159.3 XM_006711882.2 XM_006711884.3 XM_011510343.2 XM_017003900.1 XM_017003901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1004455 c.215G>A p.R72K Chr.2: 9958793-9958793 on Build GRCh38
1004456 c.231A>G p.S77S Chr.2: 9958809-9958809 on Build GRCh38
3564267 c.211C>T p.P71S Chr.2: 9958789-9958789 on Build GRCh38
4336976 c.213_214insAG p.R75fs*26 Chr.2: 9958791-9958792 on Build GRCh38
3564268 c.211C>T p.P71S Chr.2: 9958789-9958789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022043519 Chr.2: 9958926-9958926 on Build GRCh38
rs757054517 Chr.2: 9958877-9958877 on Build GRCh38
rs373148350 Chr.2: 9958807-9958807 on Build GRCh38
rs369977805 Chr.2: 9958865-9958865 on Build GRCh38
rs367953571 Chr.2: 9958749-9958749 on Build GRCh38
rs753805083 Chr.2: 9958871-9958871 on Build GRCh38
rs939926549 Chr.2: 9959011-9959011 on Build GRCh38
rs750205489 Chr.2: 9958879-9958879 on Build GRCh38
rs200624579 Chr.2: 9958845-9958845 on Build GRCh38
rs916698659 Chr.2: 9958630-9958630 on Build GRCh38
rs916832231 Chr.2: 9958999-9958999 on Build GRCh38
rs371761133 Chr.2: 9958762-9958762 on Build GRCh38
rs924029618 Chr.2: 9958649-9958649 on Build GRCh38
rs1031894088 Chr.2: 9958773-9958773 on Build GRCh38
rs150242900 Chr.2: 9958970-9958970 on Build GRCh38
rs574599452 Chr.2: 9958839-9958839 on Build GRCh38
rs908423397 Chr.2: 9959004-9959004 on Build GRCh38
rs10192887 Chr.2: 9958953-9958953 on Build GRCh38
rs1038528842 Chr.2: 9958617-9958617 on Build GRCh38
rs891047540 Chr.2: 9958743-9958743 on Build GRCh38
rs991013730 Chr.2: 9958618-9958618 on Build GRCh38
rs534808349 Chr.2: 9958642-9958642 on Build GRCh38
rs749099607 Chr.2: 9958881-9958882 on Build GRCh38
rs765521426 Chr.2: 9958757-9958757 on Build GRCh38
rs756070122 Chr.2: 9958736-9958736 on Build GRCh38
rs779739094 Chr.2: 9958774-9958774 on Build GRCh38
rs746517968 Chr.2: 9958904-9958904 on Build GRCh38
rs924038777 Chr.2: 9959078-9959078 on Build GRCh38
rs550498663 Chr.2: 9959071-9959071 on Build GRCh38
rs766540105 Chr.2: 9958846-9958846 on Build GRCh38
rs991519722 Chr.2: 9958994-9958994 on Build GRCh38
rs746466770 Chr.2: 9958783-9958783 on Build GRCh38
rs763144970 Chr.2: 9959021-9959021 on Build GRCh38
rs768274065 Chr.2: 9958905-9958905 on Build GRCh38
rs182949385 Chr.2: 9958963-9958963 on Build GRCh38
rs773353555 Chr.2: 9958840-9958840 on Build GRCh38
rs946904581 Chr.2: 9958632-9958632 on Build GRCh38
rs751653940 Chr.2: 9958850-9958850 on Build GRCh38
rs374807242 Chr.2: 9958741-9958741 on Build GRCh38
rs758142861 Chr.2: 9958891-9958891 on Build GRCh38
rs777708441 Chr.2: 9958739-9958739 on Build GRCh38
rs766641091 Chr.2: 9958660-9958660 on Build GRCh38
rs1038080099 Chr.2: 9959048-9959048 on Build GRCh38
rs779792271 Chr.2: 9958897-9958897 on Build GRCh38
rs935351363 Chr.2: 9958675-9958675 on Build GRCh38
rs771751236 Chr.2: 9958763-9958763 on Build GRCh38
rs1006722640 Chr.2: 9958744-9958744 on Build GRCh38
rs968651552 Chr.2: 9959017-9959017 on Build GRCh38
rs376660067 Chr.2: 9958861-9958861 on Build GRCh38
rs369646093 Chr.2: 9958805-9958805 on Build GRCh38
rs537040826 Chr.2: 9959028-9959028 on Build GRCh38
rs199687923 Chr.2: 9958792-9958792 on Build GRCh38
rs757074237 Chr.2: 9958748-9958748 on Build GRCh38
rs867457481 Chr.2: 9958717-9958717 on Build GRCh38

View Full Product Details