Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31180635-31181131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
-
UniGene
Hs.468059
Gene Chromosome Location
Chr.2: 31173056-31233957 on Build GRCh38
Transcripts
NM_001145122.1 NM_001321270.1 XM_011532863.2 XM_011532864.2 XM_011532865.1 XM_011532866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4530354 c.1690G>A p.G564R Chr.2: 31180956-31180956 on Build GRCh38
2941735 c.1710+1G>A p.? Chr.2: 31180935-31180935 on Build GRCh38
4312775 c.1710+1G>A p.? Chr.2: 31180935-31180935 on Build GRCh38
4530353 c.1690G>A p.G564R Chr.2: 31180956-31180956 on Build GRCh38
4312779 c.1655G>A p.S552N Chr.2: 31180991-31180991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115428238 Chr.2: 31180673-31180673 on Build GRCh38
rs149760139 Chr.2: 31180707-31180707 on Build GRCh38
rs766475493 Chr.2: 31181021-31181021 on Build GRCh38
rs748893120 Chr.2: 31180942-31180942 on Build GRCh38
rs772019423 Chr.2: 31180854-31180854 on Build GRCh38
rs944749269 Chr.2: 31180698-31180698 on Build GRCh38
rs913463697 Chr.2: 31180999-31180999 on Build GRCh38
rs781002406 Chr.2: 31181047-31181047 on Build GRCh38
rs372449814 Chr.2: 31180991-31180991 on Build GRCh38
rs370514331 Chr.2: 31180882-31180882 on Build GRCh38
rs572295701 Chr.2: 31180739-31180739 on Build GRCh38
rs982086416 Chr.2: 31180835-31180835 on Build GRCh38
rs755168692 Chr.2: 31181038-31181038 on Build GRCh38
rs952908108 Chr.2: 31180736-31180736 on Build GRCh38
rs893289134 Chr.2: 31180878-31180892 on Build GRCh38
rs199964386 Chr.2: 31180996-31180996 on Build GRCh38
rs1042620581 Chr.2: 31180958-31180958 on Build GRCh38
rs1035293480 Chr.2: 31181090-31181090 on Build GRCh38
rs540915300 Chr.2: 31180762-31180762 on Build GRCh38
rs73921565 Chr.2: 31181054-31181054 on Build GRCh38
rs1027842390 Chr.2: 31180700-31180700 on Build GRCh38
rs938021045 Chr.2: 31180866-31180866 on Build GRCh38
rs76939962 Chr.2: 31181013-31181013 on Build GRCh38
rs991259862 Chr.2: 31181100-31181100 on Build GRCh38
rs770421855 Chr.2: 31180902-31180902 on Build GRCh38
rs558454988 Chr.2: 31180737-31180737 on Build GRCh38
rs548562879 Chr.2: 31181078-31181078 on Build GRCh38
rs927966114 Chr.2: 31180838-31180838 on Build GRCh38
rs764613923 Chr.2: 31180936-31180936 on Build GRCh38
rs878924278 Chr.2: 31181012-31181012 on Build GRCh38
rs961628477 Chr.2: 31180817-31180817 on Build GRCh38
rs906296159 Chr.2: 31181046-31181046 on Build GRCh38
rs961593805 Chr.2: 31181060-31181060 on Build GRCh38
rs902389007 Chr.2: 31180973-31180973 on Build GRCh38
rs754234522 Chr.2: 31180951-31180951 on Build GRCh38
rs377551095 Chr.2: 31181049-31181049 on Build GRCh38
rs201159308 Chr.2: 31181009-31181009 on Build GRCh38
rs974726578 Chr.2: 31180891-31180891 on Build GRCh38
rs915348830 Chr.2: 31180913-31180913 on Build GRCh38
rs779578145 Chr.2: 31180815-31180815 on Build GRCh38
rs145715824 Chr.2: 31181095-31181095 on Build GRCh38
rs887532555 Chr.2: 31181041-31181041 on Build GRCh38
rs1010394126 Chr.2: 31180758-31180758 on Build GRCh38
rs1005533502 Chr.2: 31180820-31180820 on Build GRCh38
rs751785811 Chr.2: 31181025-31181025 on Build GRCh38
rs1042193447 Chr.2: 31180787-31180787 on Build GRCh38
rs543956945 Chr.2: 31180982-31180982 on Build GRCh38
rs1003219549 Chr.2: 31180800-31180800 on Build GRCh38
rs574506286 Chr.2: 31180850-31180850 on Build GRCh38
rs1027398700 Chr.2: 31181027-31181027 on Build GRCh38
rs773883138 Chr.2: 31180935-31180935 on Build GRCh38
rs762451735 Chr.2: 31180969-31180969 on Build GRCh38
rs2028678 Chr.2: 31181081-31181081 on Build GRCh38
rs773122364 Chr.2: 31181091-31181091 on Build GRCh38
rs116356768 Chr.2: 31180788-31180788 on Build GRCh38
rs984730738 Chr.2: 31180804-31180804 on Build GRCh38
rs995869681 Chr.2: 31180697-31180697 on Build GRCh38

View Full Product Details