Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38068601-38069147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1B1
Entrez Gene ID
1545
Gene Name
cytochrome P450 family 1 subfamily B member 1
Gene Aliases
CP1B, CYPIB1, GLC3A, P4501B1
UniGene
Hs.154654
Gene Chromosome Location
Chr.2: 38067603-38076181 on Build GRCh38
Transcripts
NM_000104.3

SNP Detail

SNP ID Chromosome Location
rs554309996 Chr.2: 38069092-38069092 on Build GRCh38
rs967024648 Chr.2: 38068672-38068672 on Build GRCh38
rs886055990 Chr.2: 38068827-38068827 on Build GRCh38
rs1032646115 Chr.2: 38068947-38068947 on Build GRCh38
rs950607605 Chr.2: 38068697-38068697 on Build GRCh38
rs369078176 Chr.2: 38068955-38068955 on Build GRCh38
rs946262051 Chr.2: 38069075-38069075 on Build GRCh38
rs779407183 Chr.2: 38068937-38068937 on Build GRCh38
rs9341264 Chr.2: 38068918-38068918 on Build GRCh38
rs1030202641 Chr.2: 38068656-38068656 on Build GRCh38
rs533555807 Chr.2: 38068744-38068744 on Build GRCh38
rs750990195 Chr.2: 38068941-38068941 on Build GRCh38
rs1033361792 Chr.2: 38068763-38068769 on Build GRCh38
rs927357675 Chr.2: 38068985-38068985 on Build GRCh38
rs903629356 Chr.2: 38068886-38068886 on Build GRCh38
rs572531079 Chr.2: 38069111-38069111 on Build GRCh38
rs980150505 Chr.2: 38068979-38068979 on Build GRCh38
rs997833781 Chr.2: 38068641-38068641 on Build GRCh38
rs917076692 Chr.2: 38068806-38068806 on Build GRCh38
rs938982236 Chr.2: 38068828-38068828 on Build GRCh38
rs978422251 Chr.2: 38068715-38068715 on Build GRCh38
rs900163227 Chr.2: 38068924-38068924 on Build GRCh38
rs574973848 Chr.2: 38068918-38068918 on Build GRCh38
rs776487480 Chr.2: 38068884-38068884 on Build GRCh38
rs1057504311 Chr.2: 38068842-38068842 on Build GRCh38
rs983212249 Chr.2: 38068733-38068733 on Build GRCh38
rs548724832 Chr.2: 38068714-38068714 on Build GRCh38
rs899419044 Chr.2: 38068888-38068888 on Build GRCh38
rs9341265 Chr.2: 38068869-38068869 on Build GRCh38
rs767403314 Chr.2: 38068761-38068761 on Build GRCh38
rs532324354 Chr.2: 38068963-38068963 on Build GRCh38
rs973844941 Chr.2: 38069118-38069118 on Build GRCh38
rs1008650566 Chr.2: 38068648-38068649 on Build GRCh38
rs992809675 Chr.2: 38069035-38069035 on Build GRCh38
rs34169771 Chr.2: 38068976-38068976 on Build GRCh38
rs397897606 Chr.2: 38068913-38068913 on Build GRCh38
rs34544682 Chr.2: 38068814-38068814 on Build GRCh38
rs991740470 Chr.2: 38068797-38068797 on Build GRCh38
rs747724277 Chr.2: 38068777-38068777 on Build GRCh38
rs189809220 Chr.2: 38069039-38069039 on Build GRCh38
rs1008683161 Chr.2: 38068920-38068920 on Build GRCh38
rs886055991 Chr.2: 38068992-38068992 on Build GRCh38
rs1051073866 Chr.2: 38068914-38068914 on Build GRCh38
rs886055992 Chr.2: 38069121-38069121 on Build GRCh38
rs960317584 Chr.2: 38069030-38069030 on Build GRCh38
rs982553206 Chr.2: 38069034-38069034 on Build GRCh38
rs982148193 Chr.2: 38068827-38068827 on Build GRCh38
rs570132783 Chr.2: 38068784-38068784 on Build GRCh38
rs1000604188 Chr.2: 38068913-38068913 on Build GRCh38
rs950631916 Chr.2: 38068943-38068943 on Build GRCh38
rs978953402 Chr.2: 38069098-38069098 on Build GRCh38
rs538184854 Chr.2: 38068862-38068862 on Build GRCh38
rs991402879 Chr.2: 38068972-38068972 on Build GRCh38
rs1019987880 Chr.2: 38068658-38068658 on Build GRCh38
rs546233182 Chr.2: 38069115-38069115 on Build GRCh38
rs4630747 Chr.2: 38069113-38069113 on Build GRCh38
rs9341266 Chr.2: 38068851-38068851 on Build GRCh38
rs913445988 Chr.2: 38068843-38068843 on Build GRCh38

View Full Product Details