Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 41952846-41953383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf91
Entrez Gene ID
400950
Gene Name
chromosome 2 open reading frame 91
Gene Aliases
-
UniGene
Hs.738713
Gene Chromosome Location
Chr.2: 41935368-41961382 on Build GRCh38
Transcripts
NM_001242815.1 XM_017004097.1

SNP Detail

SNP ID Chromosome Location
rs191678080 Chr.2: 41953350-41953350 on Build GRCh38
rs1044208690 Chr.2: 41952880-41952880 on Build GRCh38
rs891302957 Chr.2: 41953269-41953269 on Build GRCh38
rs200161608 Chr.2: 41953045-41953045 on Build GRCh38
rs767679064 Chr.2: 41953296-41953296 on Build GRCh38
rs772035399 Chr.2: 41953087-41953087 on Build GRCh38
rs760766486 Chr.2: 41953343-41953343 on Build GRCh38
rs979830653 Chr.2: 41953290-41953290 on Build GRCh38
rs1052292481 Chr.2: 41952896-41952896 on Build GRCh38
rs901073764 Chr.2: 41953243-41953243 on Build GRCh38
rs768599560 Chr.2: 41953246-41953246 on Build GRCh38
rs900812016 Chr.2: 41952970-41952970 on Build GRCh38
rs140518196 Chr.2: 41953284-41953284 on Build GRCh38
rs776285739 Chr.2: 41953180-41953180 on Build GRCh38
rs199686441 Chr.2: 41952919-41952919 on Build GRCh38
rs555675285 Chr.2: 41953334-41953334 on Build GRCh38
rs1025729374 Chr.2: 41953266-41953266 on Build GRCh38
rs746720399 Chr.2: 41953126-41953126 on Build GRCh38
rs917720813 Chr.2: 41953331-41953331 on Build GRCh38
rs563688485 Chr.2: 41953022-41953022 on Build GRCh38
rs935456575 Chr.2: 41953144-41953144 on Build GRCh38
rs746840721 Chr.2: 41953323-41953330 on Build GRCh38
rs1053611503 Chr.2: 41952904-41952904 on Build GRCh38
rs1044726791 Chr.2: 41952944-41952944 on Build GRCh38
rs34271023 Chr.2: 41953044-41953045 on Build GRCh38
rs893653191 Chr.2: 41952936-41952936 on Build GRCh38
rs925342946 Chr.2: 41953129-41953129 on Build GRCh38
rs151243275 Chr.2: 41953212-41953212 on Build GRCh38
rs980107168 Chr.2: 41953135-41953135 on Build GRCh38
rs558847689 Chr.2: 41953348-41953348 on Build GRCh38
rs927324544 Chr.2: 41953155-41953155 on Build GRCh38
rs756313354 Chr.2: 41952961-41952961 on Build GRCh38
rs150427867 Chr.2: 41953306-41953306 on Build GRCh38
rs971670155 Chr.2: 41953322-41953322 on Build GRCh38
rs1028432134 Chr.2: 41952995-41952995 on Build GRCh38
rs998551691 Chr.2: 41953247-41953247 on Build GRCh38
rs376589951 Chr.2: 41953201-41953201 on Build GRCh38
rs994732129 Chr.2: 41952990-41952990 on Build GRCh38
rs950379302 Chr.2: 41953108-41953108 on Build GRCh38
rs896388697 Chr.2: 41952902-41952902 on Build GRCh38
rs569340488 Chr.2: 41953297-41953297 on Build GRCh38
rs539164915 Chr.2: 41953259-41953259 on Build GRCh38
rs1044846158 Chr.2: 41952876-41952876 on Build GRCh38
rs911189916 Chr.2: 41953027-41953027 on Build GRCh38
rs760962224 Chr.2: 41953281-41953281 on Build GRCh38
rs941688216 Chr.2: 41953242-41953242 on Build GRCh38
rs760013735 Chr.2: 41953228-41953228 on Build GRCh38
rs201332915 Chr.2: 41953173-41953173 on Build GRCh38
rs547170116 Chr.2: 41953211-41953211 on Build GRCh38
rs997851028 Chr.2: 41952988-41952988 on Build GRCh38
rs749670898 Chr.2: 41952964-41952964 on Build GRCh38
rs961525502 Chr.2: 41953046-41953046 on Build GRCh38
rs775405743 Chr.2: 41953099-41953099 on Build GRCh38
rs761247575 Chr.2: 41953190-41953190 on Build GRCh38
rs753056453 Chr.2: 41953249-41953249 on Build GRCh38
rs961858619 Chr.2: 41952975-41952975 on Build GRCh38
rs897542262 Chr.2: 41953263-41953263 on Build GRCh38
rs564649586 Chr.2: 41952882-41952882 on Build GRCh38
rs767926092 Chr.2: 41953230-41953230 on Build GRCh38
rs529582911 Chr.2: 41953035-41953035 on Build GRCh38
rs187118302 Chr.2: 41952939-41952939 on Build GRCh38
rs993226810 Chr.2: 41953265-41953265 on Build GRCh38
rs777895742 Chr.2: 41953009-41953009 on Build GRCh38
rs988225373 Chr.2: 41953321-41953321 on Build GRCh38
rs936066255 Chr.2: 41953171-41953171 on Build GRCh38
rs143623092 Chr.2: 41953072-41953072 on Build GRCh38
rs927278949 Chr.2: 41953330-41953331 on Build GRCh38
rs752964570 Chr.2: 41952956-41952956 on Build GRCh38
rs1024451151 Chr.2: 41953321-41953322 on Build GRCh38
rs559675009 Chr.2: 41953057-41953057 on Build GRCh38
rs917512174 Chr.2: 41953083-41953083 on Build GRCh38
rs980770874 Chr.2: 41953326-41953326 on Build GRCh38
rs757304967 Chr.2: 41953043-41953043 on Build GRCh38
rs1016418841 Chr.2: 41952972-41952972 on Build GRCh38
rs936106184 Chr.2: 41952898-41952898 on Build GRCh38
rs955598247 Chr.2: 41953013-41953013 on Build GRCh38
rs1009448775 Chr.2: 41952948-41952948 on Build GRCh38
rs904858675 Chr.2: 41952945-41952945 on Build GRCh38
rs112075586 Chr.2: 41952883-41952883 on Build GRCh38
rs964352543 Chr.2: 41953285-41953285 on Build GRCh38
rs953819960 Chr.2: 41952998-41952998 on Build GRCh38

View Full Product Details