Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70790290-70790825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGLA
Entrez Gene ID
344018
Gene Name
folliculogenesis specific bHLH transcription factor
Gene Aliases
BHLHC8, FIGALPHA, POF6
UniGene
Hs.407636
Gene Chromosome Location
Chr.2: 70777310-70790643 on Build GRCh38
Transcripts
NM_001004311.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369193 c.62C>G p.P21R Chr.2: 70790577-70790577 on Build GRCh38
6334699 c.103G>C p.G35R Chr.2: 70790536-70790536 on Build GRCh38
5032749 c.85G>C p.V29L Chr.2: 70790554-70790554 on Build GRCh38
6043365 c.134G>A p.R45Q Chr.2: 70790505-70790505 on Build GRCh38
5690534 c.207G>T p.V69V Chr.2: 70790432-70790432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891605060 Chr.2: 70790566-70790566 on Build GRCh38
rs1021787303 Chr.2: 70790611-70790611 on Build GRCh38
rs969721494 Chr.2: 70790621-70790621 on Build GRCh38
rs373561603 Chr.2: 70790555-70790555 on Build GRCh38
rs71647803 Chr.2: 70790628-70790628 on Build GRCh38
rs528824825 Chr.2: 70790766-70790766 on Build GRCh38
rs587776535 Chr.2: 70790603-70790624 on Build GRCh38
rs781794080 Chr.2: 70790393-70790393 on Build GRCh38
rs538528747 Chr.2: 70790377-70790377 on Build GRCh38
rs932619537 Chr.2: 70790497-70790497 on Build GRCh38
rs908747634 Chr.2: 70790650-70790650 on Build GRCh38
rs960975399 Chr.2: 70790643-70790643 on Build GRCh38
rs782545480 Chr.2: 70790432-70790432 on Build GRCh38
rs908689480 Chr.2: 70790467-70790467 on Build GRCh38
rs540242376 Chr.2: 70790691-70790691 on Build GRCh38
rs77007852 Chr.2: 70790337-70790337 on Build GRCh38
rs996756609 Chr.2: 70790329-70790329 on Build GRCh38
rs569032899 Chr.2: 70790389-70790389 on Build GRCh38
rs943981542 Chr.2: 70790716-70790716 on Build GRCh38
rs947176606 Chr.2: 70790747-70790747 on Build GRCh38
rs12713717 Chr.2: 70790384-70790384 on Build GRCh38
rs749136361 Chr.2: 70790761-70790761 on Build GRCh38
rs781849225 Chr.2: 70790426-70790426 on Build GRCh38
rs553064465 Chr.2: 70790333-70790333 on Build GRCh38
rs544015252 Chr.2: 70790461-70790461 on Build GRCh38
rs782199264 Chr.2: 70790470-70790470 on Build GRCh38
rs974192635 Chr.2: 70790659-70790659 on Build GRCh38
rs782763820 Chr.2: 70790376-70790376 on Build GRCh38
rs782037656 Chr.2: 70790385-70790385 on Build GRCh38
rs782214626 Chr.2: 70790588-70790588 on Build GRCh38
rs992297731 Chr.2: 70790442-70790442 on Build GRCh38
rs782637050 Chr.2: 70790625-70790625 on Build GRCh38
rs374864485 Chr.2: 70790316-70790316 on Build GRCh38
rs940141102 Chr.2: 70790474-70790474 on Build GRCh38
rs961833537 Chr.2: 70790657-70790657 on Build GRCh38
rs992800088 Chr.2: 70790647-70790647 on Build GRCh38
rs919971886 Chr.2: 70790688-70790688 on Build GRCh38
rs573545377 Chr.2: 70790340-70790340 on Build GRCh38
rs919972350 Chr.2: 70790486-70790486 on Build GRCh38
rs932671902 Chr.2: 70790699-70790699 on Build GRCh38
rs1052988363 Chr.2: 70790560-70790560 on Build GRCh38
rs1001164504 Chr.2: 70790637-70790637 on Build GRCh38
rs782606232 Chr.2: 70790487-70790487 on Build GRCh38
rs1053442613 Chr.2: 70790733-70790733 on Build GRCh38
rs554656682 Chr.2: 70790485-70790485 on Build GRCh38
rs782665860 Chr.2: 70790450-70790450 on Build GRCh38
rs926829367 Chr.2: 70790402-70790402 on Build GRCh38
rs1038856753 Chr.2: 70790483-70790483 on Build GRCh38
rs891017930 Chr.2: 70790542-70790542 on Build GRCh38
rs1033951618 Chr.2: 70790638-70790638 on Build GRCh38
rs781954824 Chr.2: 70790363-70790363 on Build GRCh38
rs1049633950 Chr.2: 70790511-70790511 on Build GRCh38
rs1049689794 Chr.2: 70790703-70790703 on Build GRCh38
rs769370684 Chr.2: 70790349-70790349 on Build GRCh38
rs782787640 Chr.2: 70790420-70790420 on Build GRCh38
rs782387486 Chr.2: 70790593-70790593 on Build GRCh38
rs912551256 Chr.2: 70790714-70790714 on Build GRCh38
rs1011364971 Chr.2: 70790581-70790581 on Build GRCh38
rs782459218 Chr.2: 70790400-70790400 on Build GRCh38
rs575091555 Chr.2: 70790459-70790459 on Build GRCh38
rs560153492 Chr.2: 70790738-70790738 on Build GRCh38

View Full Product Details