Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73258112-73258607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
UniGene
Hs.23158
Gene Chromosome Location
Chr.2: 73254682-73284453 on Build GRCh38
Transcripts
NM_001080410.2 XM_005264158.3 XM_006711950.2 XM_011532562.1 XM_011532563.2 XM_017003421.1 XM_017003422.1 XM_017003423.1

SNP Detail

SNP ID Chromosome Location
rs534167655 Chr.2: 73258587-73258587 on Build GRCh38
rs576683967 Chr.2: 73258150-73258150 on Build GRCh38
rs941698815 Chr.2: 73258287-73258287 on Build GRCh38
rs989407422 Chr.2: 73258283-73258283 on Build GRCh38
rs1028161250 Chr.2: 73258421-73258421 on Build GRCh38
rs1040458537 Chr.2: 73258372-73258372 on Build GRCh38
rs536426462 Chr.2: 73258134-73258134 on Build GRCh38
rs1020775391 Chr.2: 73258529-73258529 on Build GRCh38
rs933228796 Chr.2: 73258351-73258351 on Build GRCh38
rs902321521 Chr.2: 73258476-73258476 on Build GRCh38
rs982300295 Chr.2: 73258291-73258291 on Build GRCh38
rs866786695 Chr.2: 73258568-73258568 on Build GRCh38
rs1009273597 Chr.2: 73258137-73258137 on Build GRCh38
rs375565432 Chr.2: 73258507-73258507 on Build GRCh38
rs559003005 Chr.2: 73258202-73258202 on Build GRCh38
rs983231270 Chr.2: 73258233-73258233 on Build GRCh38
rs1035842315 Chr.2: 73258164-73258164 on Build GRCh38
rs996144937 Chr.2: 73258396-73258396 on Build GRCh38
rs772914469 Chr.2: 73258284-73258284 on Build GRCh38
rs1008969892 Chr.2: 73258371-73258371 on Build GRCh38
rs982031683 Chr.2: 73258161-73258161 on Build GRCh38
rs957978658 Chr.2: 73258220-73258220 on Build GRCh38
rs957981236 Chr.2: 73258462-73258462 on Build GRCh38
rs559877104 Chr.2: 73258524-73258524 on Build GRCh38
rs1011563615 Chr.2: 73258554-73258554 on Build GRCh38
rs927712397 Chr.2: 73258565-73258565 on Build GRCh38
rs555269044 Chr.2: 73258145-73258145 on Build GRCh38
rs1018994519 Chr.2: 73258149-73258149 on Build GRCh38
rs958994434 Chr.2: 73258579-73258579 on Build GRCh38
rs900485694 Chr.2: 73258384-73258384 on Build GRCh38
rs908957880 Chr.2: 73258234-73258234 on Build GRCh38
rs920855445 Chr.2: 73258321-73258321 on Build GRCh38
rs930767894 Chr.2: 73258327-73258327 on Build GRCh38
rs975235713 Chr.2: 73258201-73258201 on Build GRCh38
rs990488757 Chr.2: 73258586-73258586 on Build GRCh38
rs937680716 Chr.2: 73258308-73258308 on Build GRCh38
rs1052035904 Chr.2: 73258373-73258373 on Build GRCh38
rs1053170738 Chr.2: 73258531-73258531 on Build GRCh38
rs147910875 Chr.2: 73258528-73258528 on Build GRCh38
rs480538 Chr.2: 73258390-73258390 on Build GRCh38
rs760447370 Chr.2: 73258309-73258309 on Build GRCh38
rs541588023 Chr.2: 73258380-73258380 on Build GRCh38
rs771645211 Chr.2: 73258169-73258169 on Build GRCh38
rs893205265 Chr.2: 73258552-73258552 on Build GRCh38
rs970323697 Chr.2: 73258141-73258141 on Build GRCh38
rs886641118 Chr.2: 73258341-73258341 on Build GRCh38
rs544061982 Chr.2: 73258191-73258191 on Build GRCh38
rs372281332 Chr.2: 73258306-73258306 on Build GRCh38
rs1047887869 Chr.2: 73258332-73258332 on Build GRCh38

View Full Product Details