Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73293094-73293589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR4
Entrez Gene ID
1961
Gene Name
early growth response 4
Gene Aliases
NGFI-C, NGFIC, PAT133
UniGene
Hs.3052
Gene Chromosome Location
Chr.2: 73290929-73293701 on Build GRCh38
Transcripts
NM_001965.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251026 c.315C>T p.L105L Chr.2: 73293312-73293312 on Build GRCh38
6421083 c.25G>T p.E9* Chr.2: 73293293-73293293 on Build GRCh38
6427602 c.115G>T p.A39S Chr.2: 73293203-73293203 on Build GRCh38
1205066 c.425C>T p.A142V Chr.2: 73293202-73293202 on Build GRCh38
3659946 c.298C>T p.R100W Chr.2: 73293329-73293329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748358260 Chr.2: 73293137-73293137 on Build GRCh38
rs773172505 Chr.2: 73293362-73293362 on Build GRCh38
rs986556118 Chr.2: 73293381-73293381 on Build GRCh38
rs771943471 Chr.2: 73293359-73293359 on Build GRCh38
rs747196821 Chr.2: 73293344-73293344 on Build GRCh38
rs779302657 Chr.2: 73293238-73293238 on Build GRCh38
rs747145988 Chr.2: 73293163-73293163 on Build GRCh38
rs756320734 Chr.2: 73293231-73293232 on Build GRCh38
rs760423874 Chr.2: 73293300-73293300 on Build GRCh38
rs765718128 Chr.2: 73293337-73293337 on Build GRCh38
rs762823394 Chr.2: 73293186-73293186 on Build GRCh38
rs768526709 Chr.2: 73293282-73293282 on Build GRCh38
rs774182958 Chr.2: 73293283-73293283 on Build GRCh38
rs1043972656 Chr.2: 73293227-73293227 on Build GRCh38
rs370512012 Chr.2: 73293248-73293248 on Build GRCh38
rs1033194822 Chr.2: 73293536-73293536 on Build GRCh38
rs748058847 Chr.2: 73293459-73293459 on Build GRCh38
rs765998769 Chr.2: 73293227-73293227 on Build GRCh38
rs755467704 Chr.2: 73293233-73293233 on Build GRCh38
rs550238849 Chr.2: 73293564-73293564 on Build GRCh38
rs919148649 Chr.2: 73293264-73293264 on Build GRCh38
rs967616072 Chr.2: 73293349-73293349 on Build GRCh38
rs759031935 Chr.2: 73293374-73293374 on Build GRCh38
rs527893278 Chr.2: 73293357-73293357 on Build GRCh38
rs746854936 Chr.2: 73293476-73293476 on Build GRCh38
rs896632601 Chr.2: 73293515-73293515 on Build GRCh38
rs555396652 Chr.2: 73293212-73293212 on Build GRCh38
rs969398077 Chr.2: 73293119-73293119 on Build GRCh38
rs745993894 Chr.2: 73293353-73293353 on Build GRCh38
rs768636423 Chr.2: 73293355-73293355 on Build GRCh38
rs145377731 Chr.2: 73293189-73293189 on Build GRCh38
rs942800497 Chr.2: 73293444-73293444 on Build GRCh38
rs537734263 Chr.2: 73293568-73293568 on Build GRCh38
rs771189047 Chr.2: 73293561-73293562 on Build GRCh38
rs779166677 Chr.2: 73293132-73293132 on Build GRCh38
rs780202647 Chr.2: 73293453-73293453 on Build GRCh38
rs751970791 Chr.2: 73293402-73293402 on Build GRCh38
rs1000417794 Chr.2: 73293529-73293529 on Build GRCh38
rs112387203 Chr.2: 73293393-73293393 on Build GRCh38
rs749947757 Chr.2: 73293200-73293200 on Build GRCh38
rs749219249 Chr.2: 73293276-73293276 on Build GRCh38
rs375301544 Chr.2: 73293188-73293188 on Build GRCh38
rs1018273308 Chr.2: 73293158-73293158 on Build GRCh38
rs1027049024 Chr.2: 73293120-73293120 on Build GRCh38
rs748351654 Chr.2: 73293229-73293231 on Build GRCh38
rs373746151 Chr.2: 73293147-73293147 on Build GRCh38
rs764819595 Chr.2: 73293377-73293377 on Build GRCh38
rs543174763 Chr.2: 73293373-73293373 on Build GRCh38
rs1040117991 Chr.2: 73293319-73293319 on Build GRCh38
rs543817678 Chr.2: 73293273-73293273 on Build GRCh38
rs749392575 Chr.2: 73293517-73293518 on Build GRCh38
rs140526933 Chr.2: 73293255-73293255 on Build GRCh38
rs772318013 Chr.2: 73293139-73293139 on Build GRCh38
rs567576832 Chr.2: 73293436-73293436 on Build GRCh38
rs753156729 Chr.2: 73293388-73293388 on Build GRCh38
rs978912217 Chr.2: 73293216-73293216 on Build GRCh38
rs781310010 Chr.2: 73293345-73293345 on Build GRCh38
rs1024161119 Chr.2: 73293273-73293275 on Build GRCh38
rs561568849 Chr.2: 73293413-73293413 on Build GRCh38
rs775445367 Chr.2: 73293173-73293173 on Build GRCh38
rs776189552 Chr.2: 73293370-73293370 on Build GRCh38
rs748108134 Chr.2: 73293358-73293358 on Build GRCh38
rs763487015 Chr.2: 73293391-73293391 on Build GRCh38
rs972610149 Chr.2: 73293154-73293154 on Build GRCh38
rs550654844 Chr.2: 73293127-73293127 on Build GRCh38
rs770535679 Chr.2: 73293369-73293369 on Build GRCh38
rs533954745 Chr.2: 73293187-73293187 on Build GRCh38
rs764415096 Chr.2: 73293394-73293394 on Build GRCh38
rs958441651 Chr.2: 73293232-73293232 on Build GRCh38
rs377552654 Chr.2: 73293171-73293171 on Build GRCh38
rs777841252 Chr.2: 73293504-73293505 on Build GRCh38
rs865991064 Chr.2: 73293230-73293230 on Build GRCh38
rs868088321 Chr.2: 73293553-73293553 on Build GRCh38
rs377352409 Chr.2: 73293240-73293240 on Build GRCh38
rs760368747 Chr.2: 73293368-73293368 on Build GRCh38
rs370494643 Chr.2: 73293339-73293339 on Build GRCh38
rs971544922 Chr.2: 73293566-73293566 on Build GRCh38
rs754319562 Chr.2: 73293229-73293229 on Build GRCh38
rs1013484984 Chr.2: 73293545-73293545 on Build GRCh38
rs998232165 Chr.2: 73293498-73293498 on Build GRCh38
rs755485169 Chr.2: 73293334-73293334 on Build GRCh38
rs777684527 Chr.2: 73293470-73293470 on Build GRCh38
rs756088470 Chr.2: 73293450-73293450 on Build GRCh38
rs773249919 Chr.2: 73293152-73293152 on Build GRCh38
rs772174430 Chr.2: 73293461-73293461 on Build GRCh38
rs969344128 Chr.2: 73293326-73293326 on Build GRCh38
rs972125480 Chr.2: 73293384-73293384 on Build GRCh38
rs772941753 Chr.2: 73293294-73293294 on Build GRCh38
rs910987900 Chr.2: 73293399-73293399 on Build GRCh38
rs1033139507 Chr.2: 73293363-73293363 on Build GRCh38
rs1043416897 Chr.2: 73293437-73293437 on Build GRCh38
rs767423505 Chr.2: 73293288-73293288 on Build GRCh38
rs1048871155 Chr.2: 73293281-73293281 on Build GRCh38
rs776077634 Chr.2: 73293543-73293543 on Build GRCh38
rs925727029 Chr.2: 73293218-73293218 on Build GRCh38
rs1024085391 Chr.2: 73293563-73293563 on Build GRCh38
rs151267217 Chr.2: 73293226-73293226 on Build GRCh38
rs747250056 Chr.2: 73293251-73293251 on Build GRCh38
rs1042108415 Chr.2: 73293518-73293518 on Build GRCh38
rs770610042 Chr.2: 73293527-73293527 on Build GRCh38
rs1045124670 Chr.2: 73293531-73293531 on Build GRCh38
rs374534321 Chr.2: 73293343-73293343 on Build GRCh38
rs555733817 Chr.2: 73293307-73293307 on Build GRCh38
rs140456361 Chr.2: 73293285-73293285 on Build GRCh38
rs750664156 Chr.2: 73293449-73293449 on Build GRCh38
rs925494152 Chr.2: 73293443-73293443 on Build GRCh38
rs999658445 Chr.2: 73293231-73293231 on Build GRCh38
rs577366080 Chr.2: 73293329-73293329 on Build GRCh38
rs905302700 Chr.2: 73293541-73293541 on Build GRCh38

View Full Product Details