Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101355084-101355498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREG2
Entrez Gene ID
200407
Gene Name
cellular repressor of E1A stimulated genes 2
Gene Aliases
-
UniGene
Hs.732289
Gene Chromosome Location
Chr.2: 101348354-101387970 on Build GRCh38
Transcripts
NM_153836.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635389 c.676G>A p.A226T Chr.2: 101355302-101355302 on Build GRCh38
4572094 c.643T>A p.C215S Chr.2: 101355335-101355335 on Build GRCh38
7081454 c.612-1G>A p.? Chr.2: 101355367-101355367 on Build GRCh38
1249114 c.692_694delAAG p.E231delE Chr.2: 101355284-101355286 on Build GRCh38
5017138 c.656C>T p.T219M Chr.2: 101355322-101355322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755412612 Chr.2: 101355285-101355285 on Build GRCh38
rs139294792 Chr.2: 101355141-101355141 on Build GRCh38
rs779376765 Chr.2: 101355347-101355347 on Build GRCh38
rs780554306 Chr.2: 101355225-101355225 on Build GRCh38
rs770928304 Chr.2: 101355260-101355260 on Build GRCh38
rs201819390 Chr.2: 101355241-101355241 on Build GRCh38
rs370834911 Chr.2: 101355159-101355159 on Build GRCh38
rs6753727 Chr.2: 101355156-101355156 on Build GRCh38
rs765269439 Chr.2: 101355263-101355263 on Build GRCh38
rs769029048 Chr.2: 101355238-101355238 on Build GRCh38
rs763655770 Chr.2: 101355352-101355352 on Build GRCh38
rs145267496 Chr.2: 101355302-101355302 on Build GRCh38
rs758858988 Chr.2: 101355411-101355411 on Build GRCh38
rs572302493 Chr.2: 101355167-101355167 on Build GRCh38
rs151160465 Chr.2: 101355356-101355356 on Build GRCh38
rs990224882 Chr.2: 101355180-101355180 on Build GRCh38
rs957875845 Chr.2: 101355450-101355452 on Build GRCh38
rs754325557 Chr.2: 101355273-101355273 on Build GRCh38
rs776563894 Chr.2: 101355261-101355261 on Build GRCh38
rs1040819677 Chr.2: 101355464-101355464 on Build GRCh38
rs373073657 Chr.2: 101355292-101355292 on Build GRCh38
rs751219780 Chr.2: 101355330-101355330 on Build GRCh38
rs552970133 Chr.2: 101355460-101355489 on Build GRCh38
rs564396267 Chr.2: 101355247-101355247 on Build GRCh38
rs200465701 Chr.2: 101355269-101355269 on Build GRCh38
rs982688169 Chr.2: 101355418-101355418 on Build GRCh38
rs760729104 Chr.2: 101355253-101355253 on Build GRCh38
rs771028344 Chr.2: 101355375-101355375 on Build GRCh38
rs767595597 Chr.2: 101355411-101355417 on Build GRCh38
rs958121009 Chr.2: 101355165-101355165 on Build GRCh38
rs894886268 Chr.2: 101355343-101355343 on Build GRCh38
rs776595926 Chr.2: 101355312-101355312 on Build GRCh38
rs781544858 Chr.2: 101355389-101355389 on Build GRCh38
rs967748223 Chr.2: 101355284-101355284 on Build GRCh38
rs767244146 Chr.2: 101355333-101355333 on Build GRCh38
rs966592667 Chr.2: 101355137-101355137 on Build GRCh38
rs943666028 Chr.2: 101355449-101355449 on Build GRCh38
rs755714159 Chr.2: 101355340-101355340 on Build GRCh38
rs563519121 Chr.2: 101355348-101355348 on Build GRCh38
rs755179368 Chr.2: 101355361-101355361 on Build GRCh38
rs762780528 Chr.2: 101355321-101355321 on Build GRCh38
rs749768170 Chr.2: 101355227-101355227 on Build GRCh38
rs756723041 Chr.2: 101355272-101355272 on Build GRCh38
rs976070973 Chr.2: 101355284-101355286 on Build GRCh38
rs763988406 Chr.2: 101355322-101355322 on Build GRCh38
rs759702151 Chr.2: 101355313-101355313 on Build GRCh38
rs577970303 Chr.2: 101355303-101355303 on Build GRCh38
rs752511188 Chr.2: 101355264-101355264 on Build GRCh38
rs756776478 Chr.2: 101355224-101355224 on Build GRCh38
rs902191911 Chr.2: 101355382-101355382 on Build GRCh38
rs775628443 Chr.2: 101355320-101355320 on Build GRCh38
rs773487553 Chr.2: 101355252-101355252 on Build GRCh38
rs745923249 Chr.2: 101355392-101355392 on Build GRCh38
rs149067073 Chr.2: 101355332-101355332 on Build GRCh38
rs747205559 Chr.2: 101355371-101355371 on Build GRCh38
rs1029102242 Chr.2: 101355158-101355158 on Build GRCh38
rs780659437 Chr.2: 101355309-101355309 on Build GRCh38
rs777608891 Chr.2: 101355212-101355212 on Build GRCh38
rs769614780 Chr.2: 101355318-101355318 on Build GRCh38
rs751272856 Chr.2: 101355271-101355271 on Build GRCh38
rs549498637 Chr.2: 101355430-101355430 on Build GRCh38
rs893411605 Chr.2: 101355173-101355173 on Build GRCh38
rs1013323058 Chr.2: 101355349-101355349 on Build GRCh38
rs749865570 Chr.2: 101355334-101355334 on Build GRCh38
rs763827124 Chr.2: 101355270-101355270 on Build GRCh38
rs748505516 Chr.2: 101355300-101355300 on Build GRCh38
rs775225542 Chr.2: 101355149-101355149 on Build GRCh38
rs367585203 Chr.2: 101355213-101355213 on Build GRCh38
rs778141481 Chr.2: 101355369-101355369 on Build GRCh38
rs759422727 Chr.2: 101355262-101355262 on Build GRCh38

View Full Product Details