Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102443071-102443473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18RAP
Entrez Gene ID
8807
Gene Name
interleukin 18 receptor accessory protein
Gene Aliases
ACPL, CD218b, CDw218b, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1R-7, IL-1R7, IL-1RAcPL, IL18RB
UniGene
Hs.158315
Gene Chromosome Location
Chr.2: 102418551-102452568 on Build GRCh38
Transcripts
NM_003853.3 XM_011512087.2 XM_011512088.2 XM_017005173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4083882 c.881C>A p.S294Y Chr.2: 102443284-102443284 on Build GRCh38
3797801 c.915G>A p.A305A Chr.2: 102443318-102443318 on Build GRCh38
5353951 c.920+6delA p.? Chr.2: 102443329-102443329 on Build GRCh38
5502507 c.832G>T p.G278C Chr.2: 102443235-102443235 on Build GRCh38
6685047 c.846C>A p.V282V Chr.2: 102443249-102443249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776733560 Chr.2: 102443237-102443237 on Build GRCh38
rs772680516 Chr.2: 102443329-102443329 on Build GRCh38
rs748318174 Chr.2: 102443321-102443321 on Build GRCh38
rs148917849 Chr.2: 102443317-102443317 on Build GRCh38
rs779394612 Chr.2: 102443318-102443318 on Build GRCh38
rs772380876 Chr.2: 102443335-102443335 on Build GRCh38
rs747791236 Chr.2: 102443319-102443319 on Build GRCh38
rs1026105709 Chr.2: 102443356-102443356 on Build GRCh38
rs984747432 Chr.2: 102443389-102443389 on Build GRCh38
rs894495563 Chr.2: 102443438-102443438 on Build GRCh38
rs542593932 Chr.2: 102443126-102443126 on Build GRCh38
rs1055777606 Chr.2: 102443395-102443395 on Build GRCh38
rs777347783 Chr.2: 102443171-102443171 on Build GRCh38
rs1052802958 Chr.2: 102443150-102443150 on Build GRCh38
rs762724296 Chr.2: 102443278-102443278 on Build GRCh38
rs778959766 Chr.2: 102443218-102443218 on Build GRCh38
rs910901699 Chr.2: 102443247-102443247 on Build GRCh38
rs756587246 Chr.2: 102443307-102443307 on Build GRCh38
rs200951162 Chr.2: 102443340-102443340 on Build GRCh38
rs181050185 Chr.2: 102443358-102443358 on Build GRCh38
rs1558651 Chr.2: 102443153-102443153 on Build GRCh38
rs769460898 Chr.2: 102443374-102443374 on Build GRCh38
rs772036162 Chr.2: 102443227-102443227 on Build GRCh38
rs911342203 Chr.2: 102443097-102443097 on Build GRCh38
rs942417922 Chr.2: 102443256-102443256 on Build GRCh38
rs377714979 Chr.2: 102443156-102443156 on Build GRCh38
rs752231922 Chr.2: 102443274-102443274 on Build GRCh38
rs748417529 Chr.2: 102443219-102443219 on Build GRCh38
rs1002821916 Chr.2: 102443384-102443384 on Build GRCh38
rs759530413 Chr.2: 102443372-102443372 on Build GRCh38
rs771041735 Chr.2: 102443235-102443235 on Build GRCh38
rs765083650 Chr.2: 102443264-102443264 on Build GRCh38
rs911493068 Chr.2: 102443426-102443426 on Build GRCh38
rs747037035 Chr.2: 102443348-102443348 on Build GRCh38
rs141220224 Chr.2: 102443349-102443349 on Build GRCh38
rs942893360 Chr.2: 102443099-102443099 on Build GRCh38
rs749442932 Chr.2: 102443188-102443188 on Build GRCh38
rs376399504 Chr.2: 102443314-102443314 on Build GRCh38
rs751105813 Chr.2: 102443299-102443299 on Build GRCh38
rs765914189 Chr.2: 102443282-102443282 on Build GRCh38
rs148553198 Chr.2: 102443229-102443229 on Build GRCh38
rs769486453 Chr.2: 102443328-102443329 on Build GRCh38
rs769084327 Chr.2: 102443207-102443207 on Build GRCh38
rs759213993 Chr.2: 102443246-102443246 on Build GRCh38
rs201331437 Chr.2: 102443294-102443294 on Build GRCh38
rs984303826 Chr.2: 102443096-102443096 on Build GRCh38
rs199675556 Chr.2: 102443204-102443204 on Build GRCh38
rs746361292 Chr.2: 102443179-102443179 on Build GRCh38
rs575217199 Chr.2: 102443103-102443103 on Build GRCh38
rs901613276 Chr.2: 102443122-102443122 on Build GRCh38
rs756679318 Chr.2: 102443181-102443181 on Build GRCh38
rs370355318 Chr.2: 102443198-102443198 on Build GRCh38

View Full Product Details