Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131055182-131055691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168B
Entrez Gene ID
130074
Gene Name
family with sequence similarity 168 member B
Gene Aliases
MANI
UniGene
Hs.534679
Gene Chromosome Location
Chr.2: 131047876-131093468 on Build GRCh38
Transcripts
NM_001009993.3 NM_001321743.1 NM_001321744.1 NM_001321745.1 NM_001321746.1 NM_001321747.1 NM_001321748.1 XM_011510594.2 XM_017003327.1 XM_017003328.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1006539 c.437T>C p.M146T Chr.2: 131055310-131055310 on Build GRCh38
1006540 c.393G>A p.V131V Chr.2: 131055354-131055354 on Build GRCh38
1006541 c.331G>T p.A111S Chr.2: 131055416-131055416 on Build GRCh38
1006542 c.268G>T p.A90S Chr.2: 131055582-131055582 on Build GRCh38
4852939 c.163C>G p.P55A Chr.2: 131055687-131055687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975480717 Chr.2: 131055454-131055454 on Build GRCh38
rs772741314 Chr.2: 131055223-131055223 on Build GRCh38
rs555047140 Chr.2: 131055261-131055261 on Build GRCh38
rs906974796 Chr.2: 131055329-131055329 on Build GRCh38
rs780025588 Chr.2: 131055491-131055491 on Build GRCh38
rs746422907 Chr.2: 131055614-131055614 on Build GRCh38
rs757935212 Chr.2: 131055562-131055562 on Build GRCh38
rs547326076 Chr.2: 131055507-131055507 on Build GRCh38
rs762063773 Chr.2: 131055442-131055442 on Build GRCh38
rs777001725 Chr.2: 131055441-131055441 on Build GRCh38
rs758901455 Chr.2: 131055352-131055352 on Build GRCh38
rs770923592 Chr.2: 131055294-131055294 on Build GRCh38
rs780402785 Chr.2: 131055256-131055256 on Build GRCh38
rs989120559 Chr.2: 131055649-131055649 on Build GRCh38
rs748767288 Chr.2: 131055281-131055281 on Build GRCh38
rs934120590 Chr.2: 131055486-131055486 on Build GRCh38
rs764101499 Chr.2: 131055242-131055242 on Build GRCh38
rs367659089 Chr.2: 131055321-131055321 on Build GRCh38
rs373755562 Chr.2: 131055357-131055357 on Build GRCh38
rs774246574 Chr.2: 131055611-131055611 on Build GRCh38
rs757876838 Chr.2: 131055252-131055252 on Build GRCh38
rs368726362 Chr.2: 131055387-131055387 on Build GRCh38
rs756993189 Chr.2: 131055510-131055510 on Build GRCh38
rs533360958 Chr.2: 131055218-131055219 on Build GRCh38
rs368387575 Chr.2: 131055487-131055487 on Build GRCh38
rs749412899 Chr.2: 131055604-131055604 on Build GRCh38
rs201871520 Chr.2: 131055405-131055405 on Build GRCh38
rs369972882 Chr.2: 131055497-131055497 on Build GRCh38
rs760910629 Chr.2: 131055241-131055241 on Build GRCh38
rs376172440 Chr.2: 131055587-131055587 on Build GRCh38
rs200775848 Chr.2: 131055254-131055254 on Build GRCh38
rs532255564 Chr.2: 131055502-131055502 on Build GRCh38
rs1002604156 Chr.2: 131055629-131055629 on Build GRCh38
rs371738478 Chr.2: 131055426-131055426 on Build GRCh38
rs747790249 Chr.2: 131055258-131055258 on Build GRCh38
rs764862740 Chr.2: 131055546-131055546 on Build GRCh38
rs775186050 Chr.2: 131055414-131055414 on Build GRCh38
rs574781509 Chr.2: 131055358-131055358 on Build GRCh38
rs376238800 Chr.2: 131055229-131055229 on Build GRCh38
rs778354039 Chr.2: 131055395-131055395 on Build GRCh38
rs934873886 Chr.2: 131055232-131055232 on Build GRCh38
rs374948559 Chr.2: 131055535-131055535 on Build GRCh38
rs377290351 Chr.2: 131055490-131055490 on Build GRCh38
rs982993033 Chr.2: 131055354-131055354 on Build GRCh38
rs372649519 Chr.2: 131055489-131055489 on Build GRCh38
rs995389444 Chr.2: 131055265-131055265 on Build GRCh38
rs781185018 Chr.2: 131055581-131055581 on Build GRCh38
rs764304514 Chr.2: 131055645-131055645 on Build GRCh38
rs761491819 Chr.2: 131055538-131055538 on Build GRCh38
rs773297316 Chr.2: 131055450-131055450 on Build GRCh38
rs373179431 Chr.2: 131055634-131055634 on Build GRCh38
rs145809016 Chr.2: 131055219-131055219 on Build GRCh38
rs199942804 Chr.2: 131055335-131055335 on Build GRCh38
rs1026817768 Chr.2: 131055300-131055300 on Build GRCh38
rs369333964 Chr.2: 131055537-131055537 on Build GRCh38
rs755763969 Chr.2: 131055372-131055372 on Build GRCh38
rs748406544 Chr.2: 131055503-131055503 on Build GRCh38
rs563639423 Chr.2: 131055458-131055458 on Build GRCh38
rs745589508 Chr.2: 131055622-131055624 on Build GRCh38
rs756518344 Chr.2: 131055598-131055598 on Build GRCh38
rs753659808 Chr.2: 131055245-131055246 on Build GRCh38
rs371711272 Chr.2: 131055529-131055529 on Build GRCh38
rs370288982 Chr.2: 131055356-131055356 on Build GRCh38
rs948371683 Chr.2: 131055588-131055588 on Build GRCh38
rs767891364 Chr.2: 131055536-131055536 on Build GRCh38
rs775304769 Chr.2: 131055331-131055331 on Build GRCh38
rs771726249 Chr.2: 131055628-131055628 on Build GRCh38
rs748855193 Chr.2: 131055378-131055378 on Build GRCh38
rs773206134 Chr.2: 131055506-131055506 on Build GRCh38
rs145577811 Chr.2: 131055628-131055628 on Build GRCh38
rs770121760 Chr.2: 131055504-131055504 on Build GRCh38
rs750973421 Chr.2: 131055351-131055351 on Build GRCh38
rs893319104 Chr.2: 131055205-131055205 on Build GRCh38
rs767284958 Chr.2: 131055236-131055236 on Build GRCh38
rs1012616847 Chr.2: 131055313-131055313 on Build GRCh38
rs1016143321 Chr.2: 131055427-131055427 on Build GRCh38
rs946167919 Chr.2: 131055216-131055216 on Build GRCh38
rs753944110 Chr.2: 131055243-131055243 on Build GRCh38
rs759173663 Chr.2: 131055295-131055295 on Build GRCh38
rs754922146 Chr.2: 131055568-131055568 on Build GRCh38
rs374592834 Chr.2: 131055299-131055299 on Build GRCh38
rs770028990 Chr.2: 131055445-131055445 on Build GRCh38
rs762469713 Chr.2: 131055665-131055665 on Build GRCh38
rs148566888 Chr.2: 131055253-131055253 on Build GRCh38
rs778669080 Chr.2: 131055518-131055518 on Build GRCh38
rs771400429 Chr.2: 131055518-131055518 on Build GRCh38
rs774173481 Chr.2: 131055234-131055234 on Build GRCh38
rs771047687 Chr.2: 131055606-131055606 on Build GRCh38
rs576151507 Chr.2: 131055467-131055467 on Build GRCh38
rs751937465 Chr.2: 131055463-131055463 on Build GRCh38
rs375144049 Chr.2: 131055475-131055475 on Build GRCh38
rs772918185 Chr.2: 131055655-131055655 on Build GRCh38
rs770209806 Chr.2: 131055600-131055600 on Build GRCh38
rs771793214 Chr.2: 131055322-131055322 on Build GRCh38
rs777399561 Chr.2: 131055375-131055375 on Build GRCh38
rs368846974 Chr.2: 131055601-131055601 on Build GRCh38
rs774259471 Chr.2: 131055320-131055320 on Build GRCh38
rs776913934 Chr.2: 131055336-131055336 on Build GRCh38
rs769054416 Chr.2: 131055431-131055431 on Build GRCh38
rs749957361 Chr.2: 131055560-131055560 on Build GRCh38
rs763421403 Chr.2: 131055452-131055452 on Build GRCh38
rs200659520 Chr.2: 131055563-131055563 on Build GRCh38
rs762902146 Chr.2: 131055228-131055228 on Build GRCh38
rs772022280 Chr.2: 131055411-131055411 on Build GRCh38
rs775578999 Chr.2: 131055240-131055240 on Build GRCh38
rs1026024311 Chr.2: 131055667-131055667 on Build GRCh38
rs777315371 Chr.2: 131055263-131055263 on Build GRCh38
rs990405601 Chr.2: 131055413-131055413 on Build GRCh38
rs746860693 Chr.2: 131055495-131055495 on Build GRCh38
rs553389316 Chr.2: 131055339-131055339 on Build GRCh38

View Full Product Details