Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202942522-202943068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARF
Entrez Gene ID
79800
Gene Name
calcium responsive transcription factor
Gene Aliases
ALS2CR8, NYD-SP24
UniGene
Hs.444982
Gene Chromosome Location
Chr.2: 202912218-202987856 on Build GRCh38
Transcripts
NM_001104586.2 NM_001282910.1 NM_001282911.1 NM_001282912.1 NM_001322427.1 NM_001322428.1 NM_001322429.1 NM_001322430.1 NM_024744.15 NR_136330.1 NR_136331.1 XM_005246858.3 XM_005246859.3 XM_006712760.2 XM_011511867.2 XM_011511868.2 XM_017004961.1 XM_017004962.1 XM_017004963.1 XM_017004964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584295 c.306+3G>A p.? Chr.2: 202942970-202942970 on Build GRCh38
3576450 c.234C>T p.F78F Chr.2: 202942895-202942895 on Build GRCh38
80859 c.168T>C p.N56N Chr.2: 202942829-202942829 on Build GRCh38
1195422 c.155G>A p.R52H Chr.2: 202942816-202942816 on Build GRCh38
3768917 c.226G>A p.E76K Chr.2: 202942887-202942887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776904768 Chr.2: 202942719-202942719 on Build GRCh38
rs745904615 Chr.2: 202942815-202942815 on Build GRCh38
rs781381178 Chr.2: 202942810-202942810 on Build GRCh38
rs967847355 Chr.2: 202942957-202942957 on Build GRCh38
rs368347355 Chr.2: 202942903-202942903 on Build GRCh38
rs776571239 Chr.2: 202942699-202942699 on Build GRCh38
rs762046856 Chr.2: 202942921-202942921 on Build GRCh38
rs370013702 Chr.2: 202943026-202943026 on Build GRCh38
rs752564252 Chr.2: 202942875-202942875 on Build GRCh38
rs759090790 Chr.2: 202942963-202942963 on Build GRCh38
rs765172621 Chr.2: 202942869-202942869 on Build GRCh38
rs776088204 Chr.2: 202942818-202942818 on Build GRCh38
rs760145219 Chr.2: 202942701-202942701 on Build GRCh38
rs754986358 Chr.2: 202942764-202942764 on Build GRCh38
rs778785750 Chr.2: 202942779-202942779 on Build GRCh38
rs979494181 Chr.2: 202942975-202942975 on Build GRCh38
rs367676155 Chr.2: 202942983-202942983 on Build GRCh38
rs553785258 Chr.2: 202943025-202943025 on Build GRCh38
rs766561509 Chr.2: 202942680-202942680 on Build GRCh38
rs375402022 Chr.2: 202942968-202942968 on Build GRCh38
rs934039740 Chr.2: 202942642-202942642 on Build GRCh38
rs768844305 Chr.2: 202942718-202942718 on Build GRCh38
rs758132089 Chr.2: 202942789-202942789 on Build GRCh38
rs551749480 Chr.2: 202942729-202942729 on Build GRCh38
rs149069557 Chr.2: 202942710-202942710 on Build GRCh38
rs776037125 Chr.2: 202942719-202942719 on Build GRCh38
rs375856342 Chr.2: 202942634-202942634 on Build GRCh38
rs753423814 Chr.2: 202942866-202942866 on Build GRCh38
rs34319048 Chr.2: 202942831-202942832 on Build GRCh38
rs371549950 Chr.2: 202942816-202942816 on Build GRCh38
rs549597577 Chr.2: 202942910-202942910 on Build GRCh38
rs750035696 Chr.2: 202942748-202942748 on Build GRCh38
rs975639774 Chr.2: 202942999-202942999 on Build GRCh38
rs755582319 Chr.2: 202942751-202942751 on Build GRCh38
rs888924529 Chr.2: 202942752-202942752 on Build GRCh38
rs534752945 Chr.2: 202942691-202942692 on Build GRCh38
rs760427256 Chr.2: 202942856-202942856 on Build GRCh38
rs749874262 Chr.2: 202942915-202942915 on Build GRCh38
rs1054382148 Chr.2: 202942740-202942740 on Build GRCh38
rs964643353 Chr.2: 202942984-202942984 on Build GRCh38
rs1026503490 Chr.2: 202942569-202942569 on Build GRCh38
rs769266943 Chr.2: 202942916-202942916 on Build GRCh38
rs952436203 Chr.2: 202942571-202942571 on Build GRCh38
rs201669246 Chr.2: 202942835-202942835 on Build GRCh38
rs755689850 Chr.2: 202943003-202943003 on Build GRCh38
rs777977612 Chr.2: 202942793-202942793 on Build GRCh38
rs767296805 Chr.2: 202942732-202942732 on Build GRCh38
rs747959867 Chr.2: 202942786-202942786 on Build GRCh38
rs76057377 Chr.2: 202942892-202942892 on Build GRCh38
rs771043074 Chr.2: 202942809-202942809 on Build GRCh38
rs992351690 Chr.2: 202942990-202942990 on Build GRCh38
rs773705443 Chr.2: 202942869-202942870 on Build GRCh38
rs774821788 Chr.2: 202942919-202942919 on Build GRCh38
rs190500049 Chr.2: 202942576-202942576 on Build GRCh38
rs566163221 Chr.2: 202943002-202943002 on Build GRCh38
rs758328312 Chr.2: 202942889-202942889 on Build GRCh38
rs917015368 Chr.2: 202942702-202942702 on Build GRCh38
rs776089990 Chr.2: 202942961-202942961 on Build GRCh38
rs533813274 Chr.2: 202943022-202943022 on Build GRCh38
rs758925157 Chr.2: 202942867-202942867 on Build GRCh38
rs769072780 Chr.2: 202942838-202942838 on Build GRCh38
rs763985667 Chr.2: 202943007-202943007 on Build GRCh38
rs745982342 Chr.2: 202942905-202942905 on Build GRCh38
rs756104051 Chr.2: 202943016-202943016 on Build GRCh38
rs765973936 Chr.2: 202942757-202942757 on Build GRCh38
rs527998316 Chr.2: 202943001-202943001 on Build GRCh38
rs762924613 Chr.2: 202943005-202943005 on Build GRCh38
rs753268367 Chr.2: 202942759-202942759 on Build GRCh38
rs567927407 Chr.2: 202942913-202942913 on Build GRCh38
rs756953826 Chr.2: 202943017-202943017 on Build GRCh38
rs770069952 Chr.2: 202942729-202942729 on Build GRCh38
rs765919935 Chr.2: 202942858-202942858 on Build GRCh38
rs993485355 Chr.2: 202942561-202942561 on Build GRCh38
rs750669975 Chr.2: 202942677-202942677 on Build GRCh38
rs377435192 Chr.2: 202942853-202942853 on Build GRCh38
rs202179884 Chr.2: 202942805-202942805 on Build GRCh38
rs371999855 Chr.2: 202942944-202942944 on Build GRCh38
rs965991047 Chr.2: 202942557-202942557 on Build GRCh38
rs748533903 Chr.2: 202942718-202942719 on Build GRCh38
rs935883967 Chr.2: 202942733-202942733 on Build GRCh38

View Full Product Details