Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207570061-207570592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_011510650.2 XM_011510651.2 XM_017003399.1 XM_017003400.1 XM_017003401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292769 c.547+3G>T p.? Chr.2: 207570324-207570324 on Build GRCh38
4829726 c.454G>C p.E152Q Chr.2: 207570228-207570228 on Build GRCh38
5481684 c.506C>T p.T169M Chr.2: 207570280-207570280 on Build GRCh38
6224923 c.405-1G>T p.? Chr.2: 207570178-207570178 on Build GRCh38
7328181 c.547+1G>T p.? Chr.2: 207570322-207570322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008582342 Chr.2: 207570482-207570482 on Build GRCh38
rs1031176218 Chr.2: 207570459-207570459 on Build GRCh38
rs56039637 Chr.2: 207570536-207570536 on Build GRCh38
rs1018998604 Chr.2: 207570565-207570565 on Build GRCh38
rs552888247 Chr.2: 207570372-207570372 on Build GRCh38
rs887888124 Chr.2: 207570099-207570099 on Build GRCh38
rs868493052 Chr.2: 207570469-207570469 on Build GRCh38
rs766439438 Chr.2: 207570146-207570146 on Build GRCh38
rs201642429 Chr.2: 207570281-207570281 on Build GRCh38
rs201006565 Chr.2: 207570388-207570388 on Build GRCh38
rs779699990 Chr.2: 207570260-207570260 on Build GRCh38
rs980789598 Chr.2: 207570533-207570533 on Build GRCh38
rs896948175 Chr.2: 207570329-207570329 on Build GRCh38
rs767690757 Chr.2: 207570359-207570359 on Build GRCh38
rs765492288 Chr.2: 207570505-207570505 on Build GRCh38
rs762025997 Chr.2: 207570282-207570282 on Build GRCh38
rs772427535 Chr.2: 207570302-207570302 on Build GRCh38
rs188315570 Chr.2: 207570084-207570084 on Build GRCh38
rs757047432 Chr.2: 207570365-207570365 on Build GRCh38
rs536477824 Chr.2: 207570501-207570501 on Build GRCh38
rs753834321 Chr.2: 207570346-207570346 on Build GRCh38
rs745805546 Chr.2: 207570209-207570209 on Build GRCh38
rs992334207 Chr.2: 207570472-207570472 on Build GRCh38
rs1013387089 Chr.2: 207570092-207570092 on Build GRCh38
rs957051932 Chr.2: 207570467-207570467 on Build GRCh38
rs751274381 Chr.2: 207570172-207570172 on Build GRCh38
rs967788255 Chr.2: 207570419-207570419 on Build GRCh38
rs760678248 Chr.2: 207570145-207570145 on Build GRCh38
rs377258860 Chr.2: 207570497-207570497 on Build GRCh38
rs750939694 Chr.2: 207570138-207570138 on Build GRCh38
rs752980714 Chr.2: 207570363-207570363 on Build GRCh38
rs757145008 Chr.2: 207570180-207570180 on Build GRCh38
rs760297212 Chr.2: 207570559-207570559 on Build GRCh38
rs761154605 Chr.2: 207570334-207570334 on Build GRCh38
rs771811975 Chr.2: 207570147-207570147 on Build GRCh38
rs751568736 Chr.2: 207570132-207570133 on Build GRCh38
rs942074081 Chr.2: 207570253-207570253 on Build GRCh38
rs113872103 Chr.2: 207570417-207570417 on Build GRCh38
rs765332411 Chr.2: 207570354-207570354 on Build GRCh38
rs781221818 Chr.2: 207570194-207570194 on Build GRCh38
rs766917972 Chr.2: 207570343-207570343 on Build GRCh38
rs748868942 Chr.2: 207570270-207570270 on Build GRCh38
rs918195995 Chr.2: 207570493-207570493 on Build GRCh38
rs769918733 Chr.2: 207570258-207570258 on Build GRCh38
rs995126472 Chr.2: 207570330-207570330 on Build GRCh38
rs773628752 Chr.2: 207570333-207570333 on Build GRCh38
rs752889040 Chr.2: 207570373-207570373 on Build GRCh38
rs899292076 Chr.2: 207570264-207570264 on Build GRCh38
rs759695908 Chr.2: 207570350-207570350 on Build GRCh38
rs759621519 Chr.2: 207570351-207570364 on Build GRCh38
rs779239350 Chr.2: 207570169-207570169 on Build GRCh38
rs192210277 Chr.2: 207570389-207570389 on Build GRCh38
rs755130189 Chr.2: 207570160-207570160 on Build GRCh38
rs1017878574 Chr.2: 207570405-207570405 on Build GRCh38
rs950458883 Chr.2: 207570402-207570402 on Build GRCh38
rs540441782 Chr.2: 207570203-207570203 on Build GRCh38
rs772974217 Chr.2: 207570543-207570543 on Build GRCh38
rs768152463 Chr.2: 207570272-207570272 on Build GRCh38
rs979561678 Chr.2: 207570456-207570456 on Build GRCh38

View Full Product Details