Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215332980-215333502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATIC
Entrez Gene ID
471
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
UniGene
Hs.90280
Gene Chromosome Location
Chr.2: 215311956-215359745 on Build GRCh38
Transcripts
NM_004044.6 XM_017004187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016145 c.850G>T p.A284S Chr.2: 215333385-215333385 on Build GRCh38
5642402 c.876C>A p.L292L Chr.2: 215333411-215333411 on Build GRCh38
1016144 c.844G>T p.D282Y Chr.2: 215333379-215333379 on Build GRCh38
3953031 c.922G>A p.G308R Chr.2: 215333457-215333457 on Build GRCh38
4154792 c.920G>C p.R307T Chr.2: 215333455-215333455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000552584 Chr.2: 215333103-215333103 on Build GRCh38
rs766893403 Chr.2: 215333303-215333303 on Build GRCh38
rs113644379 Chr.2: 215333296-215333296 on Build GRCh38
rs758755562 Chr.2: 215333337-215333337 on Build GRCh38
rs374860324 Chr.2: 215333421-215333421 on Build GRCh38
rs986277621 Chr.2: 215333238-215333238 on Build GRCh38
rs778069788 Chr.2: 215333351-215333351 on Build GRCh38
rs970813283 Chr.2: 215333166-215333166 on Build GRCh38
rs12995526 Chr.2: 215333248-215333248 on Build GRCh38
rs773792940 Chr.2: 215333404-215333404 on Build GRCh38
rs924579949 Chr.2: 215333030-215333030 on Build GRCh38
rs3821353 Chr.2: 215333056-215333056 on Build GRCh38
rs371518493 Chr.2: 215333397-215333397 on Build GRCh38
rs1024111267 Chr.2: 215333162-215333162 on Build GRCh38
rs902833053 Chr.2: 215333102-215333102 on Build GRCh38
rs146593641 Chr.2: 215333087-215333087 on Build GRCh38
rs375460932 Chr.2: 215333316-215333316 on Build GRCh38
rs372300174 Chr.2: 215333438-215333438 on Build GRCh38
rs1015363238 Chr.2: 215333182-215333182 on Build GRCh38
rs73089311 Chr.2: 215333153-215333153 on Build GRCh38
rs769666403 Chr.2: 215333383-215333383 on Build GRCh38
rs751270833 Chr.2: 215333272-215333272 on Build GRCh38
rs990195919 Chr.2: 215333059-215333062 on Build GRCh38
rs190688403 Chr.2: 215333307-215333307 on Build GRCh38
rs1017060849 Chr.2: 215333155-215333155 on Build GRCh38
rs527898971 Chr.2: 215333025-215333025 on Build GRCh38
rs760972324 Chr.2: 215333302-215333302 on Build GRCh38
rs777028878 Chr.2: 215333431-215333431 on Build GRCh38
rs572284878 Chr.2: 215333104-215333104 on Build GRCh38
rs765123991 Chr.2: 215333190-215333190 on Build GRCh38
rs188057519 Chr.2: 215333221-215333221 on Build GRCh38
rs771716973 Chr.2: 215333410-215333410 on Build GRCh38
rs10179873 Chr.2: 215333326-215333326 on Build GRCh38
rs868458651 Chr.2: 215333211-215333211 on Build GRCh38
rs181312746 Chr.2: 215333239-215333239 on Build GRCh38
rs1038021651 Chr.2: 215333428-215333428 on Build GRCh38
rs778345919 Chr.2: 215333471-215333471 on Build GRCh38
rs916022898 Chr.2: 215333255-215333255 on Build GRCh38
rs143359980 Chr.2: 215333458-215333458 on Build GRCh38
rs929545425 Chr.2: 215333101-215333101 on Build GRCh38
rs779764592 Chr.2: 215333460-215333460 on Build GRCh38
rs77534283 Chr.2: 215333256-215333256 on Build GRCh38
rs996579932 Chr.2: 215333434-215333434 on Build GRCh38
rs760015676 Chr.2: 215333435-215333435 on Build GRCh38
rs142401012 Chr.2: 215333441-215333441 on Build GRCh38
rs747653411 Chr.2: 215333406-215333406 on Build GRCh38
rs775474198 Chr.2: 215333206-215333206 on Build GRCh38
rs546639237 Chr.2: 215333027-215333027 on Build GRCh38
rs141406980 Chr.2: 215333243-215333244 on Build GRCh38
rs765766705 Chr.2: 215333437-215333437 on Build GRCh38
rs766883760 Chr.2: 215333342-215333342 on Build GRCh38
rs547703964 Chr.2: 215333301-215333301 on Build GRCh38
rs982194573 Chr.2: 215333179-215333179 on Build GRCh38
rs894352753 Chr.2: 215333120-215333120 on Build GRCh38
rs34416522 Chr.2: 215333006-215333007 on Build GRCh38
rs962055843 Chr.2: 215333185-215333185 on Build GRCh38
rs566284744 Chr.2: 215333253-215333253 on Build GRCh38
rs779345676 Chr.2: 215333467-215333468 on Build GRCh38
rs3821354 Chr.2: 215333074-215333074 on Build GRCh38
rs780074840 Chr.2: 215333386-215333386 on Build GRCh38
rs767144865 Chr.2: 215333300-215333300 on Build GRCh38
rs371182050 Chr.2: 215333299-215333299 on Build GRCh38
rs899608772 Chr.2: 215333433-215333433 on Build GRCh38
rs766691786 Chr.2: 215333429-215333429 on Build GRCh38
rs948937685 Chr.2: 215333068-215333068 on Build GRCh38
rs1030032312 Chr.2: 215333024-215333025 on Build GRCh38
rs113864137 Chr.2: 215333061-215333061 on Build GRCh38
rs886751206 Chr.2: 215333148-215333148 on Build GRCh38
rs772803024 Chr.2: 215333411-215333411 on Build GRCh38
rs902881606 Chr.2: 215333261-215333261 on Build GRCh38
rs755476983 Chr.2: 215333315-215333315 on Build GRCh38
rs751794606 Chr.2: 215333448-215333448 on Build GRCh38
rs757560807 Chr.2: 215333395-215333399 on Build GRCh38
rs1050171167 Chr.2: 215333036-215333036 on Build GRCh38
rs558723404 Chr.2: 215333044-215333044 on Build GRCh38
rs749225851 Chr.2: 215333395-215333395 on Build GRCh38
rs778906526 Chr.2: 215333317-215333317 on Build GRCh38
rs745674983 Chr.2: 215333353-215333353 on Build GRCh38
rs569770353 Chr.2: 215333023-215333023 on Build GRCh38

View Full Product Details