Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230012368-230012882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO36
Entrez Gene ID
130888
Gene Name
F-box protein 36
Gene Aliases
Fbx36
UniGene
Hs.140666
Gene Chromosome Location
Chr.2: 229922491-230013109 on Build GRCh38
Transcripts
NM_174899.4 XM_005246317.1

SNP Detail

SNP ID Chromosome Location
rs912272563 Chr.2: 230012742-230012742 on Build GRCh38
rs924896313 Chr.2: 230012845-230012845 on Build GRCh38
rs139646192 Chr.2: 230012715-230012715 on Build GRCh38
rs755139702 Chr.2: 230012703-230012703 on Build GRCh38
rs982636167 Chr.2: 230012738-230012738 on Build GRCh38
rs183967282 Chr.2: 230012634-230012634 on Build GRCh38
rs1026946085 Chr.2: 230012721-230012721 on Build GRCh38
rs113935612 Chr.2: 230012580-230012580 on Build GRCh38
rs890791755 Chr.2: 230012694-230012694 on Build GRCh38
rs549458959 Chr.2: 230012517-230012517 on Build GRCh38
rs936172498 Chr.2: 230012590-230012590 on Build GRCh38
rs1004569850 Chr.2: 230012702-230012702 on Build GRCh38
rs751539931 Chr.2: 230012542-230012542 on Build GRCh38
rs1022986258 Chr.2: 230012620-230012620 on Build GRCh38
rs943801282 Chr.2: 230012754-230012754 on Build GRCh38
rs947092364 Chr.2: 230012793-230012793 on Build GRCh38
rs531688331 Chr.2: 230012431-230012431 on Build GRCh38
rs951283860 Chr.2: 230012734-230012734 on Build GRCh38
rs958790604 Chr.2: 230012795-230012795 on Build GRCh38
rs569311455 Chr.2: 230012705-230012705 on Build GRCh38
rs1043312640 Chr.2: 230012430-230012430 on Build GRCh38
rs1036149911 Chr.2: 230012576-230012576 on Build GRCh38
rs557565686 Chr.2: 230012651-230012651 on Build GRCh38
rs879839856 Chr.2: 230012623-230012623 on Build GRCh38
rs142638322 Chr.2: 230012482-230012482 on Build GRCh38
rs567878855 Chr.2: 230012818-230012818 on Build GRCh38
rs920910226 Chr.2: 230012783-230012783 on Build GRCh38
rs371970317 Chr.2: 230012491-230012491 on Build GRCh38
rs111895699 Chr.2: 230012597-230012597 on Build GRCh38
rs566127273 Chr.2: 230012605-230012605 on Build GRCh38
rs991461487 Chr.2: 230012472-230012472 on Build GRCh38
rs1043281381 Chr.2: 230012807-230012807 on Build GRCh38
rs958758520 Chr.2: 230012452-230012452 on Build GRCh38
rs554960630 Chr.2: 230012856-230012856 on Build GRCh38
rs932362361 Chr.2: 230012619-230012619 on Build GRCh38
rs924808334 Chr.2: 230012584-230012584 on Build GRCh38
rs375304058 Chr.2: 230012661-230012661 on Build GRCh38
rs568736296 Chr.2: 230012555-230012555 on Build GRCh38
rs754668238 Chr.2: 230012768-230012768 on Build GRCh38
rs962565429 Chr.2: 230012408-230012408 on Build GRCh38
rs963401714 Chr.2: 230012675-230012675 on Build GRCh38

View Full Product Details