Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232766776-232767276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
UniGene
Hs.467338
Gene Chromosome Location
Chr.2: 232765802-232776565 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

SNP Detail

SNP ID Chromosome Location
rs762142938 Chr.2: 232766876-232766876 on Build GRCh38
rs376571065 Chr.2: 232767208-232767208 on Build GRCh38
rs972195729 Chr.2: 232767023-232767023 on Build GRCh38
rs1016045931 Chr.2: 232766996-232766996 on Build GRCh38
rs1040122587 Chr.2: 232766979-232766979 on Build GRCh38
rs560074051 Chr.2: 232766827-232766827 on Build GRCh38
rs116740510 Chr.2: 232766990-232766990 on Build GRCh38
rs527706386 Chr.2: 232767206-232767206 on Build GRCh38
rs768069588 Chr.2: 232767103-232767103 on Build GRCh38
rs1013792535 Chr.2: 232767117-232767117 on Build GRCh38
rs920191282 Chr.2: 232766831-232766831 on Build GRCh38
rs969558495 Chr.2: 232767141-232767141 on Build GRCh38
rs923138366 Chr.2: 232767051-232767055 on Build GRCh38
rs141561309 Chr.2: 232767069-232767069 on Build GRCh38
rs933112110 Chr.2: 232767051-232767051 on Build GRCh38
rs201810122 Chr.2: 232766894-232766894 on Build GRCh38
rs986106356 Chr.2: 232767062-232767062 on Build GRCh38
rs908580305 Chr.2: 232767226-232767226 on Build GRCh38
rs952684250 Chr.2: 232767215-232767215 on Build GRCh38
rs750978844 Chr.2: 232767165-232767165 on Build GRCh38
rs903500073 Chr.2: 232767028-232767028 on Build GRCh38
rs980971006 Chr.2: 232767205-232767205 on Build GRCh38
rs901710627 Chr.2: 232766983-232766983 on Build GRCh38
rs886055793 Chr.2: 232767234-232767234 on Build GRCh38
rs191034075 Chr.2: 232766843-232766843 on Build GRCh38
rs931542186 Chr.2: 232766859-232766859 on Build GRCh38
rs887271781 Chr.2: 232766963-232766963 on Build GRCh38
rs1025133291 Chr.2: 232767126-232767126 on Build GRCh38
rs1000548585 Chr.2: 232767032-232767032 on Build GRCh38
rs1024267411 Chr.2: 232766974-232766974 on Build GRCh38
rs1031135636 Chr.2: 232767078-232767078 on Build GRCh38
rs532524928 Chr.2: 232767187-232767187 on Build GRCh38
rs200075792 Chr.2: 232766976-232766976 on Build GRCh38
rs559498619 Chr.2: 232766909-232766909 on Build GRCh38
rs999670544 Chr.2: 232766809-232766809 on Build GRCh38
rs561799289 Chr.2: 232767080-232767080 on Build GRCh38
rs896615648 Chr.2: 232766866-232766866 on Build GRCh38
rs182131998 Chr.2: 232767005-232767005 on Build GRCh38
rs377117377 Chr.2: 232766826-232766826 on Build GRCh38
rs202203672 Chr.2: 232766901-232766901 on Build GRCh38
rs944132200 Chr.2: 232767234-232767234 on Build GRCh38
rs982752400 Chr.2: 232767221-232767221 on Build GRCh38
rs1047245172 Chr.2: 232766949-232766949 on Build GRCh38

View Full Product Details