Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6076355-6076855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
UniGene
Hs.472054
Gene Chromosome Location
Chr.20: 6074845-6123544 on Build GRCh38
Transcripts
NM_017671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161046 c.1650A>T p.R550S Chr.20: 6076456-6076456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576322873 Chr.20: 6076734-6076734 on Build GRCh38
rs781160740 Chr.20: 6076483-6076483 on Build GRCh38
rs542709881 Chr.20: 6076448-6076448 on Build GRCh38
rs969989950 Chr.20: 6076659-6076659 on Build GRCh38
rs770846215 Chr.20: 6076479-6076479 on Build GRCh38
rs565407541 Chr.20: 6076606-6076606 on Build GRCh38
rs545057383 Chr.20: 6076810-6076810 on Build GRCh38
rs1037009708 Chr.20: 6076540-6076540 on Build GRCh38
rs528619608 Chr.20: 6076667-6076667 on Build GRCh38
rs775060736 Chr.20: 6076405-6076406 on Build GRCh38
rs370906888 Chr.20: 6076443-6076443 on Build GRCh38
rs6117067 Chr.20: 6076647-6076647 on Build GRCh38
rs188548418 Chr.20: 6076390-6076390 on Build GRCh38
rs550396481 Chr.20: 6076389-6076389 on Build GRCh38
rs978445167 Chr.20: 6076751-6076751 on Build GRCh38
rs898106780 Chr.20: 6076618-6076618 on Build GRCh38
rs750904222 Chr.20: 6076395-6076395 on Build GRCh38
rs1009430299 Chr.20: 6076723-6076723 on Build GRCh38
rs959892100 Chr.20: 6076672-6076672 on Build GRCh38
rs748927308 Chr.20: 6076418-6076418 on Build GRCh38
rs761138761 Chr.20: 6076819-6076819 on Build GRCh38
rs942947229 Chr.20: 6076784-6076784 on Build GRCh38
rs536245011 Chr.20: 6076623-6076623 on Build GRCh38
rs576410071 Chr.20: 6076735-6076735 on Build GRCh38
rs1035509380 Chr.20: 6076668-6076668 on Build GRCh38
rs768192573 Chr.20: 6076640-6076640 on Build GRCh38
rs766683404 Chr.20: 6076822-6076822 on Build GRCh38
rs768309440 Chr.20: 6076474-6076474 on Build GRCh38
rs910918601 Chr.20: 6076781-6076781 on Build GRCh38
rs374876522 Chr.20: 6076420-6076420 on Build GRCh38
rs901439562 Chr.20: 6076830-6076830 on Build GRCh38
rs192292204 Chr.20: 6076476-6076476 on Build GRCh38
rs536947318 Chr.20: 6076740-6076740 on Build GRCh38
rs920894502 Chr.20: 6076753-6076753 on Build GRCh38
rs745634549 Chr.20: 6076510-6076510 on Build GRCh38
rs371657740 Chr.20: 6076430-6076430 on Build GRCh38
rs183993931 Chr.20: 6076535-6076535 on Build GRCh38
rs755419437 Chr.20: 6076441-6076441 on Build GRCh38
rs767059789 Chr.20: 6076424-6076424 on Build GRCh38
rs1032444979 Chr.20: 6076681-6076681 on Build GRCh38
rs747864690 Chr.20: 6076458-6076458 on Build GRCh38
rs994241301 Chr.20: 6076505-6076505 on Build GRCh38
rs958081652 Chr.20: 6076704-6076704 on Build GRCh38
rs756695878 Chr.20: 6076423-6076423 on Build GRCh38
rs865955953 Chr.20: 6076739-6076739 on Build GRCh38
rs373866950 Chr.20: 6076537-6076537 on Build GRCh38
rs976405045 Chr.20: 6076750-6076750 on Build GRCh38
rs1013538347 Chr.20: 6076638-6076638 on Build GRCh38
rs112442363 Chr.20: 6076651-6076651 on Build GRCh38
rs6107754 Chr.20: 6076746-6076746 on Build GRCh38
rs1048148033 Chr.20: 6076523-6076524 on Build GRCh38
rs770481049 Chr.20: 6076542-6076543 on Build GRCh38
rs775250903 Chr.20: 6076408-6076408 on Build GRCh38
rs757795824 Chr.20: 6076467-6076467 on Build GRCh38
rs762424632 Chr.20: 6076411-6076412 on Build GRCh38
rs367688066 Chr.20: 6076534-6076534 on Build GRCh38
rs953713964 Chr.20: 6076829-6076829 on Build GRCh38
rs888548988 Chr.20: 6076539-6076539 on Build GRCh38
rs1043120859 Chr.20: 6076401-6076401 on Build GRCh38

View Full Product Details