Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6088985-6089532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
UniGene
Hs.472054
Gene Chromosome Location
Chr.20: 6074845-6123544 on Build GRCh38
Transcripts
NM_017671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478364 c.1181T>G p.F394C Chr.20: 6089048-6089048 on Build GRCh38
1713154 c.1199C>T p.S400F Chr.20: 6089030-6089030 on Build GRCh38
3548577 c.1185C>T p.I395I Chr.20: 6089044-6089044 on Build GRCh38
3673152 c.1222G>T p.E408* Chr.20: 6089007-6089007 on Build GRCh38
4684341 c.1205C>T p.A402V Chr.20: 6089024-6089024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575485797 Chr.20: 6089391-6089391 on Build GRCh38
rs962995565 Chr.20: 6089338-6089338 on Build GRCh38
rs927239737 Chr.20: 6089065-6089065 on Build GRCh38
rs970780376 Chr.20: 6089487-6089487 on Build GRCh38
rs564199459 Chr.20: 6089507-6089507 on Build GRCh38
rs189183428 Chr.20: 6089237-6089237 on Build GRCh38
rs553114149 Chr.20: 6089263-6089263 on Build GRCh38
rs926463344 Chr.20: 6089156-6089156 on Build GRCh38
rs963699132 Chr.20: 6089418-6089418 on Build GRCh38
rs937899032 Chr.20: 6089187-6089188 on Build GRCh38
rs4815888 Chr.20: 6089405-6089405 on Build GRCh38
rs768469368 Chr.20: 6089242-6089242 on Build GRCh38
rs181819823 Chr.20: 6089324-6089324 on Build GRCh38
rs958635347 Chr.20: 6089388-6089388 on Build GRCh38
rs780581206 Chr.20: 6089101-6089101 on Build GRCh38
rs536285387 Chr.20: 6089081-6089081 on Build GRCh38
rs558951165 Chr.20: 6089200-6089200 on Build GRCh38
rs138706973 Chr.20: 6089137-6089137 on Build GRCh38
rs773939268 Chr.20: 6089113-6089113 on Build GRCh38
rs780468359 Chr.20: 6089311-6089311 on Build GRCh38
rs925734742 Chr.20: 6089491-6089491 on Build GRCh38
rs914874488 Chr.20: 6089287-6089287 on Build GRCh38
rs757299220 Chr.20: 6089085-6089085 on Build GRCh38
rs746023395 Chr.20: 6089088-6089088 on Build GRCh38
rs753662479 Chr.20: 6089068-6089068 on Build GRCh38
rs770269738 Chr.20: 6089094-6089094 on Build GRCh38
rs1051230996 Chr.20: 6089301-6089301 on Build GRCh38
rs781132211 Chr.20: 6089087-6089087 on Build GRCh38
rs750308235 Chr.20: 6089051-6089051 on Build GRCh38
rs3827699 Chr.20: 6089458-6089458 on Build GRCh38
rs749553968 Chr.20: 6089362-6089362 on Build GRCh38
rs116175000 Chr.20: 6089129-6089129 on Build GRCh38
rs917056577 Chr.20: 6089416-6089416 on Build GRCh38
rs746392391 Chr.20: 6089167-6089167 on Build GRCh38
rs35413391 Chr.20: 6089076-6089076 on Build GRCh38
rs869312718 Chr.20: 6089031-6089031 on Build GRCh38
rs113748453 Chr.20: 6089143-6089143 on Build GRCh38
rs756568918 Chr.20: 6089290-6089290 on Build GRCh38
rs374069028 Chr.20: 6089062-6089062 on Build GRCh38
rs778100302 Chr.20: 6089080-6089080 on Build GRCh38
rs373454595 Chr.20: 6089111-6089111 on Build GRCh38
rs376957545 Chr.20: 6089115-6089115 on Build GRCh38
rs869312729 Chr.20: 6089068-6089068 on Build GRCh38
rs749615948 Chr.20: 6089105-6089105 on Build GRCh38
rs766950162 Chr.20: 6089046-6089046 on Build GRCh38
rs998308598 Chr.20: 6089296-6089296 on Build GRCh38
rs775333932 Chr.20: 6089126-6089126 on Build GRCh38
rs869312730 Chr.20: 6089053-6089053 on Build GRCh38
rs963433241 Chr.20: 6089128-6089128 on Build GRCh38
rs1041499074 Chr.20: 6089144-6089144 on Build GRCh38
rs6038352 Chr.20: 6089323-6089323 on Build GRCh38
rs1002242727 Chr.20: 6089342-6089342 on Build GRCh38
rs761341202 Chr.20: 6089020-6089020 on Build GRCh38
rs977128144 Chr.20: 6089468-6089468 on Build GRCh38
rs145606109 Chr.20: 6089234-6089234 on Build GRCh38
rs1034676911 Chr.20: 6089369-6089369 on Build GRCh38
rs1026319354 Chr.20: 6089480-6089480 on Build GRCh38
rs1017038958 Chr.20: 6089332-6089332 on Build GRCh38
rs935658886 Chr.20: 6089495-6089495 on Build GRCh38
rs547871353 Chr.20: 6089389-6089389 on Build GRCh38
rs889318198 Chr.20: 6089392-6089392 on Build GRCh38

View Full Product Details