Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32447276-32447813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL4L
Entrez Gene ID
140688
Gene Name
nucleolar protein 4 like
Gene Aliases
C20orf112, C20orf113, dJ1184F4.2, dJ1184F4.4
UniGene
Hs.516978
Gene Chromosome Location
Chr.20: 32443059-32585073 on Build GRCh38
Transcripts
NM_001256798.1 NM_080616.4 XM_005260282.4 XM_005260283.4 XM_005260284.4 XM_005260285.4 XM_005260286.4 XM_005260288.2 XM_005260289.4 XM_006723702.3 XM_006723703.3 XM_011528562.2 XM_011528563.1 XM_017027669.1 XM_017027670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025748 c.1168A>G p.T390A Chr.20: 32447739-32447739 on Build GRCh38
6252279 c.1116C>A p.G372G Chr.20: 32447791-32447791 on Build GRCh38
6615503 c.1106G>A p.S369N Chr.20: 32447801-32447801 on Build GRCh38
4164669 c.1120G>A p.A374T Chr.20: 32447787-32447787 on Build GRCh38
478007 c.1108A>G p.M370V Chr.20: 32447799-32447799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761283119 Chr.20: 32447777-32447777 on Build GRCh38
rs867919106 Chr.20: 32447587-32447587 on Build GRCh38
rs533426847 Chr.20: 32447541-32447541 on Build GRCh38
rs758494565 Chr.20: 32447689-32447689 on Build GRCh38
rs201552093 Chr.20: 32447611-32447611 on Build GRCh38
rs750955868 Chr.20: 32447339-32447339 on Build GRCh38
rs745812555 Chr.20: 32447762-32447762 on Build GRCh38
rs749152465 Chr.20: 32447768-32447768 on Build GRCh38
rs764917691 Chr.20: 32447610-32447610 on Build GRCh38
rs779721841 Chr.20: 32447767-32447767 on Build GRCh38
rs768920742 Chr.20: 32447566-32447566 on Build GRCh38
rs747061655 Chr.20: 32447756-32447756 on Build GRCh38
rs1054629969 Chr.20: 32447663-32447663 on Build GRCh38
rs143292226 Chr.20: 32447692-32447692 on Build GRCh38
rs545820363 Chr.20: 32447556-32447556 on Build GRCh38
rs775877396 Chr.20: 32447552-32447553 on Build GRCh38
rs935723095 Chr.20: 32447638-32447638 on Build GRCh38
rs907185695 Chr.20: 32447547-32447547 on Build GRCh38
rs962060950 Chr.20: 32447723-32447723 on Build GRCh38
rs891469019 Chr.20: 32447386-32447386 on Build GRCh38
rs1737887 Chr.20: 32447326-32447326 on Build GRCh38
rs777147266 Chr.20: 32447784-32447784 on Build GRCh38
rs996050637 Chr.20: 32447429-32447429 on Build GRCh38
rs779063979 Chr.20: 32447569-32447569 on Build GRCh38
rs779165706 Chr.20: 32447685-32447685 on Build GRCh38
rs375174652 Chr.20: 32447671-32447671 on Build GRCh38
rs766890241 Chr.20: 32447728-32447728 on Build GRCh38
rs773244243 Chr.20: 32447577-32447577 on Build GRCh38
rs901670410 Chr.20: 32447404-32447404 on Build GRCh38
rs778198218 Chr.20: 32447754-32447754 on Build GRCh38
rs758688029 Chr.20: 32447749-32447749 on Build GRCh38
rs774040982 Chr.20: 32447771-32447771 on Build GRCh38
rs762405985 Chr.20: 32447668-32447668 on Build GRCh38
rs962378798 Chr.20: 32447513-32447513 on Build GRCh38
rs145067613 Chr.20: 32447357-32447357 on Build GRCh38
rs770782194 Chr.20: 32447584-32447584 on Build GRCh38
rs368933653 Chr.20: 32447790-32447790 on Build GRCh38
rs887823645 Chr.20: 32447456-32447456 on Build GRCh38
rs769503846 Chr.20: 32447706-32447706 on Build GRCh38
rs557850410 Chr.20: 32447318-32447318 on Build GRCh38
rs746955555 Chr.20: 32447578-32447578 on Build GRCh38
rs748349050 Chr.20: 32447688-32447688 on Build GRCh38
rs925461940 Chr.20: 32447620-32447620 on Build GRCh38
rs763531949 Chr.20: 32447720-32447720 on Build GRCh38
rs772319246 Chr.20: 32447576-32447576 on Build GRCh38
rs386393630 Chr.20: 32447404-32447405 on Build GRCh38
rs544392323 Chr.20: 32447389-32447389 on Build GRCh38
rs773849504 Chr.20: 32447722-32447722 on Build GRCh38
rs766940645 Chr.20: 32447674-32447674 on Build GRCh38
rs755331445 Chr.20: 32447736-32447736 on Build GRCh38
rs992407879 Chr.20: 32447545-32447545 on Build GRCh38
rs776479346 Chr.20: 32447698-32447698 on Build GRCh38
rs76752983 Chr.20: 32447432-32447432 on Build GRCh38
rs1040290757 Chr.20: 32447677-32447677 on Build GRCh38
rs776479949 Chr.20: 32447588-32447588 on Build GRCh38
rs71338906 Chr.20: 32447783-32447783 on Build GRCh38
rs749406674 Chr.20: 32447558-32447558 on Build GRCh38
rs540393376 Chr.20: 32447350-32447350 on Build GRCh38
rs771514773 Chr.20: 32447779-32447779 on Build GRCh38
rs754219166 Chr.20: 32447676-32447676 on Build GRCh38
rs752236350 Chr.20: 32447323-32447323 on Build GRCh38
rs774601152 Chr.20: 32447733-32447744 on Build GRCh38
rs745641509 Chr.20: 32447702-32447702 on Build GRCh38
rs563943986 Chr.20: 32447573-32447573 on Build GRCh38
rs759230082 Chr.20: 32447317-32447317 on Build GRCh38
rs757891913 Chr.20: 32447325-32447325 on Build GRCh38
rs753096212 Chr.20: 32447791-32447791 on Build GRCh38
rs969576664 Chr.20: 32447753-32447753 on Build GRCh38
rs1014904798 Chr.20: 32447497-32447497 on Build GRCh38
rs191891712 Chr.20: 32447446-32447446 on Build GRCh38
rs774612473 Chr.20: 32447403-32447404 on Build GRCh38
rs144695451 Chr.20: 32447701-32447701 on Build GRCh38
rs765543770 Chr.20: 32447738-32447738 on Build GRCh38
rs764863723 Chr.20: 32447319-32447319 on Build GRCh38
rs752899897 Chr.20: 32447746-32447746 on Build GRCh38
rs984888906 Chr.20: 32447484-32447484 on Build GRCh38
rs762177818 Chr.20: 32447743-32447766 on Build GRCh38
rs770976546 Chr.20: 32447694-32447694 on Build GRCh38
rs980864582 Chr.20: 32447595-32447595 on Build GRCh38
rs148090404 Chr.20: 32447760-32447760 on Build GRCh38
rs372314583 Chr.20: 32447639-32447639 on Build GRCh38
rs373464965 Chr.20: 32447719-32447719 on Build GRCh38
rs6057583 Chr.20: 32447366-32447366 on Build GRCh38
rs749342775 Chr.20: 32447732-32447733 on Build GRCh38
rs1015464818 Chr.20: 32447517-32447518 on Build GRCh38
rs756621752 Chr.20: 32447673-32447673 on Build GRCh38
rs757451105 Chr.20: 32447758-32447758 on Build GRCh38
rs769570581 Chr.20: 32447763-32447763 on Build GRCh38
rs1054092169 Chr.20: 32447367-32447367 on Build GRCh38
rs573906564 Chr.20: 32447358-32447358 on Build GRCh38
rs568149628 Chr.20: 32447761-32447761 on Build GRCh38
rs1006290048 Chr.20: 32447713-32447713 on Build GRCh38
rs759964502 Chr.20: 32447787-32447787 on Build GRCh38
rs766673725 Chr.20: 32447300-32447300 on Build GRCh38
rs775154968 Chr.20: 32447718-32447718 on Build GRCh38
rs756167827 Chr.20: 32447433-32447433 on Build GRCh38
rs1049259656 Chr.20: 32447448-32447448 on Build GRCh38
rs374212206 Chr.20: 32447429-32447430 on Build GRCh38
rs867512673 Chr.20: 32447403-32447403 on Build GRCh38
rs766264637 Chr.20: 32447762-32447770 on Build GRCh38
rs546385167 Chr.20: 32447691-32447691 on Build GRCh38
rs761373270 Chr.20: 32447727-32447727 on Build GRCh38
rs1010129143 Chr.20: 32447388-32447388 on Build GRCh38
rs6057582 Chr.20: 32447364-32447364 on Build GRCh38
rs759305982 Chr.20: 32447589-32447589 on Build GRCh38
rs369968679 Chr.20: 32447683-32447683 on Build GRCh38
rs768651158 Chr.20: 32447733-32447738 on Build GRCh38

View Full Product Details