Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34852063-34852565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
UniGene
Hs.433738
Gene Chromosome Location
Chr.20: 34844720-34872858 on Build GRCh38
Transcripts
NM_178026.2 XM_011528782.2 XM_011528783.2 XM_017027809.1 XM_017027810.1 XM_017027811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588852 c.1473C>T p.S491S Chr.20: 34852269-34852269 on Build GRCh38
219364 c.1421C>T p.A474V Chr.20: 34852437-34852437 on Build GRCh38
6562751 c.1436T>A p.M479K Chr.20: 34852422-34852422 on Build GRCh38
6525755 c.1407A>T p.G469G Chr.20: 34852451-34852451 on Build GRCh38
4819137 c.1361C>T p.S454F Chr.20: 34852497-34852497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891071056 Chr.20: 34852478-34852478 on Build GRCh38
rs537150694 Chr.20: 34852275-34852275 on Build GRCh38
rs986048766 Chr.20: 34852428-34852428 on Build GRCh38
rs558657735 Chr.20: 34852320-34852320 on Build GRCh38
rs563550376 Chr.20: 34852468-34852468 on Build GRCh38
rs368265165 Chr.20: 34852526-34852526 on Build GRCh38
rs778644849 Chr.20: 34852234-34852234 on Build GRCh38
rs756059919 Chr.20: 34852137-34852137 on Build GRCh38
rs748483752 Chr.20: 34852446-34852446 on Build GRCh38
rs373815703 Chr.20: 34852112-34852112 on Build GRCh38
rs764414645 Chr.20: 34852202-34852202 on Build GRCh38
rs941354599 Chr.20: 34852126-34852126 on Build GRCh38
rs752627042 Chr.20: 34852472-34852472 on Build GRCh38
rs754719912 Chr.20: 34852151-34852151 on Build GRCh38
rs772658673 Chr.20: 34852183-34852183 on Build GRCh38
rs150105441 Chr.20: 34852166-34852166 on Build GRCh38
rs139011248 Chr.20: 34852447-34852447 on Build GRCh38
rs138441861 Chr.20: 34852204-34852204 on Build GRCh38
rs991009167 Chr.20: 34852331-34852331 on Build GRCh38
rs779436496 Chr.20: 34852444-34852444 on Build GRCh38
rs776256348 Chr.20: 34852261-34852261 on Build GRCh38
rs749111962 Chr.20: 34852140-34852140 on Build GRCh38
rs866226326 Chr.20: 34852525-34852525 on Build GRCh38
rs746458249 Chr.20: 34852184-34852184 on Build GRCh38
rs764017297 Chr.20: 34852493-34852493 on Build GRCh38
rs758829060 Chr.20: 34852215-34852215 on Build GRCh38
rs764404000 Chr.20: 34852106-34852106 on Build GRCh38
rs377137760 Chr.20: 34852365-34852365 on Build GRCh38
rs1017963365 Chr.20: 34852161-34852161 on Build GRCh38
rs773376870 Chr.20: 34852304-34852304 on Build GRCh38
rs370540483 Chr.20: 34852375-34852375 on Build GRCh38
rs767765280 Chr.20: 34852214-34852214 on Build GRCh38
rs770514344 Chr.20: 34852257-34852257 on Build GRCh38
rs776096524 Chr.20: 34852186-34852186 on Build GRCh38
rs534641558 Chr.20: 34852364-34852364 on Build GRCh38
rs751303314 Chr.20: 34852408-34852408 on Build GRCh38
rs898350731 Chr.20: 34852489-34852489 on Build GRCh38
rs778406384 Chr.20: 34852233-34852233 on Build GRCh38
rs569819094 Chr.20: 34852235-34852235 on Build GRCh38
rs760956407 Chr.20: 34852453-34852453 on Build GRCh38
rs780933324 Chr.20: 34852499-34852499 on Build GRCh38
rs756149488 Chr.20: 34852222-34852222 on Build GRCh38
rs750496453 Chr.20: 34852121-34852121 on Build GRCh38
rs771222124 Chr.20: 34852312-34852312 on Build GRCh38
rs777413502 Chr.20: 34852242-34852242 on Build GRCh38
rs923079275 Chr.20: 34852084-34852084 on Build GRCh38
rs777588293 Chr.20: 34852405-34852405 on Build GRCh38
rs762005863 Chr.20: 34852209-34852209 on Build GRCh38
rs766326804 Chr.20: 34852227-34852227 on Build GRCh38
rs749933146 Chr.20: 34852503-34852503 on Build GRCh38
rs370270142 Chr.20: 34852107-34852107 on Build GRCh38
rs748731847 Chr.20: 34852536-34852536 on Build GRCh38
rs1030634700 Chr.20: 34852326-34852326 on Build GRCh38
rs756937363 Chr.20: 34852433-34852433 on Build GRCh38
rs866176443 Chr.20: 34852236-34852236 on Build GRCh38
rs878895916 Chr.20: 34852283-34852283 on Build GRCh38
rs975781895 Chr.20: 34852343-34852343 on Build GRCh38
rs773536419 Chr.20: 34852450-34852450 on Build GRCh38
rs762172271 Chr.20: 34852282-34852282 on Build GRCh38
rs766488910 Chr.20: 34852313-34852313 on Build GRCh38
rs746544534 Chr.20: 34852248-34852248 on Build GRCh38
rs774636144 Chr.20: 34852279-34852279 on Build GRCh38
rs374047068 Chr.20: 34852435-34852435 on Build GRCh38
rs946760966 Chr.20: 34852359-34852359 on Build GRCh38
rs770426550 Chr.20: 34852185-34852185 on Build GRCh38
rs953202460 Chr.20: 34852395-34852395 on Build GRCh38
rs778749978 Chr.20: 34852159-34852159 on Build GRCh38
rs755601936 Chr.20: 34852516-34852516 on Build GRCh38
rs201339481 Chr.20: 34852191-34852191 on Build GRCh38
rs201177862 Chr.20: 34852336-34852336 on Build GRCh38
rs747530699 Chr.20: 34852237-34852237 on Build GRCh38
rs202228985 Chr.20: 34852496-34852496 on Build GRCh38
rs200008306 Chr.20: 34852114-34852114 on Build GRCh38
rs201017577 Chr.20: 34852443-34852443 on Build GRCh38
rs763430219 Chr.20: 34852187-34852187 on Build GRCh38
rs759597912 Chr.20: 34852460-34852460 on Build GRCh38
rs768780046 Chr.20: 34852298-34852299 on Build GRCh38
rs1025353649 Chr.20: 34852509-34852509 on Build GRCh38
rs751393594 Chr.20: 34852495-34852495 on Build GRCh38
rs142938362 Chr.20: 34852231-34852231 on Build GRCh38
rs750508603 Chr.20: 34852218-34852218 on Build GRCh38
rs758168191 Chr.20: 34852379-34852379 on Build GRCh38
rs771754497 Chr.20: 34852168-34852168 on Build GRCh38
rs760866289 Chr.20: 34852307-34852307 on Build GRCh38
rs772215890 Chr.20: 34852292-34852292 on Build GRCh38
rs145189899 Chr.20: 34852442-34852442 on Build GRCh38
rs918569895 Chr.20: 34852466-34852466 on Build GRCh38
rs1007124242 Chr.20: 34852479-34852479 on Build GRCh38
rs769202495 Chr.20: 34852278-34852278 on Build GRCh38
rs984073016 Chr.20: 34852303-34852303 on Build GRCh38
rs200443812 Chr.20: 34852454-34852454 on Build GRCh38
rs753913102 Chr.20: 34852318-34852318 on Build GRCh38
rs774746188 Chr.20: 34852205-34852205 on Build GRCh38

View Full Product Details