Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44202874-44203383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSER1
Entrez Gene ID
51526
Gene Name
oxidative stress responsive serine rich 1
Gene Aliases
C20orf111, HSPC207, Osr1, Perit1, dJ1183I21.1
UniGene
Hs.75798
Gene Chromosome Location
Chr.20: 44195939-44217229 on Build GRCh38
Transcripts
NM_016470.7 XM_011528851.1 XM_011528853.2 XM_011528854.2 XM_017027873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667455 c.191+2T>C p.? Chr.20: 44202959-44202959 on Build GRCh38
1483656 c.191+1G>T p.? Chr.20: 44202960-44202960 on Build GRCh38
3292666 c.168T>C p.C56C Chr.20: 44202984-44202984 on Build GRCh38
1632302 c.102A>T p.G34G Chr.20: 44203050-44203050 on Build GRCh38
6735851 c.166T>C p.C56R Chr.20: 44202986-44202986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999252908 Chr.20: 44203082-44203082 on Build GRCh38
rs537824349 Chr.20: 44203280-44203280 on Build GRCh38
rs760034281 Chr.20: 44203043-44203043 on Build GRCh38
rs751859241 Chr.20: 44202920-44202922 on Build GRCh38
rs999429435 Chr.20: 44203111-44203111 on Build GRCh38
rs780655038 Chr.20: 44203036-44203037 on Build GRCh38
rs900447545 Chr.20: 44203315-44203315 on Build GRCh38
rs889497018 Chr.20: 44203052-44203052 on Build GRCh38
rs920566489 Chr.20: 44203244-44203244 on Build GRCh38
rs749996517 Chr.20: 44203099-44203099 on Build GRCh38
rs759194157 Chr.20: 44203119-44203121 on Build GRCh38
rs992992718 Chr.20: 44203159-44203159 on Build GRCh38
rs182156066 Chr.20: 44203310-44203310 on Build GRCh38
rs918866353 Chr.20: 44203285-44203285 on Build GRCh38
rs996469700 Chr.20: 44203320-44203320 on Build GRCh38
rs779641135 Chr.20: 44202962-44202962 on Build GRCh38
rs896807673 Chr.20: 44203044-44203044 on Build GRCh38
rs749800468 Chr.20: 44203042-44203043 on Build GRCh38
rs768925628 Chr.20: 44203045-44203045 on Build GRCh38
rs772625460 Chr.20: 44203048-44203048 on Build GRCh38
rs569115284 Chr.20: 44203108-44203108 on Build GRCh38
rs556820020 Chr.20: 44203340-44203340 on Build GRCh38
rs531384048 Chr.20: 44203168-44203168 on Build GRCh38
rs555859885 Chr.20: 44203168-44203169 on Build GRCh38
rs998285163 Chr.20: 44203045-44203045 on Build GRCh38
rs941564060 Chr.20: 44203245-44203245 on Build GRCh38
rs772142720 Chr.20: 44203083-44203085 on Build GRCh38
rs151077407 Chr.20: 44202963-44202963 on Build GRCh38
rs764743068 Chr.20: 44202928-44202928 on Build GRCh38
rs549045334 Chr.20: 44202988-44202988 on Build GRCh38
rs930745305 Chr.20: 44203249-44203249 on Build GRCh38
rs762778107 Chr.20: 44202976-44202976 on Build GRCh38
rs188226615 Chr.20: 44202915-44202915 on Build GRCh38
rs776489371 Chr.20: 44203103-44203104 on Build GRCh38
rs934265974 Chr.20: 44203324-44203324 on Build GRCh38
rs192662483 Chr.20: 44203169-44203169 on Build GRCh38
rs369429802 Chr.20: 44203068-44203068 on Build GRCh38
rs747890788 Chr.20: 44202986-44202986 on Build GRCh38
rs775990830 Chr.20: 44203056-44203056 on Build GRCh38
rs372129103 Chr.20: 44202926-44202926 on Build GRCh38
rs2232287 Chr.20: 44202910-44202910 on Build GRCh38
rs1006641254 Chr.20: 44203054-44203054 on Build GRCh38
rs774531388 Chr.20: 44203034-44203034 on Build GRCh38
rs199780305 Chr.20: 44202979-44202979 on Build GRCh38
rs6073368 Chr.20: 44203114-44203114 on Build GRCh38
rs575527188 Chr.20: 44203339-44203339 on Build GRCh38
rs553746454 Chr.20: 44203187-44203233 on Build GRCh38
rs3177803 Chr.20: 44202960-44202960 on Build GRCh38
rs753987773 Chr.20: 44203287-44203287 on Build GRCh38
rs4564837 Chr.20: 44203276-44203276 on Build GRCh38
rs944583789 Chr.20: 44203290-44203290 on Build GRCh38
rs555813327 Chr.20: 44203337-44203337 on Build GRCh38
rs765799812 Chr.20: 44203112-44203112 on Build GRCh38
rs766381497 Chr.20: 44202989-44202989 on Build GRCh38
rs763134182 Chr.20: 44202897-44202900 on Build GRCh38
rs751484881 Chr.20: 44203016-44203016 on Build GRCh38
rs529204339 Chr.20: 44202987-44202987 on Build GRCh38
rs750099437 Chr.20: 44202929-44202929 on Build GRCh38
rs748453959 Chr.20: 44203052-44203054 on Build GRCh38
rs114321134 Chr.20: 44202982-44202982 on Build GRCh38
rs114554184 Chr.20: 44203358-44203358 on Build GRCh38
rs764555069 Chr.20: 44203069-44203069 on Build GRCh38
rs189978694 Chr.20: 44203266-44203266 on Build GRCh38
rs536031581 Chr.20: 44203335-44203335 on Build GRCh38
rs752963054 Chr.20: 44203180-44203180 on Build GRCh38
rs770891733 Chr.20: 44203033-44203033 on Build GRCh38
rs980138471 Chr.20: 44203186-44203186 on Build GRCh38
rs759345813 Chr.20: 44203140-44203140 on Build GRCh38
rs757809843 Chr.20: 44202939-44202939 on Build GRCh38
rs751170460 Chr.20: 44203124-44203124 on Build GRCh38
rs774542814 Chr.20: 44203050-44203051 on Build GRCh38
rs967839312 Chr.20: 44203081-44203081 on Build GRCh38
rs185005566 Chr.20: 44203205-44203205 on Build GRCh38

View Full Product Details