Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62415029-62415521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP8NL
Entrez Gene ID
140893
Gene Name
RBBP8 N-terminal like
Gene Aliases
C20orf151
UniGene
Hs.180374
Gene Chromosome Location
Chr.20: 62410237-62427573 on Build GRCh38
Transcripts
NM_080833.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721056 c.708A>G p.R236R Chr.20: 62415207-62415207 on Build GRCh38
1713765 c.668C>T p.A223V Chr.20: 62415247-62415247 on Build GRCh38
7232488 c.794+7G>A p.? Chr.20: 62415114-62415114 on Build GRCh38
4648416 c.753C>T p.S251S Chr.20: 62415162-62415162 on Build GRCh38
4646023 c.725C>T p.T242M Chr.20: 62415190-62415190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145071825 Chr.20: 62415481-62415481 on Build GRCh38
rs937449034 Chr.20: 62415266-62415266 on Build GRCh38
rs374980436 Chr.20: 62415319-62415319 on Build GRCh38
rs1021942185 Chr.20: 62415391-62415391 on Build GRCh38
rs977010774 Chr.20: 62415434-62415434 on Build GRCh38
rs764012856 Chr.20: 62415252-62415252 on Build GRCh38
rs764066164 Chr.20: 62415154-62415154 on Build GRCh38
rs760499152 Chr.20: 62415251-62415251 on Build GRCh38
rs759363914 Chr.20: 62415450-62415450 on Build GRCh38
rs759285229 Chr.20: 62415137-62415137 on Build GRCh38
rs988187582 Chr.20: 62415352-62415352 on Build GRCh38
rs953210181 Chr.20: 62415378-62415378 on Build GRCh38
rs36042460 Chr.20: 62415204-62415204 on Build GRCh38
rs922817021 Chr.20: 62415436-62415436 on Build GRCh38
rs1008687038 Chr.20: 62415388-62415388 on Build GRCh38
rs942806436 Chr.20: 62415427-62415427 on Build GRCh38
rs141285142 Chr.20: 62415315-62415315 on Build GRCh38
rs543077183 Chr.20: 62415078-62415078 on Build GRCh38
rs372018613 Chr.20: 62415200-62415200 on Build GRCh38
rs770503692 Chr.20: 62415129-62415129 on Build GRCh38
rs6121577 Chr.20: 62415235-62415235 on Build GRCh38
rs766362259 Chr.20: 62415278-62415278 on Build GRCh38
rs751590926 Chr.20: 62415288-62415288 on Build GRCh38
rs35934795 Chr.20: 62415173-62415173 on Build GRCh38
rs911079 Chr.20: 62415114-62415114 on Build GRCh38
rs1044922932 Chr.20: 62415214-62415214 on Build GRCh38
rs202237856 Chr.20: 62415320-62415320 on Build GRCh38
rs757851520 Chr.20: 62415325-62415325 on Build GRCh38
rs1005619247 Chr.20: 62415308-62415308 on Build GRCh38
rs1003300497 Chr.20: 62415500-62415500 on Build GRCh38
rs946072724 Chr.20: 62415486-62415486 on Build GRCh38
rs1034916573 Chr.20: 62415501-62415501 on Build GRCh38
rs1029138852 Chr.20: 62415115-62415115 on Build GRCh38
rs911565599 Chr.20: 62415184-62415186 on Build GRCh38
rs763763369 Chr.20: 62415078-62415078 on Build GRCh38
rs767674245 Chr.20: 62415303-62415303 on Build GRCh38
rs985869521 Chr.20: 62415420-62415420 on Build GRCh38
rs961736272 Chr.20: 62415405-62415405 on Build GRCh38
rs756436131 Chr.20: 62415317-62415317 on Build GRCh38
rs376884862 Chr.20: 62415239-62415239 on Build GRCh38
rs62196317 Chr.20: 62415106-62415106 on Build GRCh38
rs369153183 Chr.20: 62415140-62415140 on Build GRCh38
rs555537037 Chr.20: 62415417-62415417 on Build GRCh38
rs773841918 Chr.20: 62415136-62415136 on Build GRCh38
rs770795292 Chr.20: 62415277-62415277 on Build GRCh38
rs956410150 Chr.20: 62415328-62415328 on Build GRCh38
rs776509098 Chr.20: 62415449-62415449 on Build GRCh38
rs762316630 Chr.20: 62415121-62415121 on Build GRCh38
rs368034371 Chr.20: 62415444-62415444 on Build GRCh38
rs940586083 Chr.20: 62415189-62415189 on Build GRCh38
rs878888099 Chr.20: 62415205-62415205 on Build GRCh38
rs767409193 Chr.20: 62415139-62415139 on Build GRCh38
rs117333532 Chr.20: 62415323-62415323 on Build GRCh38
rs551678730 Chr.20: 62415174-62415174 on Build GRCh38
rs781592550 Chr.20: 62415105-62415105 on Build GRCh38
rs768880033 Chr.20: 62415098-62415098 on Build GRCh38
rs575962044 Chr.20: 62415433-62415433 on Build GRCh38
rs373953870 Chr.20: 62415223-62415223 on Build GRCh38
rs930014062 Chr.20: 62415442-62415442 on Build GRCh38
rs911433160 Chr.20: 62415421-62415421 on Build GRCh38
rs952585037 Chr.20: 62415051-62415051 on Build GRCh38
rs888443744 Chr.20: 62415458-62415458 on Build GRCh38
rs536111897 Chr.20: 62415326-62415326 on Build GRCh38
rs1029172947 Chr.20: 62415367-62415367 on Build GRCh38
rs6121576 Chr.20: 62415112-62415112 on Build GRCh38
rs1045086669 Chr.20: 62415482-62415482 on Build GRCh38
rs571629533 Chr.20: 62415212-62415212 on Build GRCh38
rs920485802 Chr.20: 62415432-62415432 on Build GRCh38
rs550183119 Chr.20: 62415208-62415208 on Build GRCh38
rs560779290 Chr.20: 62415175-62415175 on Build GRCh38
rs1000745131 Chr.20: 62415349-62415349 on Build GRCh38
rs368889241 Chr.20: 62415245-62415245 on Build GRCh38
rs751048977 Chr.20: 62415106-62415107 on Build GRCh38
rs527948115 Chr.20: 62415190-62415190 on Build GRCh38
rs868335904 Chr.20: 62415413-62415413 on Build GRCh38
rs927670597 Chr.20: 62415147-62415147 on Build GRCh38
rs200432639 Chr.20: 62415201-62415201 on Build GRCh38
rs914944496 Chr.20: 62415184-62415184 on Build GRCh38
rs981983060 Chr.20: 62415145-62415145 on Build GRCh38
rs755062476 Chr.20: 62415295-62415295 on Build GRCh38
rs905039531 Chr.20: 62415490-62415490 on Build GRCh38

View Full Product Details