Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37266930-37267454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR3
Entrez Gene ID
10311
Gene Name
DSCR3 arrestin fold containing
Gene Aliases
DCRA, DSCRA
UniGene
Hs.369488
Gene Chromosome Location
Chr.21: 37223425-37268124 on Build GRCh38
Transcripts
NM_006052.1 XM_005260909.4 XM_005260911.4 XM_011529423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5823464 c.15G>T p.L5L Chr.21: 37267280-37267280 on Build GRCh38
4101523 c.11C>T p.A4V Chr.21: 37267284-37267284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959595554 Chr.21: 37267154-37267154 on Build GRCh38
rs1013967279 Chr.21: 37267382-37267382 on Build GRCh38
rs9976050 Chr.21: 37266993-37266993 on Build GRCh38
rs1004763067 Chr.21: 37267113-37267113 on Build GRCh38
rs936655079 Chr.21: 37267240-37267240 on Build GRCh38
rs76622052 Chr.21: 37267068-37267068 on Build GRCh38
rs372752053 Chr.21: 37267114-37267114 on Build GRCh38
rs189656605 Chr.21: 37267357-37267357 on Build GRCh38
rs200214927 Chr.21: 37267309-37267309 on Build GRCh38
rs56348240 Chr.21: 37266965-37266966 on Build GRCh38
rs746374750 Chr.21: 37267226-37267226 on Build GRCh38
rs397800001 Chr.21: 37266966-37266967 on Build GRCh38
rs780502738 Chr.21: 37267321-37267321 on Build GRCh38
rs934650655 Chr.21: 37267053-37267053 on Build GRCh38
rs1015944731 Chr.21: 37267123-37267123 on Build GRCh38
rs776778925 Chr.21: 37267258-37267258 on Build GRCh38
rs762842167 Chr.21: 37267287-37267287 on Build GRCh38
rs575762356 Chr.21: 37267166-37267166 on Build GRCh38
rs916179150 Chr.21: 37266964-37266964 on Build GRCh38
rs528542556 Chr.21: 37267082-37267082 on Build GRCh38
rs761744217 Chr.21: 37267267-37267267 on Build GRCh38
rs372683532 Chr.21: 37267306-37267306 on Build GRCh38
rs938158612 Chr.21: 37266999-37266999 on Build GRCh38
rs779503518 Chr.21: 37267219-37267219 on Build GRCh38
rs1042802980 Chr.21: 37267054-37267054 on Build GRCh38
rs1039088764 Chr.21: 37267078-37267078 on Build GRCh38
rs370620632 Chr.21: 37267400-37267400 on Build GRCh38
rs369181934 Chr.21: 37266968-37266968 on Build GRCh38
rs921063442 Chr.21: 37267225-37267225 on Build GRCh38
rs914674583 Chr.21: 37267367-37267367 on Build GRCh38
rs751261400 Chr.21: 37267194-37267194 on Build GRCh38
rs373126930 Chr.21: 37267334-37267334 on Build GRCh38
rs56994384 Chr.21: 37267213-37267213 on Build GRCh38
rs767515796 Chr.21: 37267203-37267214 on Build GRCh38
rs376885484 Chr.21: 37267238-37267238 on Build GRCh38
rs545211089 Chr.21: 37267150-37267150 on Build GRCh38
rs780776663 Chr.21: 37267209-37267209 on Build GRCh38
rs979813877 Chr.21: 37266980-37266980 on Build GRCh38
rs960078243 Chr.21: 37267361-37267361 on Build GRCh38
rs750130766 Chr.21: 37267214-37267215 on Build GRCh38
rs769897716 Chr.21: 37267332-37267332 on Build GRCh38
rs1039122837 Chr.21: 37267241-37267241 on Build GRCh38
rs369799571 Chr.21: 37267356-37267356 on Build GRCh38
rs754690709 Chr.21: 37267291-37267291 on Build GRCh38
rs945623791 Chr.21: 37267180-37267180 on Build GRCh38
rs947434078 Chr.21: 37267391-37267391 on Build GRCh38
rs755908735 Chr.21: 37267313-37267315 on Build GRCh38
rs948410748 Chr.21: 37267080-37267094 on Build GRCh38
rs963030501 Chr.21: 37267125-37267125 on Build GRCh38
rs548366586 Chr.21: 37267401-37267401 on Build GRCh38
rs978335389 Chr.21: 37267188-37267188 on Build GRCh38
rs899693434 Chr.21: 37267424-37267424 on Build GRCh38
rs536080445 Chr.21: 37267307-37267307 on Build GRCh38
rs547158340 Chr.21: 37267297-37267297 on Build GRCh38
rs375075922 Chr.21: 37266967-37266967 on Build GRCh38
rs35802298 Chr.21: 37267262-37267263 on Build GRCh38
rs766263304 Chr.21: 37267342-37267342 on Build GRCh38
rs979887426 Chr.21: 37267126-37267126 on Build GRCh38
rs865880796 Chr.21: 37267100-37267100 on Build GRCh38
rs559316845 Chr.21: 37267077-37267077 on Build GRCh38
rs776779815 Chr.21: 37267329-37267329 on Build GRCh38
rs372120868 Chr.21: 37267162-37267162 on Build GRCh38
rs537038829 Chr.21: 37267396-37267396 on Build GRCh38
rs374521794 Chr.21: 37267293-37267293 on Build GRCh38
rs762897818 Chr.21: 37267340-37267340 on Build GRCh38
rs528442495 Chr.21: 37267324-37267324 on Build GRCh38
rs373146302 Chr.21: 37267408-37267408 on Build GRCh38
rs539786230 Chr.21: 37267232-37267232 on Build GRCh38
rs970187356 Chr.21: 37267353-37267353 on Build GRCh38
rs1029569726 Chr.21: 37267155-37267155 on Build GRCh38
rs368458335 Chr.21: 37267160-37267160 on Build GRCh38
rs1045944696 Chr.21: 37266991-37266991 on Build GRCh38
rs979490728 Chr.21: 37267099-37267099 on Build GRCh38
rs747571859 Chr.21: 37267211-37267211 on Build GRCh38
rs530772715 Chr.21: 37267163-37267163 on Build GRCh38
rs1015353058 Chr.21: 37267256-37267256 on Build GRCh38
rs1043548405 Chr.21: 37267104-37267104 on Build GRCh38
rs918150439 Chr.21: 37267012-37267012 on Build GRCh38
rs773198574 Chr.21: 37267276-37267276 on Build GRCh38
rs768724850 Chr.21: 37267243-37267243 on Build GRCh38
rs371217621 Chr.21: 37267316-37267316 on Build GRCh38
rs866113570 Chr.21: 37267261-37267261 on Build GRCh38
rs981693406 Chr.21: 37267354-37267354 on Build GRCh38
rs561433791 Chr.21: 37267374-37267374 on Build GRCh38
rs554418774 Chr.21: 37267352-37267373 on Build GRCh38
rs770778764 Chr.21: 37267360-37267360 on Build GRCh38
rs899766995 Chr.21: 37267117-37267117 on Build GRCh38
rs563549175 Chr.21: 37266985-37266985 on Build GRCh38
rs75445357 Chr.21: 37267156-37267156 on Build GRCh38
rs369058750 Chr.21: 37267271-37267271 on Build GRCh38
rs766206465 Chr.21: 37267290-37267290 on Build GRCh38
rs748394701 Chr.21: 37267331-37267331 on Build GRCh38
rs997743861 Chr.21: 37267091-37267091 on Build GRCh38
rs992447283 Chr.21: 37267001-37267001 on Build GRCh38
rs886371490 Chr.21: 37267112-37267112 on Build GRCh38

View Full Product Details